Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197486772-197487273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPD1
Entrez Gene ID
3329
Gene Name
heat shock protein family D (Hsp60) member 1
Gene Aliases
CPN60, GROEL, HLD4, HSP-60, HSP60, HSP65, HuCHA60, SPG13
UniGene
Hs.595053
Gene Chromosome Location
Chr.2: 197486584-197500274 on Build GRCh38
Transcripts
NM_002156.4 NM_199440.1 NR_132788.1

Gene Symbol
SNORA105B
Entrez Gene ID
106635540
Gene Name
small nucleolar RNA, H/ACA box 105B
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.2: 197486788-197486903 on Build GRCh38
Transcripts
NM_002156.4 NM_199440.1 NR_132788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1177574 c.1708G>A p.G570S Chr.2: 197487060-197487060 on Build GRCh38
6377071 c.1685G>T p.G562V Chr.2: 197487083-197487083 on Build GRCh38
1014525 c.1646T>G p.I549S Chr.2: 197487122-197487122 on Build GRCh38
6049720 c.1688G>C p.G563A Chr.2: 197487080-197487080 on Build GRCh38
7191192 c.1621A>G p.T541A Chr.2: 197487147-197487147 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780286521 Chr.2: 197487092-197487092 on Build GRCh38
rs932315571 Chr.2: 197486912-197486912 on Build GRCh38
rs920966690 Chr.2: 197486906-197486906 on Build GRCh38
rs1002130353 Chr.2: 197487049-197487049 on Build GRCh38
rs564507442 Chr.2: 197486977-197486977 on Build GRCh38
rs749640265 Chr.2: 197487137-197487137 on Build GRCh38
rs766526223 Chr.2: 197487238-197487238 on Build GRCh38
rs779569096 Chr.2: 197487148-197487148 on Build GRCh38
rs201988193 Chr.2: 197487063-197487063 on Build GRCh38
rs939855653 Chr.2: 197486958-197486958 on Build GRCh38
rs367873403 Chr.2: 197487211-197487211 on Build GRCh38
rs767629305 Chr.2: 197487226-197487226 on Build GRCh38
rs977430026 Chr.2: 197487215-197487215 on Build GRCh38
rs553206228 Chr.2: 197487244-197487244 on Build GRCh38
rs863224878 Chr.2: 197487161-197487161 on Build GRCh38
rs368341206 Chr.2: 197486948-197486948 on Build GRCh38
rs761722635 Chr.2: 197487222-197487222 on Build GRCh38
rs755339378 Chr.2: 197487138-197487138 on Build GRCh38
rs769100059 Chr.2: 197487021-197487026 on Build GRCh38
rs767576463 Chr.2: 197487078-197487078 on Build GRCh38
rs375736101 Chr.2: 197487210-197487210 on Build GRCh38
rs888170776 Chr.2: 197486956-197486956 on Build GRCh38
rs551123918 Chr.2: 197487057-197487057 on Build GRCh38
rs760763468 Chr.2: 197487232-197487232 on Build GRCh38
rs1022880686 Chr.2: 197487028-197487028 on Build GRCh38
rs755229081 Chr.2: 197487008-197487009 on Build GRCh38
rs775116514 Chr.2: 197487047-197487047 on Build GRCh38
rs977022325 Chr.2: 197486881-197486881 on Build GRCh38
rs768371703 Chr.2: 197487218-197487218 on Build GRCh38
rs955 Chr.2: 197486851-197486851 on Build GRCh38
rs746573079 Chr.2: 197486995-197486995 on Build GRCh38
rs3860443 Chr.2: 197486849-197486849 on Build GRCh38
rs879486164 Chr.2: 197487055-197487056 on Build GRCh38
rs774337079 Chr.2: 197487221-197487221 on Build GRCh38
rs757331948 Chr.2: 197487066-197487066 on Build GRCh38
rs559668759 Chr.2: 197486796-197486796 on Build GRCh38
rs944251028 Chr.2: 197486855-197486855 on Build GRCh38
rs775983510 Chr.2: 197487162-197487162 on Build GRCh38
rs374322039 Chr.2: 197487056-197487056 on Build GRCh38
rs745574190 Chr.2: 197487177-197487177 on Build GRCh38
rs548560176 Chr.2: 197486880-197486880 on Build GRCh38
rs886055385 Chr.2: 197486990-197486990 on Build GRCh38
rs3860442 Chr.2: 197486799-197486799 on Build GRCh38
rs12392 Chr.2: 197486805-197486805 on Build GRCh38
rs201713666 Chr.2: 197487081-197487081 on Build GRCh38
rs776425415 Chr.2: 197487030-197487030 on Build GRCh38
rs535634000 Chr.2: 197487067-197487068 on Build GRCh38
rs759099027 Chr.2: 197487032-197487032 on Build GRCh38
rs1048203594 Chr.2: 197486920-197486920 on Build GRCh38
rs762688563 Chr.2: 197487211-197487214 on Build GRCh38
rs3877628 Chr.2: 197486922-197486922 on Build GRCh38
rs770624807 Chr.2: 197487156-197487156 on Build GRCh38
rs200363537 Chr.2: 197487055-197487055 on Build GRCh38
rs201030114 Chr.2: 197487060-197487060 on Build GRCh38
rs374037426 Chr.2: 197487150-197487150 on Build GRCh38
rs563146858 Chr.2: 197487217-197487217 on Build GRCh38
rs761316562 Chr.2: 197487051-197487052 on Build GRCh38
rs748662234 Chr.2: 197487154-197487154 on Build GRCh38
rs772837296 Chr.2: 197487155-197487156 on Build GRCh38
rs765128726 Chr.2: 197487036-197487036 on Build GRCh38
rs41265953 Chr.2: 197487080-197487080 on Build GRCh38
rs371969794 Chr.2: 197487087-197487087 on Build GRCh38
rs2889364 Chr.2: 197486845-197486845 on Build GRCh38
rs35288727 Chr.2: 197487203-197487203 on Build GRCh38
rs988244005 Chr.2: 197486820-197486820 on Build GRCh38
rs751438836 Chr.2: 197487065-197487065 on Build GRCh38
rs544458991 Chr.2: 197487107-197487107 on Build GRCh38
rs770559250 Chr.2: 197487011-197487011 on Build GRCh38
rs568459774 Chr.2: 197486921-197486921 on Build GRCh38
rs777031722 Chr.2: 197487027-197487028 on Build GRCh38
rs202168359 Chr.2: 197487037-197487037 on Build GRCh38
rs914158050 Chr.2: 197486829-197486829 on Build GRCh38
rs772384118 Chr.2: 197487013-197487013 on Build GRCh38

View Full Product Details