Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32900939-32901484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB1
Entrez Gene ID
3688
Gene Name
integrin subunit beta 1
Gene Aliases
CD29, FNRB, GPIIA, MDF2, MSK12, VLA-BETA, VLAB
UniGene
Hs.643813
Gene Chromosome Location
Chr.10: 32900318-32958365 on Build GRCh38
Transcripts
NM_002211.3 NM_033668.2 NM_133376.2 NR_132768.1

Gene Symbol
SNORA86
Entrez Gene ID
106633814
Gene Name
small nucleolar RNA, H/ACA box 86
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.10: 32901334-32901472 on Build GRCh38
Transcripts
NM_002211.3 NM_033668.2 NM_133376.2 NR_132768.1

SNP Detail

SNP ID Chromosome Location
rs962987006 Chr.10: 32901098-32901098 on Build GRCh38
rs202044859 Chr.10: 32901264-32901264 on Build GRCh38
rs199630493 Chr.10: 32901453-32901453 on Build GRCh38
rs201214013 Chr.10: 32900971-32900974 on Build GRCh38
rs1016703578 Chr.10: 32901087-32901087 on Build GRCh38
rs2891367 Chr.10: 32901410-32901410 on Build GRCh38
rs201358761 Chr.10: 32901335-32901335 on Build GRCh38
rs199877926 Chr.10: 32901265-32901265 on Build GRCh38
rs201465247 Chr.10: 32901404-32901404 on Build GRCh38
rs1029345672 Chr.10: 32900987-32900987 on Build GRCh38
rs919256722 Chr.10: 32901144-32901144 on Build GRCh38
rs531879881 Chr.10: 32901324-32901324 on Build GRCh38
rs992930609 Chr.10: 32901175-32901176 on Build GRCh38
rs1044624812 Chr.10: 32901233-32901233 on Build GRCh38
rs200016869 Chr.10: 32901241-32901241 on Build GRCh38
rs377677604 Chr.10: 32901405-32901407 on Build GRCh38
rs201793528 Chr.10: 32901305-32901305 on Build GRCh38
rs547790661 Chr.10: 32901448-32901448 on Build GRCh38
rs200793083 Chr.10: 32901275-32901275 on Build GRCh38
rs200458932 Chr.10: 32901425-32901425 on Build GRCh38
rs201638551 Chr.10: 32901210-32901210 on Build GRCh38
rs370409455 Chr.10: 32901381-32901381 on Build GRCh38
rs201101183 Chr.10: 32901251-32901251 on Build GRCh38
rs186026916 Chr.10: 32901313-32901313 on Build GRCh38
rs199762334 Chr.10: 32901415-32901415 on Build GRCh38
rs538178773 Chr.10: 32901402-32901404 on Build GRCh38
rs543068435 Chr.10: 32901209-32901209 on Build GRCh38
rs565103451 Chr.10: 32901391-32901391 on Build GRCh38
rs17468 Chr.10: 32901357-32901357 on Build GRCh38
rs972665782 Chr.10: 32901140-32901140 on Build GRCh38
rs529900671 Chr.10: 32901432-32901432 on Build GRCh38
rs953895468 Chr.10: 32901055-32901055 on Build GRCh38
rs950707136 Chr.10: 32901160-32901160 on Build GRCh38
rs201278521 Chr.10: 32901429-32901429 on Build GRCh38
rs997779423 Chr.10: 32901403-32901403 on Build GRCh38
rs201352037 Chr.10: 32901071-32901071 on Build GRCh38
rs191301347 Chr.10: 32901399-32901399 on Build GRCh38
rs948769000 Chr.10: 32901228-32901228 on Build GRCh38
rs889554468 Chr.10: 32901433-32901433 on Build GRCh38

View Full Product Details