Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67207090-67207635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2A
Entrez Gene ID
22992
Gene Name
lysine demethylase 2A
Gene Aliases
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
UniGene
Hs.124147
Gene Chromosome Location
Chr.11: 67119210-67258082 on Build GRCh38
Transcripts
NM_012308.2 NR_027473.1 XM_006718480.3 XM_011544860.2 XM_011544861.1 XM_017017391.1 XM_017017392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6277828 c.337G>A p.V113M Chr.11: 67207539-67207539 on Build GRCh38
4590278 c.310A>G p.S104G Chr.11: 67207512-67207512 on Build GRCh38
5567213 c.312T>G p.S104R Chr.11: 67207514-67207514 on Build GRCh38
2041333 c.324G>A p.V108V Chr.11: 67207526-67207526 on Build GRCh38
931012 c.350A>C p.K117T Chr.11: 67207552-67207552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993438213 Chr.11: 67207308-67207308 on Build GRCh38
rs773836437 Chr.11: 67207600-67207600 on Build GRCh38
rs978452549 Chr.11: 67207143-67207143 on Build GRCh38
rs751076811 Chr.11: 67207482-67207482 on Build GRCh38
rs918201099 Chr.11: 67207470-67207470 on Build GRCh38
rs185719548 Chr.11: 67207463-67207463 on Build GRCh38
rs765890930 Chr.11: 67207258-67207258 on Build GRCh38
rs770994964 Chr.11: 67207464-67207464 on Build GRCh38
rs181517217 Chr.11: 67207259-67207259 on Build GRCh38
rs552189934 Chr.11: 67207267-67207267 on Build GRCh38
rs745757617 Chr.11: 67207517-67207517 on Build GRCh38
rs918098700 Chr.11: 67207225-67207225 on Build GRCh38
rs960514885 Chr.11: 67207392-67207392 on Build GRCh38
rs999923182 Chr.11: 67207406-67207406 on Build GRCh38
rs761811784 Chr.11: 67207508-67207509 on Build GRCh38
rs751752033 Chr.11: 67207453-67207453 on Build GRCh38
rs993981624 Chr.11: 67207214-67207214 on Build GRCh38
rs1038248072 Chr.11: 67207207-67207207 on Build GRCh38
rs1037687383 Chr.11: 67207284-67207284 on Build GRCh38
rs759397670 Chr.11: 67207477-67207477 on Build GRCh38
rs1017406242 Chr.11: 67207126-67207126 on Build GRCh38
rs201296173 Chr.11: 67207490-67207490 on Build GRCh38
rs766987445 Chr.11: 67207591-67207591 on Build GRCh38
rs756541399 Chr.11: 67207503-67207503 on Build GRCh38
rs377136367 Chr.11: 67207520-67207520 on Build GRCh38
rs762506362 Chr.11: 67207479-67207479 on Build GRCh38
rs890522417 Chr.11: 67207251-67207251 on Build GRCh38
rs763650055 Chr.11: 67207480-67207480 on Build GRCh38
rs964701666 Chr.11: 67207263-67207263 on Build GRCh38
rs1011204056 Chr.11: 67207320-67207320 on Build GRCh38
rs781239345 Chr.11: 67207515-67207515 on Build GRCh38
rs761275225 Chr.11: 67207489-67207489 on Build GRCh38
rs779208233 Chr.11: 67207511-67207511 on Build GRCh38
rs758502917 Chr.11: 67207514-67207514 on Build GRCh38
rs758697579 Chr.11: 67207585-67207585 on Build GRCh38
rs776670262 Chr.11: 67207475-67207475 on Build GRCh38
rs890943355 Chr.11: 67207543-67207543 on Build GRCh38
rs1048291090 Chr.11: 67207215-67207215 on Build GRCh38
rs761365159 Chr.11: 67207583-67207583 on Build GRCh38
rs541776975 Chr.11: 67207115-67207115 on Build GRCh38
rs897793158 Chr.11: 67207291-67207291 on Build GRCh38
rs753147591 Chr.11: 67207245-67207245 on Build GRCh38
rs775131036 Chr.11: 67207525-67207525 on Build GRCh38
rs924245046 Chr.11: 67207170-67207170 on Build GRCh38
rs761073927 Chr.11: 67207608-67207608 on Build GRCh38
rs916233451 Chr.11: 67207192-67207192 on Build GRCh38
rs138807496 Chr.11: 67207359-67207359 on Build GRCh38
rs764416773 Chr.11: 67207405-67207405 on Build GRCh38
rs968439188 Chr.11: 67207142-67207142 on Build GRCh38
rs766648893 Chr.11: 67207505-67207505 on Build GRCh38
rs554189801 Chr.11: 67207544-67207544 on Build GRCh38
rs896995933 Chr.11: 67207209-67207209 on Build GRCh38
rs942124385 Chr.11: 67207521-67207521 on Build GRCh38
rs769450375 Chr.11: 67207478-67207478 on Build GRCh38
rs748267809 Chr.11: 67207512-67207512 on Build GRCh38
rs773947282 Chr.11: 67207581-67207581 on Build GRCh38
rs1022517743 Chr.11: 67207326-67207326 on Build GRCh38
rs749052730 Chr.11: 67207538-67207538 on Build GRCh38
rs527658784 Chr.11: 67207262-67207262 on Build GRCh38
rs1007828992 Chr.11: 67207345-67207345 on Build GRCh38
rs928102708 Chr.11: 67207365-67207365 on Build GRCh38
rs941314554 Chr.11: 67207195-67207195 on Build GRCh38
rs754149469 Chr.11: 67207506-67207506 on Build GRCh38
rs73501800 Chr.11: 67207331-67207331 on Build GRCh38
rs1054821561 Chr.11: 67207156-67207156 on Build GRCh38
rs747646695 Chr.11: 67207326-67207328 on Build GRCh38
rs890377333 Chr.11: 67207342-67207342 on Build GRCh38
rs939031301 Chr.11: 67207152-67207152 on Build GRCh38
rs768331087 Chr.11: 67207574-67207574 on Build GRCh38
rs755265889 Chr.11: 67207507-67207507 on Build GRCh38

View Full Product Details