Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 66986438-66986965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAST4
Entrez Gene ID
375449
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
-
UniGene
Hs.595458
Gene Chromosome Location
Chr.5: 66596348-67169595 on Build GRCh38
Transcripts
NM_001164664.1 NM_001290226.1 NM_001290228.1 NM_015183.2 NM_198828.2 XM_011543382.2 XM_011543385.2 XM_017009447.1 XM_017009448.1 XM_017009449.1 XM_017009450.1 XM_017009451.1 XM_017009452.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5547053 c.748delG p.G250fs*7 Chr.5: 66986481-66986481 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs948079462 Chr.5: 66986676-66986676 on Build GRCh38
rs781587585 Chr.5: 66986667-66986667 on Build GRCh38
rs1052525426 Chr.5: 66986737-66986737 on Build GRCh38
rs1015103514 Chr.5: 66986772-66986772 on Build GRCh38
rs1027902614 Chr.5: 66986470-66986470 on Build GRCh38
rs755293244 Chr.5: 66986665-66986665 on Build GRCh38
rs868323822 Chr.5: 66986464-66986464 on Build GRCh38
rs977238165 Chr.5: 66986591-66986591 on Build GRCh38
rs928465466 Chr.5: 66986613-66986620 on Build GRCh38
rs955213135 Chr.5: 66986622-66986622 on Build GRCh38
rs977272413 Chr.5: 66986923-66986923 on Build GRCh38
rs966203607 Chr.5: 66986520-66986520 on Build GRCh38
rs979444754 Chr.5: 66986696-66986696 on Build GRCh38
rs754740059 Chr.5: 66986473-66986473 on Build GRCh38
rs1006917967 Chr.5: 66986830-66986830 on Build GRCh38
rs371285941 Chr.5: 66986624-66986624 on Build GRCh38
rs747802621 Chr.5: 66986701-66986701 on Build GRCh38
rs989762513 Chr.5: 66986845-66986845 on Build GRCh38
rs748765002 Chr.5: 66986897-66986897 on Build GRCh38
rs372332179 Chr.5: 66986514-66986514 on Build GRCh38
rs967285656 Chr.5: 66986922-66986922 on Build GRCh38
rs58502962 Chr.5: 66986620-66986621 on Build GRCh38
rs149234910 Chr.5: 66986616-66986617 on Build GRCh38
rs553822212 Chr.5: 66986546-66986549 on Build GRCh38
rs942637615 Chr.5: 66986673-66986673 on Build GRCh38
rs541718605 Chr.5: 66986761-66986761 on Build GRCh38
rs748055189 Chr.5: 66986498-66986498 on Build GRCh38
rs777215480 Chr.5: 66986812-66986812 on Build GRCh38
rs192012897 Chr.5: 66986537-66986537 on Build GRCh38
rs962428077 Chr.5: 66986602-66986602 on Build GRCh38
rs914473225 Chr.5: 66986569-66986569 on Build GRCh38
rs993843277 Chr.5: 66986915-66986915 on Build GRCh38
rs746517867 Chr.5: 66986510-66986510 on Build GRCh38
rs200184662 Chr.5: 66986873-66986873 on Build GRCh38
rs112765509 Chr.5: 66986601-66986602 on Build GRCh38
rs866000532 Chr.5: 66986594-66986594 on Build GRCh38
rs1025358547 Chr.5: 66986916-66986916 on Build GRCh38
rs548477839 Chr.5: 66986600-66986601 on Build GRCh38
rs772116803 Chr.5: 66986501-66986501 on Build GRCh38
rs114675228 Chr.5: 66986803-66986803 on Build GRCh38
rs1023898348 Chr.5: 66986539-66986539 on Build GRCh38
rs3073601 Chr.5: 66986616-66986616 on Build GRCh38
rs57088072 Chr.5: 66986509-66986509 on Build GRCh38
rs898167354 Chr.5: 66986903-66986903 on Build GRCh38
rs539192528 Chr.5: 66986809-66986809 on Build GRCh38
rs953187423 Chr.5: 66986821-66986821 on Build GRCh38
rs59915268 Chr.5: 66986619-66986620 on Build GRCh38
rs949545185 Chr.5: 66986795-66986795 on Build GRCh38
rs1017039402 Chr.5: 66986598-66986598 on Build GRCh38
rs888040098 Chr.5: 66986722-66986722 on Build GRCh38
rs905502638 Chr.5: 66986540-66986540 on Build GRCh38
rs918056829 Chr.5: 66986745-66986745 on Build GRCh38
rs1001190206 Chr.5: 66986597-66986597 on Build GRCh38
rs750612322 Chr.5: 66986462-66986462 on Build GRCh38
rs755394806 Chr.5: 66986684-66986684 on Build GRCh38
rs986649068 Chr.5: 66986647-66986647 on Build GRCh38
rs536642871 Chr.5: 66986478-66986478 on Build GRCh38
rs796572426 Chr.5: 66986619-66986620 on Build GRCh38
rs534668535 Chr.5: 66986579-66986579 on Build GRCh38
rs887967903 Chr.5: 66986487-66986487 on Build GRCh38
rs377295387 Chr.5: 66986523-66986523 on Build GRCh38
rs775687452 Chr.5: 66986536-66986536 on Build GRCh38
rs770158088 Chr.5: 66986532-66986532 on Build GRCh38
rs776289928 Chr.5: 66986517-66986517 on Build GRCh38
rs187604346 Chr.5: 66986660-66986660 on Build GRCh38
rs769472718 Chr.5: 66986712-66986712 on Build GRCh38
rs1645250 Chr.5: 66986620-66986620 on Build GRCh38
rs945914249 Chr.5: 66986570-66986570 on Build GRCh38

View Full Product Details