Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 1792085-1792587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYT1L
Entrez Gene ID
23040
Gene Name
myelin transcription factor 1 like
Gene Aliases
MRD39, NZF1, ZC2H2C2, ZC2HC4B
UniGene
Hs.434418
Gene Chromosome Location
Chr.2: 1789113-2331388 on Build GRCh38
Transcripts
NM_001303052.1 NM_015025.3 XM_011510318.1 XM_011510319.1 XM_011510320.2 XM_011510321.2 XM_011510322.2 XM_011510323.1 XM_011510324.1 XM_011510325.1 XM_011510326.1 XM_011510327.1 XM_011510328.2 XM_017003604.1 XM_017003605.1 XM_017003606.1 XM_017003607.1 XM_017003609.1 XM_017003610.1 XM_017003611.1 XM_017003612.1 XM_017003613.1 XM_017003614.1 XM_017003615.1 XM_017003616.1 XM_017003617.1 XM_017003618.1 XM_017003619.1 XM_017003620.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4859508 c.3348C>T p.L1116L Chr.2: 1792387-1792387 on Build GRCh38
7213789 c.361A>C p.N121H Chr.2: 1792374-1792374 on Build GRCh38
5829339 c.351C>T p.H117H Chr.2: 1792384-1792384 on Build GRCh38
4949005 c.3351C>T p.H1117H Chr.2: 1792384-1792384 on Build GRCh38
4801035 c.3305A>C p.E1102A Chr.2: 1792430-1792430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761976331 Chr.2: 1792274-1792274 on Build GRCh38
rs778341828 Chr.2: 1792386-1792388 on Build GRCh38
rs548064082 Chr.2: 1792230-1792230 on Build GRCh38
rs754873838 Chr.2: 1792483-1792483 on Build GRCh38
rs1043133579 Chr.2: 1792447-1792447 on Build GRCh38
rs764364073 Chr.2: 1792461-1792461 on Build GRCh38
rs1016543178 Chr.2: 1792183-1792183 on Build GRCh38
rs994783337 Chr.2: 1792209-1792209 on Build GRCh38
rs775893555 Chr.2: 1792117-1792117 on Build GRCh38
rs968012777 Chr.2: 1792456-1792456 on Build GRCh38
rs760935170 Chr.2: 1792453-1792453 on Build GRCh38
rs1048834884 Chr.2: 1792232-1792232 on Build GRCh38
rs553936183 Chr.2: 1792479-1792479 on Build GRCh38
rs755797259 Chr.2: 1792497-1792497 on Build GRCh38
rs775734266 Chr.2: 1792444-1792444 on Build GRCh38
rs151085015 Chr.2: 1792384-1792384 on Build GRCh38
rs746396957 Chr.2: 1792416-1792416 on Build GRCh38
rs752503638 Chr.2: 1792484-1792484 on Build GRCh38
rs937513639 Chr.2: 1792522-1792522 on Build GRCh38
rs944521961 Chr.2: 1792125-1792125 on Build GRCh38
rs774876723 Chr.2: 1792414-1792414 on Build GRCh38
rs755889769 Chr.2: 1792375-1792375 on Build GRCh38
rs954575705 Chr.2: 1792308-1792308 on Build GRCh38
rs565764893 Chr.2: 1792516-1792516 on Build GRCh38
rs531865082 Chr.2: 1792194-1792194 on Build GRCh38
rs913208512 Chr.2: 1792408-1792408 on Build GRCh38
rs756792071 Chr.2: 1792508-1792508 on Build GRCh38
rs751288751 Chr.2: 1792290-1792290 on Build GRCh38
rs34259720 Chr.2: 1792140-1792141 on Build GRCh38
rs1020420815 Chr.2: 1792426-1792426 on Build GRCh38
rs190967170 Chr.2: 1792552-1792552 on Build GRCh38
rs895927119 Chr.2: 1792329-1792329 on Build GRCh38
rs971867381 Chr.2: 1792536-1792536 on Build GRCh38
rs888460697 Chr.2: 1792270-1792270 on Build GRCh38
rs549758932 Chr.2: 1792245-1792245 on Build GRCh38
rs373725845 Chr.2: 1792225-1792225 on Build GRCh38
rs778293492 Chr.2: 1792514-1792514 on Build GRCh38
rs368807335 Chr.2: 1792377-1792377 on Build GRCh38
rs761926891 Chr.2: 1792473-1792473 on Build GRCh38
rs754753972 Chr.2: 1792297-1792297 on Build GRCh38
rs753571569 Chr.2: 1792506-1792506 on Build GRCh38
rs1024570209 Chr.2: 1792123-1792123 on Build GRCh38
rs1055856127 Chr.2: 1792551-1792551 on Build GRCh38
rs1004619562 Chr.2: 1792161-1792161 on Build GRCh38
rs771542746 Chr.2: 1792389-1792389 on Build GRCh38
rs911626214 Chr.2: 1792523-1792523 on Build GRCh38
rs776656662 Chr.2: 1792471-1792471 on Build GRCh38
rs921846569 Chr.2: 1792284-1792284 on Build GRCh38
rs780878610 Chr.2: 1792327-1792327 on Build GRCh38
rs747768727 Chr.2: 1792367-1792367 on Build GRCh38
rs764099993 Chr.2: 1792272-1792272 on Build GRCh38
rs772416903 Chr.2: 1792423-1792423 on Build GRCh38
rs530061364 Chr.2: 1792207-1792207 on Build GRCh38
rs950540688 Chr.2: 1792124-1792124 on Build GRCh38
rs754253029 Chr.2: 1792546-1792546 on Build GRCh38
rs774537123 Chr.2: 1792254-1792254 on Build GRCh38
rs758323992 Chr.2: 1792275-1792275 on Build GRCh38
rs777618852 Chr.2: 1792503-1792503 on Build GRCh38
rs944468324 Chr.2: 1792528-1792528 on Build GRCh38
rs115972918 Chr.2: 1792276-1792276 on Build GRCh38

View Full Product Details