Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42762680-42763187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXER1
Entrez Gene ID
165140
Gene Name
oxoeicosanoid receptor 1
Gene Aliases
GPCR, GPR170, TG1019
UniGene
Hs.168439
Gene Chromosome Location
Chr.2: 42762499-42764261 on Build GRCh38
Transcripts
NM_148962.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6258901 c.1267G>T p.G423C Chr.2: 42762913-42762913 on Build GRCh38
331996 c.993C>A p.D331E Chr.2: 42763187-42763187 on Build GRCh38
331995 c.995T>C p.L332P Chr.2: 42763185-42763185 on Build GRCh38
6091517 c.1058T>A p.V353E Chr.2: 42763122-42763122 on Build GRCh38
6542537 c.1255G>A p.G419S Chr.2: 42762925-42762925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781062404 Chr.2: 42762852-42762852 on Build GRCh38
rs111706530 Chr.2: 42762987-42762987 on Build GRCh38
rs1046828651 Chr.2: 42762710-42762710 on Build GRCh38
rs370202578 Chr.2: 42762998-42762998 on Build GRCh38
rs143944527 Chr.2: 42763085-42763085 on Build GRCh38
rs369004247 Chr.2: 42763109-42763109 on Build GRCh38
rs777119606 Chr.2: 42762946-42762946 on Build GRCh38
rs994250035 Chr.2: 42762757-42762757 on Build GRCh38
rs750743290 Chr.2: 42762927-42762927 on Build GRCh38
rs1037993608 Chr.2: 42762866-42762866 on Build GRCh38
rs754291238 Chr.2: 42762936-42762936 on Build GRCh38
rs373771713 Chr.2: 42762978-42762978 on Build GRCh38
rs574380144 Chr.2: 42762995-42762995 on Build GRCh38
rs145274773 Chr.2: 42763002-42763002 on Build GRCh38
rs770749658 Chr.2: 42762873-42762873 on Build GRCh38
rs377351782 Chr.2: 42763081-42763081 on Build GRCh38
rs767745953 Chr.2: 42762924-42762924 on Build GRCh38
rs1019416411 Chr.2: 42762761-42762761 on Build GRCh38
rs765905637 Chr.2: 42763161-42763161 on Build GRCh38
rs775294834 Chr.2: 42762886-42762886 on Build GRCh38
rs760234997 Chr.2: 42763091-42763091 on Build GRCh38
rs532254056 Chr.2: 42763033-42763033 on Build GRCh38
rs775947502 Chr.2: 42763135-42763135 on Build GRCh38
rs538553281 Chr.2: 42762879-42762879 on Build GRCh38
rs764075054 Chr.2: 42762908-42762908 on Build GRCh38
rs995127862 Chr.2: 42762703-42762703 on Build GRCh38
rs556344195 Chr.2: 42763093-42763093 on Build GRCh38
rs754272344 Chr.2: 42762996-42762996 on Build GRCh38
rs1007944333 Chr.2: 42762923-42762923 on Build GRCh38
rs766652626 Chr.2: 42762993-42762993 on Build GRCh38
rs182457573 Chr.2: 42763124-42763124 on Build GRCh38
rs921040803 Chr.2: 42763142-42763142 on Build GRCh38
rs2278586 Chr.2: 42762961-42762961 on Build GRCh38
rs148146219 Chr.2: 42762992-42762992 on Build GRCh38
rs746099064 Chr.2: 42763127-42763127 on Build GRCh38
rs529593797 Chr.2: 42762804-42762804 on Build GRCh38
rs560232632 Chr.2: 42762831-42762831 on Build GRCh38
rs764718309 Chr.2: 42763090-42763090 on Build GRCh38
rs568811676 Chr.2: 42763061-42763061 on Build GRCh38
rs760394888 Chr.2: 42762828-42762828 on Build GRCh38
rs750095060 Chr.2: 42763007-42763007 on Build GRCh38
rs749698585 Chr.2: 42763141-42763141 on Build GRCh38
rs903055757 Chr.2: 42763154-42763154 on Build GRCh38
rs745681118 Chr.2: 42762881-42762881 on Build GRCh38
rs12617777 Chr.2: 42762870-42762870 on Build GRCh38
rs768543138 Chr.2: 42762920-42762920 on Build GRCh38
rs767104653 Chr.2: 42763059-42763059 on Build GRCh38
rs75802518 Chr.2: 42763066-42763066 on Build GRCh38
rs370218708 Chr.2: 42763088-42763088 on Build GRCh38
rs111369512 Chr.2: 42762801-42762801 on Build GRCh38
rs747117114 Chr.2: 42763079-42763079 on Build GRCh38
rs145580414 Chr.2: 42763034-42763034 on Build GRCh38
rs113066186 Chr.2: 42762885-42762885 on Build GRCh38
rs769216048 Chr.2: 42763145-42763145 on Build GRCh38
rs145145793 Chr.2: 42763062-42763062 on Build GRCh38
rs755579802 Chr.2: 42762944-42762944 on Build GRCh38
rs772121715 Chr.2: 42763031-42763031 on Build GRCh38
rs746787844 Chr.2: 42762864-42762865 on Build GRCh38
rs776049075 Chr.2: 42763165-42763165 on Build GRCh38
rs368188901 Chr.2: 42762864-42762864 on Build GRCh38
rs778911259 Chr.2: 42763028-42763028 on Build GRCh38
rs922593198 Chr.2: 42762874-42762874 on Build GRCh38
rs370591048 Chr.2: 42763053-42763053 on Build GRCh38
rs147970991 Chr.2: 42763036-42763036 on Build GRCh38
rs143925997 Chr.2: 42762969-42762969 on Build GRCh38
rs996735948 Chr.2: 42762785-42762785 on Build GRCh38
rs747986811 Chr.2: 42763016-42763017 on Build GRCh38
rs564918578 Chr.2: 42762745-42762745 on Build GRCh38
rs370198807 Chr.2: 42762997-42762997 on Build GRCh38
rs775010531 Chr.2: 42763151-42763151 on Build GRCh38
rs544076125 Chr.2: 42762791-42762791 on Build GRCh38
rs776608682 Chr.2: 42762988-42762999 on Build GRCh38
rs748078243 Chr.2: 42763074-42763074 on Build GRCh38
rs758285274 Chr.2: 42763103-42763103 on Build GRCh38
rs752541591 Chr.2: 42763101-42763101 on Build GRCh38
rs906951321 Chr.2: 42762780-42762780 on Build GRCh38
rs142162926 Chr.2: 42762983-42762983 on Build GRCh38
rs200032038 Chr.2: 42763060-42763060 on Build GRCh38
rs770189297 Chr.2: 42763130-42763130 on Build GRCh38
rs768685734 Chr.2: 42762977-42762977 on Build GRCh38
rs145238995 Chr.2: 42762773-42762773 on Build GRCh38
rs577065981 Chr.2: 42762863-42762863 on Build GRCh38
rs746640098 Chr.2: 42762872-42762872 on Build GRCh38
rs2278585 Chr.2: 42762854-42762854 on Build GRCh38
rs539531390 Chr.2: 42762966-42762966 on Build GRCh38
rs369046833 Chr.2: 42762970-42762970 on Build GRCh38
rs575064937 Chr.2: 42762973-42762973 on Build GRCh38
rs371721042 Chr.2: 42762935-42762935 on Build GRCh38
rs985442697 Chr.2: 42763047-42763047 on Build GRCh38
rs756470893 Chr.2: 42762934-42762934 on Build GRCh38
rs145926558 Chr.2: 42763027-42763027 on Build GRCh38
rs554325300 Chr.2: 42763123-42763123 on Build GRCh38
rs914008461 Chr.2: 42762833-42762833 on Build GRCh38
rs761998660 Chr.2: 42762921-42762921 on Build GRCh38
rs200272835 Chr.2: 42762889-42762889 on Build GRCh38
rs759572081 Chr.2: 42763029-42763029 on Build GRCh38
rs747624085 Chr.2: 42762865-42762865 on Build GRCh38
rs199600588 Chr.2: 42763049-42763049 on Build GRCh38
rs547799865 Chr.2: 42763067-42763067 on Build GRCh38
rs1053594595 Chr.2: 42763058-42763058 on Build GRCh38
rs774527594 Chr.2: 42762916-42762916 on Build GRCh38
rs1001605615 Chr.2: 42762986-42762986 on Build GRCh38
rs374950829 Chr.2: 42763098-42763098 on Build GRCh38
rs779690889 Chr.2: 42763018-42763018 on Build GRCh38
rs763004929 Chr.2: 42762891-42762891 on Build GRCh38
rs563106071 Chr.2: 42763037-42763037 on Build GRCh38
rs201387408 Chr.2: 42763044-42763044 on Build GRCh38
rs1001260527 Chr.2: 42762794-42762794 on Build GRCh38
rs546182661 Chr.2: 42762807-42762807 on Build GRCh38
rs762281525 Chr.2: 42763153-42763153 on Build GRCh38
rs771020476 Chr.2: 42763080-42763080 on Build GRCh38
rs755869412 Chr.2: 42763016-42763016 on Build GRCh38
rs76606481 Chr.2: 42763077-42763077 on Build GRCh38
rs115559602 Chr.2: 42762728-42762728 on Build GRCh38
rs374132247 Chr.2: 42763003-42763003 on Build GRCh38
rs375310268 Chr.2: 42763054-42763054 on Build GRCh38
rs897396531 Chr.2: 42762747-42762747 on Build GRCh38
rs750833314 Chr.2: 42762862-42762862 on Build GRCh38
rs79735204 Chr.2: 42762947-42762947 on Build GRCh38
rs149479883 Chr.2: 42762948-42762948 on Build GRCh38
rs374714113 Chr.2: 42762959-42762959 on Build GRCh38
rs144634528 Chr.2: 42763097-42763097 on Build GRCh38
rs377669097 Chr.2: 42763021-42763021 on Build GRCh38
rs200218777 Chr.2: 42763082-42763082 on Build GRCh38

View Full Product Details