Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 128135422-128135921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A2
Entrez Gene ID
6558
Gene Name
solute carrier family 12 member 2
Gene Aliases
BSC, BSC2, NKCC1, PPP1R141
UniGene
Hs.162585
Gene Chromosome Location
Chr.5: 128083791-128189688 on Build GRCh38
Transcripts
NM_001046.2 NM_001256461.1 NR_046207.1 XM_011543588.2 XM_017009771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
297968 c.1347C>T p.F449F Chr.5: 128135747-128135747 on Build GRCh38
3135824 c.1392_1393insT p.G467fs*3 Chr.5: 128135792-128135793 on Build GRCh38
3135823 c.1348G>A p.V450I Chr.5: 128135748-128135748 on Build GRCh38
7264649 c.1327_1329delCTT p.L444delL Chr.5: 128135727-128135729 on Build GRCh38
4525811 c.1355G>T p.G452V Chr.5: 128135755-128135755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144533824 Chr.5: 128135722-128135722 on Build GRCh38
rs372179434 Chr.5: 128135789-128135789 on Build GRCh38
rs749800306 Chr.5: 128135736-128135736 on Build GRCh38
rs999371919 Chr.5: 128135824-128135824 on Build GRCh38
rs148897238 Chr.5: 128135786-128135786 on Build GRCh38
rs554821539 Chr.5: 128135748-128135748 on Build GRCh38
rs976755019 Chr.5: 128135664-128135664 on Build GRCh38
rs986494256 Chr.5: 128135543-128135543 on Build GRCh38
rs1017938659 Chr.5: 128135888-128135888 on Build GRCh38
rs372943536 Chr.5: 128135718-128135718 on Build GRCh38
rs896362413 Chr.5: 128135853-128135853 on Build GRCh38
rs1002511711 Chr.5: 128135461-128135461 on Build GRCh38
rs753598198 Chr.5: 128135492-128135493 on Build GRCh38
rs780482894 Chr.5: 128135724-128135724 on Build GRCh38
rs756405226 Chr.5: 128135827-128135827 on Build GRCh38
rs369857269 Chr.5: 128135485-128135485 on Build GRCh38
rs781382457 Chr.5: 128135717-128135717 on Build GRCh38
rs758121421 Chr.5: 128135711-128135711 on Build GRCh38
rs777591827 Chr.5: 128135617-128135617 on Build GRCh38
rs973846233 Chr.5: 128135604-128135604 on Build GRCh38
rs894583586 Chr.5: 128135468-128135468 on Build GRCh38
rs375344286 Chr.5: 128135483-128135483 on Build GRCh38
rs752246285 Chr.5: 128135703-128135703 on Build GRCh38
rs778885215 Chr.5: 128135552-128135553 on Build GRCh38
rs147843214 Chr.5: 128135743-128135743 on Build GRCh38
rs759572983 Chr.5: 128135798-128135798 on Build GRCh38
rs942407514 Chr.5: 128135568-128135568 on Build GRCh38
rs760485260 Chr.5: 128135784-128135784 on Build GRCh38
rs955334486 Chr.5: 128135851-128135851 on Build GRCh38
rs972032829 Chr.5: 128135522-128135522 on Build GRCh38
rs774396024 Chr.5: 128135659-128135659 on Build GRCh38
rs767193375 Chr.5: 128135676-128135676 on Build GRCh38
rs760683207 Chr.5: 128135683-128135683 on Build GRCh38
rs574715299 Chr.5: 128135871-128135871 on Build GRCh38
rs542019784 Chr.5: 128135613-128135613 on Build GRCh38
rs1011642256 Chr.5: 128135474-128135474 on Build GRCh38
rs1002013567 Chr.5: 128135820-128135820 on Build GRCh38
rs987145128 Chr.5: 128135877-128135877 on Build GRCh38
rs1021705822 Chr.5: 128135761-128135761 on Build GRCh38
rs1011611143 Chr.5: 128135752-128135752 on Build GRCh38
rs568900988 Chr.5: 128135567-128135567 on Build GRCh38
rs377735249 Chr.5: 128135747-128135747 on Build GRCh38
rs746870677 Chr.5: 128135641-128135641 on Build GRCh38
rs755693918 Chr.5: 128135872-128135872 on Build GRCh38
rs374869158 Chr.5: 128135860-128135860 on Build GRCh38
rs767218303 Chr.5: 128135631-128135631 on Build GRCh38
rs913687418 Chr.5: 128135755-128135755 on Build GRCh38
rs371376296 Chr.5: 128135482-128135482 on Build GRCh38
rs778853382 Chr.5: 128135852-128135852 on Build GRCh38
rs984998547 Chr.5: 128135628-128135628 on Build GRCh38
rs369058298 Chr.5: 128135697-128135697 on Build GRCh38
rs753836840 Chr.5: 128135685-128135685 on Build GRCh38
rs952103090 Chr.5: 128135579-128135579 on Build GRCh38
rs558955818 Chr.5: 128135517-128135517 on Build GRCh38
rs369460358 Chr.5: 128135684-128135684 on Build GRCh38
rs762117057 Chr.5: 128135667-128135667 on Build GRCh38
rs905147015 Chr.5: 128135454-128135454 on Build GRCh38
rs750271025 Chr.5: 128135679-128135679 on Build GRCh38
rs566914673 Chr.5: 128135464-128135464 on Build GRCh38
rs777542577 Chr.5: 128135815-128135815 on Build GRCh38
rs755547281 Chr.5: 128135843-128135843 on Build GRCh38
rs749773320 Chr.5: 128135840-128135840 on Build GRCh38
rs752295596 Chr.5: 128135805-128135805 on Build GRCh38
rs757983842 Chr.5: 128135814-128135814 on Build GRCh38
rs780417796 Chr.5: 128135830-128135830 on Build GRCh38
rs770844015 Chr.5: 128135788-128135788 on Build GRCh38
rs938659395 Chr.5: 128135721-128135721 on Build GRCh38
rs965020779 Chr.5: 128135645-128135645 on Build GRCh38
rs773503469 Chr.5: 128135860-128135862 on Build GRCh38
rs372961515 Chr.5: 128135686-128135686 on Build GRCh38
rs768736307 Chr.5: 128135654-128135654 on Build GRCh38
rs567860471 Chr.5: 128135492-128135492 on Build GRCh38
rs751226380 Chr.5: 128135825-128135825 on Build GRCh38
rs772866879 Chr.5: 128135767-128135767 on Build GRCh38
rs1022073719 Chr.5: 128135485-128135485 on Build GRCh38
rs6875842 Chr.5: 128135462-128135462 on Build GRCh38
rs1026400794 Chr.5: 128135547-128135547 on Build GRCh38
rs963741210 Chr.5: 128135891-128135891 on Build GRCh38

View Full Product Details