Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32043759-32044267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNXB
Entrez Gene ID
7148
Gene Name
tenascin XB
Gene Aliases
EDS3, HXBL, TENX, TN-X, TNX, TNXB1, TNXB2, TNXBS, VUR8, XB, XBS
UniGene
Hs.485104
Gene Chromosome Location
Chr.6: 32041155-32109374 on Build GRCh38
Transcripts
NM_019105.6 NM_032470.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7195812 c.11712C>T p.G3904G Chr.6: 32043762-32043762 on Build GRCh38
4899835 c.11644C>T p.P3882S Chr.6: 32043830-32043830 on Build GRCh38
4899834 c.736C>T p.P246S Chr.6: 32043830-32043830 on Build GRCh38
3347712 c.11458C>A p.R3820S Chr.6: 32043821-32043821 on Build GRCh38
3861953 c.707C>G p.A236G Chr.6: 32043859-32043859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774638630 Chr.6: 32043976-32043976 on Build GRCh38
rs765212228 Chr.6: 32044061-32044061 on Build GRCh38
rs2894233 Chr.6: 32043901-32043901 on Build GRCh38
rs769074772 Chr.6: 32044044-32044044 on Build GRCh38
rs930155266 Chr.6: 32044060-32044060 on Build GRCh38
rs533020881 Chr.6: 32044217-32044217 on Build GRCh38
rs549209308 Chr.6: 32043902-32043902 on Build GRCh38
rs781668158 Chr.6: 32043960-32043960 on Build GRCh38
rs567422768 Chr.6: 32043911-32043911 on Build GRCh38
rs747604607 Chr.6: 32043837-32043837 on Build GRCh38
rs538684999 Chr.6: 32043993-32043993 on Build GRCh38
rs542870641 Chr.6: 32044048-32044048 on Build GRCh38
rs756087047 Chr.6: 32043900-32043900 on Build GRCh38
rs535029125 Chr.6: 32043961-32043961 on Build GRCh38
rs971252129 Chr.6: 32043880-32043880 on Build GRCh38
rs397302 Chr.6: 32044184-32044184 on Build GRCh38
rs2894232 Chr.6: 32043867-32043867 on Build GRCh38
rs749933982 Chr.6: 32044117-32044117 on Build GRCh38
rs763132768 Chr.6: 32043965-32043965 on Build GRCh38
rs752319931 Chr.6: 32043942-32043942 on Build GRCh38
rs754085722 Chr.6: 32043817-32043817 on Build GRCh38
rs562475886 Chr.6: 32044155-32044155 on Build GRCh38
rs770131586 Chr.6: 32043928-32043928 on Build GRCh38
rs753758829 Chr.6: 32043939-32043939 on Build GRCh38
rs761027837 Chr.6: 32043812-32043812 on Build GRCh38
rs371882323 Chr.6: 32043825-32043825 on Build GRCh38
rs954367012 Chr.6: 32043833-32043853 on Build GRCh38
rs765702815 Chr.6: 32043841-32043841 on Build GRCh38
rs201948559 Chr.6: 32044167-32044167 on Build GRCh38
rs775589079 Chr.6: 32043929-32043929 on Build GRCh38
rs149197999 Chr.6: 32043857-32043857 on Build GRCh38
rs138558351 Chr.6: 32043821-32043821 on Build GRCh38
rs777041397 Chr.6: 32043923-32043923 on Build GRCh38
rs775891374 Chr.6: 32043964-32043964 on Build GRCh38
rs183465462 Chr.6: 32043819-32043819 on Build GRCh38
rs768984974 Chr.6: 32043972-32043972 on Build GRCh38
rs767061316 Chr.6: 32044114-32044114 on Build GRCh38
rs762430096 Chr.6: 32043805-32043805 on Build GRCh38
rs759816596 Chr.6: 32043927-32043927 on Build GRCh38
rs776953580 Chr.6: 32043860-32043860 on Build GRCh38
rs753737731 Chr.6: 32043984-32043984 on Build GRCh38
rs773691198 Chr.6: 32043809-32043809 on Build GRCh38
rs9378120 Chr.6: 32044223-32044223 on Build GRCh38
rs780632328 Chr.6: 32044020-32044020 on Build GRCh38
rs761985795 Chr.6: 32043981-32043981 on Build GRCh38
rs889575324 Chr.6: 32043793-32043793 on Build GRCh38
rs1024022512 Chr.6: 32044156-32044156 on Build GRCh38
rs550355338 Chr.6: 32043832-32043832 on Build GRCh38
rs756621012 Chr.6: 32044025-32044025 on Build GRCh38
rs760750881 Chr.6: 32043936-32043936 on Build GRCh38
rs773701143 Chr.6: 32043798-32043798 on Build GRCh38
rs750439153 Chr.6: 32044198-32044198 on Build GRCh38
rs780765306 Chr.6: 32044056-32044057 on Build GRCh38
rs763429682 Chr.6: 32043931-32043931 on Build GRCh38
rs768313092 Chr.6: 32043791-32043791 on Build GRCh38
rs746394659 Chr.6: 32044024-32044024 on Build GRCh38
rs148379439 Chr.6: 32043941-32043941 on Build GRCh38
rs768169058 Chr.6: 32043829-32043829 on Build GRCh38
rs561147357 Chr.6: 32044074-32044074 on Build GRCh38
rs756774851 Chr.6: 32044014-32044014 on Build GRCh38
rs897296884 Chr.6: 32044003-32044003 on Build GRCh38
rs572418615 Chr.6: 32044035-32044035 on Build GRCh38
rs758822132 Chr.6: 32043959-32043960 on Build GRCh38
rs376486710 Chr.6: 32043856-32043856 on Build GRCh38
rs765400545 Chr.6: 32043820-32043820 on Build GRCh38
rs1008389232 Chr.6: 32043799-32043799 on Build GRCh38
rs774724727 Chr.6: 32044045-32044045 on Build GRCh38
rs1040815807 Chr.6: 32043999-32043999 on Build GRCh38
rs943748943 Chr.6: 32043994-32043994 on Build GRCh38
rs369016292 Chr.6: 32043977-32043977 on Build GRCh38
rs554107135 Chr.6: 32044007-32044007 on Build GRCh38
rs771496120 Chr.6: 32043959-32043959 on Build GRCh38
rs761117807 Chr.6: 32043858-32043858 on Build GRCh38
rs758041540 Chr.6: 32043956-32043956 on Build GRCh38
rs77466839 Chr.6: 32044149-32044149 on Build GRCh38
rs2856450 Chr.6: 32044177-32044177 on Build GRCh38
rs751667690 Chr.6: 32043932-32043932 on Build GRCh38
rs371670513 Chr.6: 32043910-32043910 on Build GRCh38
rs747774925 Chr.6: 32043905-32043905 on Build GRCh38
rs566797504 Chr.6: 32043870-32043870 on Build GRCh38
rs766668761 Chr.6: 32043813-32043813 on Build GRCh38
rs745733950 Chr.6: 32043823-32043823 on Build GRCh38
rs142610250 Chr.6: 32043824-32043824 on Build GRCh38
rs756864717 Chr.6: 32044108-32044108 on Build GRCh38
rs756645662 Chr.6: 32044120-32044120 on Build GRCh38
rs397948 Chr.6: 32043937-32043937 on Build GRCh38
rs758046999 Chr.6: 32043995-32043995 on Build GRCh38
rs576272360 Chr.6: 32044075-32044075 on Build GRCh38
rs759475078 Chr.6: 32043989-32043989 on Build GRCh38
rs748899355 Chr.6: 32043785-32043785 on Build GRCh38
rs751236645 Chr.6: 32044087-32044087 on Build GRCh38
rs578036611 Chr.6: 32043992-32043992 on Build GRCh38
rs751146752 Chr.6: 32044011-32044011 on Build GRCh38
rs550641852 Chr.6: 32043806-32043806 on Build GRCh38
rs373894152 Chr.6: 32044143-32044143 on Build GRCh38
rs773307742 Chr.6: 32044049-32044049 on Build GRCh38
rs767828367 Chr.6: 32044173-32044173 on Build GRCh38
rs750416041 Chr.6: 32043894-32043894 on Build GRCh38
rs771408261 Chr.6: 32043922-32043922 on Build GRCh38
rs746510955 Chr.6: 32043958-32043958 on Build GRCh38
rs749658381 Chr.6: 32044040-32044040 on Build GRCh38
rs767894911 Chr.6: 32043807-32043807 on Build GRCh38
rs750082283 Chr.6: 32044173-32044173 on Build GRCh38
rs771500868 Chr.6: 32043907-32043907 on Build GRCh38
rs375776922 Chr.6: 32043921-32043921 on Build GRCh38
rs759445043 Chr.6: 32044055-32044055 on Build GRCh38
rs775218605 Chr.6: 32044070-32044070 on Build GRCh38

View Full Product Details