Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 47366916-47366423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF488
Entrez Gene ID
118738
Gene Name
zinc finger protein 488
Gene Aliases
-
UniGene
Hs.27788
Gene Chromosome Location
Chr.10: 47365482-47384339 on Build GRCh38
Transcripts
NM_153034.2 XM_006717617.3 XM_011539244.2 XM_017015642.1 XM_017015643.1

View Full Product Details