Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75401624-75402124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF503
Entrez Gene ID
84858
Gene Name
zinc finger protein 503
Gene Aliases
NOLZ-1, NOLZ1, Nlz2
UniGene
Hs.195710
Gene Chromosome Location
Chr.10: 75279726-75401894 on Build GRCh38
Transcripts
NM_032772.5 NR_120651.1 NR_024421.1

Gene Symbol
ZNF503-AS2
Entrez Gene ID
100131213
Gene Name
ZNF503 antisense RNA 2
Gene Aliases
C10orf41, NCRNA00245
UniGene
Hs.534598
Gene Chromosome Location
Chr.10: 75401528-75408982 on Build GRCh38
Transcripts
NM_032772.5 NR_120651.1 NR_024421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6861885 c.624C>A p.G208G Chr.10: 75401624-75401624 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs945570037 Chr.10: 75401804-75401804 on Build GRCh38
rs759155569 Chr.10: 75401757-75401757 on Build GRCh38
rs1026085982 Chr.10: 75402057-75402057 on Build GRCh38
rs1034786627 Chr.10: 75401755-75401755 on Build GRCh38
rs905001015 Chr.10: 75401857-75401857 on Build GRCh38
rs755325857 Chr.10: 75401733-75401733 on Build GRCh38
rs1017563932 Chr.10: 75401989-75401989 on Build GRCh38
rs1048090300 Chr.10: 75401963-75401963 on Build GRCh38
rs546695592 Chr.10: 75401768-75401768 on Build GRCh38
rs890346602 Chr.10: 75401968-75401968 on Build GRCh38
rs544101706 Chr.10: 75401671-75401671 on Build GRCh38
rs563598299 Chr.10: 75401654-75401654 on Build GRCh38
rs868148344 Chr.10: 75401802-75401802 on Build GRCh38
rs369229080 Chr.10: 75401882-75401882 on Build GRCh38
rs896231808 Chr.10: 75402014-75402014 on Build GRCh38
rs892384143 Chr.10: 75401711-75401711 on Build GRCh38
rs528211436 Chr.10: 75401765-75401765 on Build GRCh38
rs7085324 Chr.10: 75401901-75401901 on Build GRCh38
rs934111681 Chr.10: 75401918-75401918 on Build GRCh38
rs1039687649 Chr.10: 75401664-75401664 on Build GRCh38
rs1056916068 Chr.10: 75401686-75401686 on Build GRCh38
rs1026266543 Chr.10: 75401890-75401890 on Build GRCh38
rs1043549702 Chr.10: 75401852-75401852 on Build GRCh38
rs984753735 Chr.10: 75401897-75401897 on Build GRCh38
rs1017564097 Chr.10: 75402097-75402097 on Build GRCh38
rs951758726 Chr.10: 75401792-75401792 on Build GRCh38
rs951641678 Chr.10: 75401896-75401896 on Build GRCh38
rs938334036 Chr.10: 75401937-75401937 on Build GRCh38
rs376626822 Chr.10: 75401919-75401919 on Build GRCh38
rs964858585 Chr.10: 75401782-75401783 on Build GRCh38
rs556839255 Chr.10: 75402079-75402079 on Build GRCh38
rs894296640 Chr.10: 75401691-75401691 on Build GRCh38
rs1040161903 Chr.10: 75401812-75401812 on Build GRCh38
rs774766435 Chr.10: 75401791-75401791 on Build GRCh38
rs954874600 Chr.10: 75401893-75401901 on Build GRCh38
rs960335038 Chr.10: 75401760-75401760 on Build GRCh38
rs12780400 Chr.10: 75401709-75401709 on Build GRCh38
rs928261402 Chr.10: 75401934-75401934 on Build GRCh38
rs111533383 Chr.10: 75401718-75401718 on Build GRCh38
rs754156773 Chr.10: 75401928-75401928 on Build GRCh38
rs933953374 Chr.10: 75401821-75401821 on Build GRCh38
rs1057265728 Chr.10: 75401841-75401841 on Build GRCh38
rs560022052 Chr.10: 75401775-75401775 on Build GRCh38
rs560688833 Chr.10: 75401754-75401754 on Build GRCh38
rs1041379168 Chr.10: 75401656-75401656 on Build GRCh38
rs567533792 Chr.10: 75401910-75401910 on Build GRCh38
rs1002408731 Chr.10: 75401972-75401972 on Build GRCh38
rs1026695948 Chr.10: 75401744-75401744 on Build GRCh38
rs910063116 Chr.10: 75401800-75401800 on Build GRCh38
rs548424892 Chr.10: 75402038-75402038 on Build GRCh38
rs964765094 Chr.10: 75401899-75401901 on Build GRCh38
rs922696594 Chr.10: 75401815-75401815 on Build GRCh38
rs1003001504 Chr.10: 75401683-75401683 on Build GRCh38
rs901156171 Chr.10: 75401670-75401670 on Build GRCh38
rs80057415 Chr.10: 75401953-75401953 on Build GRCh38
rs946757523 Chr.10: 75401948-75401948 on Build GRCh38
rs555023176 Chr.10: 75401756-75401756 on Build GRCh38
rs998151625 Chr.10: 75401678-75401678 on Build GRCh38
rs541008614 Chr.10: 75401882-75401883 on Build GRCh38
rs1022426407 Chr.10: 75402054-75402054 on Build GRCh38
rs190375594 Chr.10: 75401982-75401982 on Build GRCh38
rs993268614 Chr.10: 75402041-75402041 on Build GRCh38
rs1011270446 Chr.10: 75401850-75401850 on Build GRCh38
rs993342804 Chr.10: 75401883-75401885 on Build GRCh38
rs915429430 Chr.10: 75401803-75401803 on Build GRCh38
rs969153591 Chr.10: 75401739-75401739 on Build GRCh38
rs991482077 Chr.10: 75402099-75402099 on Build GRCh38
rs964365019 Chr.10: 75402104-75402104 on Build GRCh38
rs1042669397 Chr.10: 75401805-75401805 on Build GRCh38
rs367698599 Chr.10: 75401721-75401722 on Build GRCh38
rs910146254 Chr.10: 75401898-75401898 on Build GRCh38
rs980952787 Chr.10: 75401741-75401741 on Build GRCh38
rs984290844 Chr.10: 75401799-75401799 on Build GRCh38
rs1012727370 Chr.10: 75401693-75401693 on Build GRCh38
rs376287796 Chr.10: 75402001-75402001 on Build GRCh38
rs375974337 Chr.10: 75402096-75402096 on Build GRCh38
rs550002110 Chr.10: 75401954-75401955 on Build GRCh38
rs960448667 Chr.10: 75401876-75401876 on Build GRCh38
rs963237484 Chr.10: 75401725-75401725 on Build GRCh38
rs552148046 Chr.10: 75401826-75401826 on Build GRCh38
rs951771677 Chr.10: 75402083-75402083 on Build GRCh38
rs562574074 Chr.10: 75401877-75401877 on Build GRCh38
rs1007851962 Chr.10: 75401654-75401676 on Build GRCh38
rs1022583407 Chr.10: 75401737-75401737 on Build GRCh38
rs1002057771 Chr.10: 75401860-75401860 on Build GRCh38
rs577463817 Chr.10: 75401956-75401956 on Build GRCh38
rs575403442 Chr.10: 75401679-75401679 on Build GRCh38
rs922686747 Chr.10: 75401913-75401913 on Build GRCh38
rs755564401 Chr.10: 75401955-75401955 on Build GRCh38
rs905119027 Chr.10: 75401967-75401967 on Build GRCh38
rs1034735421 Chr.10: 75401863-75401863 on Build GRCh38
rs372064586 Chr.10: 75401806-75401806 on Build GRCh38
rs536270097 Chr.10: 75401732-75401732 on Build GRCh38
rs892374710 Chr.10: 75401849-75401849 on Build GRCh38
rs866011897 Chr.10: 75401668-75401668 on Build GRCh38
rs111731434 Chr.10: 75402038-75402038 on Build GRCh38

View Full Product Details