Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 168421997-168422496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100505918
Entrez Gene ID
100505918
Gene Name
uncharacterized LOC100505918
Gene Aliases
-
UniGene
Hs.568494
Gene Chromosome Location
Chr.1: 168400188-168422656 on Build GRCh38
Transcripts
NR_037851.1

SNP Detail

SNP ID Chromosome Location
rs979135309 Chr.1: 168422050-168422050 on Build GRCh38
rs896371771 Chr.1: 168422428-168422428 on Build GRCh38
rs1019870572 Chr.1: 168422246-168422246 on Build GRCh38
rs140849572 Chr.1: 168422437-168422437 on Build GRCh38
rs75439666 Chr.1: 168422177-168422177 on Build GRCh38
rs768592477 Chr.1: 168422060-168422061 on Build GRCh38
rs755421121 Chr.1: 168422457-168422457 on Build GRCh38
rs566107957 Chr.1: 168422073-168422088 on Build GRCh38
rs758905269 Chr.1: 168422032-168422032 on Build GRCh38
rs1045175704 Chr.1: 168422183-168422183 on Build GRCh38
rs569685159 Chr.1: 168422231-168422231 on Build GRCh38
rs984658523 Chr.1: 168422100-168422100 on Build GRCh38
rs946079977 Chr.1: 168422045-168422045 on Build GRCh38
rs1048747359 Chr.1: 168422371-168422371 on Build GRCh38
rs1000974114 Chr.1: 168422201-168422201 on Build GRCh38
rs964347384 Chr.1: 168422256-168422256 on Build GRCh38
rs565181952 Chr.1: 168422466-168422466 on Build GRCh38
rs868751846 Chr.1: 168422091-168422091 on Build GRCh38
rs890611800 Chr.1: 168422113-168422113 on Build GRCh38
rs1001801444 Chr.1: 168422403-168422403 on Build GRCh38
rs986402749 Chr.1: 168422265-168422265 on Build GRCh38
rs535226890 Chr.1: 168422360-168422360 on Build GRCh38
rs940549432 Chr.1: 168422456-168422456 on Build GRCh38
rs766865230 Chr.1: 168422426-168422426 on Build GRCh38
rs564966050 Chr.1: 168422069-168422069 on Build GRCh38
rs201242975 Chr.1: 168422045-168422048 on Build GRCh38
rs151076517 Chr.1: 168422092-168422093 on Build GRCh38
rs559195172 Chr.1: 168422293-168422293 on Build GRCh38
rs542718570 Chr.1: 168422423-168422423 on Build GRCh38
rs1034913106 Chr.1: 168422405-168422405 on Build GRCh38
rs904849372 Chr.1: 168422399-168422399 on Build GRCh38
rs944834823 Chr.1: 168422114-168422114 on Build GRCh38
rs56231785 Chr.1: 168422088-168422089 on Build GRCh38
rs1036129485 Chr.1: 168422115-168422115 on Build GRCh38
rs947753823 Chr.1: 168422181-168422181 on Build GRCh38
rs572870022 Chr.1: 168422137-168422137 on Build GRCh38
rs865881949 Chr.1: 168422092-168422092 on Build GRCh38
rs796928050 Chr.1: 168422090-168422092 on Build GRCh38
rs925935470 Chr.1: 168422253-168422253 on Build GRCh38
rs926227471 Chr.1: 168422061-168422061 on Build GRCh38
rs1050613584 Chr.1: 168422086-168422089 on Build GRCh38
rs571007153 Chr.1: 168422329-168422329 on Build GRCh38
rs932216752 Chr.1: 168422085-168422085 on Build GRCh38
rs199913755 Chr.1: 168422045-168422052 on Build GRCh38
rs77622928 Chr.1: 168422275-168422275 on Build GRCh38
rs984606678 Chr.1: 168422409-168422409 on Build GRCh38
rs897535878 Chr.1: 168422121-168422121 on Build GRCh38
rs866596733 Chr.1: 168422089-168422089 on Build GRCh38
rs115568205 Chr.1: 168422458-168422458 on Build GRCh38
rs151058109 Chr.1: 168422230-168422230 on Build GRCh38
rs146589534 Chr.1: 168422078-168422079 on Build GRCh38
rs60417712 Chr.1: 168422073-168422073 on Build GRCh38
rs952803831 Chr.1: 168422382-168422382 on Build GRCh38
rs185236704 Chr.1: 168422392-168422392 on Build GRCh38
rs1033075965 Chr.1: 168422462-168422462 on Build GRCh38
rs1027061713 Chr.1: 168422357-168422357 on Build GRCh38
rs866528056 Chr.1: 168422073-168422073 on Build GRCh38
rs543718160 Chr.1: 168422039-168422039 on Build GRCh38
rs67235331 Chr.1: 168422045-168422060 on Build GRCh38
rs907781746 Chr.1: 168422108-168422108 on Build GRCh38
rs187301220 Chr.1: 168422204-168422204 on Build GRCh38
rs944864214 Chr.1: 168422281-168422281 on Build GRCh38
rs1021203471 Chr.1: 168422440-168422440 on Build GRCh38
rs940629079 Chr.1: 168422124-168422124 on Build GRCh38
rs147951520 Chr.1: 168422077-168422077 on Build GRCh38
rs932247469 Chr.1: 168422254-168422254 on Build GRCh38
rs538488704 Chr.1: 168422197-168422197 on Build GRCh38
rs78615681 Chr.1: 168422033-168422033 on Build GRCh38
rs1036898257 Chr.1: 168422335-168422335 on Build GRCh38
rs532324657 Chr.1: 168422172-168422172 on Build GRCh38
rs553250702 Chr.1: 168422361-168422361 on Build GRCh38
rs772029649 Chr.1: 168422044-168422045 on Build GRCh38

View Full Product Details