Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26649206-26649737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00202-2
Entrez Gene ID
731789
Gene Name
long intergenic non-protein coding RNA 202-2
Gene Aliases
-
UniGene
Hs.178186
Gene Chromosome Location
Chr.10: 26643108-26653454 on Build GRCh38
Transcripts
NR_026794.1

SNP Detail

SNP ID Chromosome Location
rs920804379 Chr.10: 26649240-26649240 on Build GRCh38
rs561652026 Chr.10: 26649245-26649245 on Build GRCh38
rs931163498 Chr.10: 26649363-26649363 on Build GRCh38
rs867913684 Chr.10: 26649248-26649248 on Build GRCh38
rs185379262 Chr.10: 26649588-26649588 on Build GRCh38
rs112210419 Chr.10: 26649283-26649283 on Build GRCh38
rs562806150 Chr.10: 26649333-26649333 on Build GRCh38
rs188238813 Chr.10: 26649683-26649683 on Build GRCh38
rs547463820 Chr.10: 26649455-26649455 on Build GRCh38
rs540268198 Chr.10: 26649271-26649271 on Build GRCh38
rs902675564 Chr.10: 26649456-26649456 on Build GRCh38
rs1042432562 Chr.10: 26649652-26649652 on Build GRCh38
rs942575203 Chr.10: 26649583-26649583 on Build GRCh38
rs2448100 Chr.10: 26649400-26649400 on Build GRCh38
rs111421964 Chr.10: 26649324-26649324 on Build GRCh38
rs901810181 Chr.10: 26649632-26649635 on Build GRCh38
rs574774739 Chr.10: 26649250-26649250 on Build GRCh38
rs190199942 Chr.10: 26649464-26649464 on Build GRCh38
rs574837769 Chr.10: 26649527-26649527 on Build GRCh38
rs369043448 Chr.10: 26649702-26649702 on Build GRCh38
rs1021504182 Chr.10: 26649272-26649272 on Build GRCh38
rs2477269 Chr.10: 26649329-26649329 on Build GRCh38
rs972511985 Chr.10: 26649348-26649348 on Build GRCh38
rs150038442 Chr.10: 26649419-26649419 on Build GRCh38
rs35659207 Chr.10: 26649522-26649522 on Build GRCh38
rs949120973 Chr.10: 26649340-26649340 on Build GRCh38
rs548061544 Chr.10: 26649377-26649377 on Build GRCh38
rs4749172 Chr.10: 26649468-26649468 on Build GRCh38
rs370168205 Chr.10: 26649469-26649469 on Build GRCh38
rs1025022975 Chr.10: 26649487-26649487 on Build GRCh38
rs71403866 Chr.10: 26649239-26649240 on Build GRCh38
rs779938382 Chr.10: 26649634-26649634 on Build GRCh38
rs567035867 Chr.10: 26649433-26649433 on Build GRCh38
rs1039414028 Chr.10: 26649401-26649401 on Build GRCh38
rs931133765 Chr.10: 26649577-26649577 on Build GRCh38
rs181408803 Chr.10: 26649482-26649482 on Build GRCh38
rs556241553 Chr.10: 26649472-26649472 on Build GRCh38
rs917666358 Chr.10: 26649337-26649337 on Build GRCh38
rs918395116 Chr.10: 26649350-26649350 on Build GRCh38
rs991095207 Chr.10: 26649481-26649481 on Build GRCh38
rs17558423 Chr.10: 26649365-26649365 on Build GRCh38
rs1032437683 Chr.10: 26649708-26649708 on Build GRCh38
rs11817226 Chr.10: 26649707-26649707 on Build GRCh38
rs576841494 Chr.10: 26649592-26649592 on Build GRCh38
rs933363014 Chr.10: 26649640-26649640 on Build GRCh38
rs911067905 Chr.10: 26649582-26649582 on Build GRCh38
rs2489579 Chr.10: 26649580-26649580 on Build GRCh38
rs970433017 Chr.10: 26649502-26649502 on Build GRCh38
rs901848994 Chr.10: 26649406-26649406 on Build GRCh38
rs889748438 Chr.10: 26649395-26649395 on Build GRCh38
rs979623311 Chr.10: 26649319-26649319 on Build GRCh38
rs201596271 Chr.10: 26649501-26649501 on Build GRCh38
rs576030629 Chr.10: 26649473-26649473 on Build GRCh38
rs11817200 Chr.10: 26649505-26649505 on Build GRCh38

View Full Product Details