Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26652861-26653405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00202-2
Entrez Gene ID
731789
Gene Name
long intergenic non-protein coding RNA 202-2
Gene Aliases
-
UniGene
Hs.178186
Gene Chromosome Location
Chr.10: 26643108-26653454 on Build GRCh38
Transcripts
NR_026794.1

SNP Detail

SNP ID Chromosome Location
rs867316316 Chr.10: 26652931-26652931 on Build GRCh38
rs368927860 Chr.10: 26653296-26653296 on Build GRCh38
rs917391589 Chr.10: 26653165-26653165 on Build GRCh38
rs927408155 Chr.10: 26652904-26652904 on Build GRCh38
rs968745253 Chr.10: 26653117-26653117 on Build GRCh38
rs565404729 Chr.10: 26653274-26653274 on Build GRCh38
rs992929061 Chr.10: 26653161-26653161 on Build GRCh38
rs978851043 Chr.10: 26652902-26652902 on Build GRCh38
rs543831872 Chr.10: 26653002-26653002 on Build GRCh38
rs930331166 Chr.10: 26653038-26653038 on Build GRCh38
rs116016002 Chr.10: 26652891-26652891 on Build GRCh38
rs1022656273 Chr.10: 26653111-26653111 on Build GRCh38
rs1050480251 Chr.10: 26653291-26653291 on Build GRCh38
rs190434991 Chr.10: 26653275-26653275 on Build GRCh38
rs537893052 Chr.10: 26653386-26653386 on Build GRCh38
rs561081024 Chr.10: 26653327-26653327 on Build GRCh38
rs112914380 Chr.10: 26653323-26653323 on Build GRCh38
rs546439337 Chr.10: 26653385-26653385 on Build GRCh38
rs558529024 Chr.10: 26652895-26652895 on Build GRCh38
rs71403867 Chr.10: 26653380-26653381 on Build GRCh38
rs1013069193 Chr.10: 26653144-26653144 on Build GRCh38
rs554731289 Chr.10: 26653085-26653085 on Build GRCh38
rs147906002 Chr.10: 26653153-26653153 on Build GRCh38
rs866306217 Chr.10: 26653055-26653055 on Build GRCh38
rs182887182 Chr.10: 26652915-26652915 on Build GRCh38
rs972324710 Chr.10: 26653215-26653215 on Build GRCh38
rs917424117 Chr.10: 26652934-26652934 on Build GRCh38
rs892281245 Chr.10: 26653370-26653370 on Build GRCh38
rs375471096 Chr.10: 26652996-26652996 on Build GRCh38
rs1009424800 Chr.10: 26653105-26653105 on Build GRCh38
rs930320132 Chr.10: 26653228-26653228 on Build GRCh38
rs888898040 Chr.10: 26653065-26653065 on Build GRCh38
rs112122210 Chr.10: 26653221-26653221 on Build GRCh38
rs530127366 Chr.10: 26653358-26653358 on Build GRCh38
rs1008608938 Chr.10: 26653080-26653080 on Build GRCh38
rs892319168 Chr.10: 26653103-26653103 on Build GRCh38
rs12260579 Chr.10: 26653196-26653196 on Build GRCh38
rs1036630341 Chr.10: 26652964-26652964 on Build GRCh38
rs140838110 Chr.10: 26653241-26653255 on Build GRCh38
rs528648534 Chr.10: 26653201-26653201 on Build GRCh38
rs780962577 Chr.10: 26653255-26653255 on Build GRCh38
rs920262069 Chr.10: 26652982-26652983 on Build GRCh38
rs1000637902 Chr.10: 26653140-26653140 on Build GRCh38
rs971623821 Chr.10: 26653227-26653227 on Build GRCh38
rs531113168 Chr.10: 26653254-26653254 on Build GRCh38
rs574568158 Chr.10: 26653145-26653145 on Build GRCh38
rs910222155 Chr.10: 26653305-26653305 on Build GRCh38
rs940897599 Chr.10: 26652938-26652938 on Build GRCh38

View Full Product Details