Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125705928-125706427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ37035
Entrez Gene ID
399821
Gene Name
uncharacterized LOC399821
Gene Aliases
-
UniGene
Hs.535286
Gene Chromosome Location
Chr.10: 125705290-125719493 on Build GRCh38
Transcripts
NR_033847.1 NR_033848.1

Gene Symbol
LOC283038
Entrez Gene ID
283038
Gene Name
uncharacterized LOC283038
Gene Aliases
-
UniGene
Hs.144911
Gene Chromosome Location
Chr.10: 125683243-125709677 on Build GRCh38
Transcripts
NR_033847.1 NR_033848.1

SNP Detail

SNP ID Chromosome Location
rs1015932564 Chr.10: 125705955-125705955 on Build GRCh38
rs990606920 Chr.10: 125706137-125706137 on Build GRCh38
rs10794017 Chr.10: 125706176-125706176 on Build GRCh38
rs991308480 Chr.10: 125706116-125706116 on Build GRCh38
rs76679523 Chr.10: 125706060-125706060 on Build GRCh38
rs770782503 Chr.10: 125706344-125706344 on Build GRCh38
rs186249577 Chr.10: 125706001-125706001 on Build GRCh38
rs752801186 Chr.10: 125706174-125706174 on Build GRCh38
rs141270125 Chr.10: 125706211-125706211 on Build GRCh38
rs201550106 Chr.10: 125706000-125706000 on Build GRCh38
rs903893863 Chr.10: 125706245-125706245 on Build GRCh38
rs370332058 Chr.10: 125705997-125705998 on Build GRCh38
rs911349634 Chr.10: 125706066-125706066 on Build GRCh38
rs562992763 Chr.10: 125706125-125706125 on Build GRCh38
rs773788663 Chr.10: 125706051-125706051 on Build GRCh38
rs955555724 Chr.10: 125706042-125706042 on Build GRCh38
rs573743918 Chr.10: 125705997-125705997 on Build GRCh38
rs74857349 Chr.10: 125706313-125706313 on Build GRCh38
rs1001258207 Chr.10: 125706266-125706266 on Build GRCh38
rs201808395 Chr.10: 125705962-125705963 on Build GRCh38
rs559000240 Chr.10: 125706038-125706038 on Build GRCh38
rs550955370 Chr.10: 125706121-125706121 on Build GRCh38
rs148092036 Chr.10: 125706145-125706145 on Build GRCh38
rs1009565588 Chr.10: 125706227-125706227 on Build GRCh38
rs183248774 Chr.10: 125706396-125706396 on Build GRCh38
rs45462095 Chr.10: 125705967-125705967 on Build GRCh38
rs921497967 Chr.10: 125706193-125706193 on Build GRCh38
rs755239516 Chr.10: 125706140-125706140 on Build GRCh38
rs191861363 Chr.10: 125706331-125706331 on Build GRCh38
rs575552742 Chr.10: 125706348-125706348 on Build GRCh38
rs1042859342 Chr.10: 125706233-125706233 on Build GRCh38
rs10794018 Chr.10: 125706219-125706219 on Build GRCh38
rs111411348 Chr.10: 125706014-125706014 on Build GRCh38
rs932848381 Chr.10: 125706205-125706205 on Build GRCh38
rs942765416 Chr.10: 125706087-125706087 on Build GRCh38
rs17153448 Chr.10: 125706143-125706143 on Build GRCh38
rs549221753 Chr.10: 125706276-125706276 on Build GRCh38
rs71868116 Chr.10: 125705972-125705973 on Build GRCh38
rs760252252 Chr.10: 125705996-125705997 on Build GRCh38
rs11244616 Chr.10: 125705971-125705971 on Build GRCh38
rs1033342333 Chr.10: 125706040-125706040 on Build GRCh38
rs71029267 Chr.10: 125705999-125706000 on Build GRCh38
rs1025929952 Chr.10: 125706366-125706366 on Build GRCh38
rs941015597 Chr.10: 125706146-125706146 on Build GRCh38
rs12354855 Chr.10: 125706142-125706142 on Build GRCh38
rs748170358 Chr.10: 125705971-125705972 on Build GRCh38

View Full Product Details