Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76153419-76153925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF157
Entrez Gene ID
114804
Gene Name
ring finger protein 157
Gene Aliases
-
UniGene
Hs.500643
Gene Chromosome Location
Chr.17: 76142453-76240368 on Build GRCh38
Transcripts
NM_052916.2 XM_005257007.4 XM_017024117.1 XM_017024118.1 XM_017024119.1 XM_017024120.1 NR_040017.1

Gene Symbol
RNF157-AS1
Entrez Gene ID
100507218
Gene Name
RNF157 antisense RNA 1
Gene Aliases
-
UniGene
Hs.654866
Gene Chromosome Location
Chr.17: 76140556-76154648 on Build GRCh38
Transcripts
NM_052916.2 XM_005257007.4 XM_017024117.1 XM_017024118.1 XM_017024119.1 XM_017024120.1 NR_040017.1

SNP Detail

SNP ID Chromosome Location
rs969061015 Chr.17: 76153477-76153477 on Build GRCh38
rs538631260 Chr.17: 76153772-76153772 on Build GRCh38
rs1037633953 Chr.17: 76153636-76153636 on Build GRCh38
rs569218497 Chr.17: 76153766-76153766 on Build GRCh38
rs960438908 Chr.17: 76153487-76153487 on Build GRCh38
rs572474749 Chr.17: 76153886-76153886 on Build GRCh38
rs963313586 Chr.17: 76153462-76153462 on Build GRCh38
rs989309668 Chr.17: 76153875-76153875 on Build GRCh38
rs547013459 Chr.17: 76153515-76153515 on Build GRCh38
rs755659505 Chr.17: 76153463-76153463 on Build GRCh38
rs534944503 Chr.17: 76153559-76153559 on Build GRCh38
rs970272379 Chr.17: 76153820-76153820 on Build GRCh38
rs913615828 Chr.17: 76153793-76153793 on Build GRCh38
rs748039419 Chr.17: 76153729-76153729 on Build GRCh38
rs772021408 Chr.17: 76153792-76153792 on Build GRCh38
rs1004233899 Chr.17: 76153609-76153609 on Build GRCh38
rs933205143 Chr.17: 76153890-76153890 on Build GRCh38
rs112237060 Chr.17: 76153526-76153526 on Build GRCh38
rs1023611518 Chr.17: 76153476-76153476 on Build GRCh38
rs988421164 Chr.17: 76153489-76153489 on Build GRCh38
rs1023138006 Chr.17: 76153505-76153505 on Build GRCh38
rs116286995 Chr.17: 76153781-76153781 on Build GRCh38
rs1038973016 Chr.17: 76153634-76153634 on Build GRCh38
rs113973281 Chr.17: 76153677-76153677 on Build GRCh38
rs1034684590 Chr.17: 76153707-76153707 on Build GRCh38
rs1868821 Chr.17: 76153608-76153608 on Build GRCh38
rs921212752 Chr.17: 76153860-76153860 on Build GRCh38
rs934725384 Chr.17: 76153561-76153561 on Build GRCh38
rs921137750 Chr.17: 76153570-76153570 on Build GRCh38
rs981568745 Chr.17: 76153558-76153559 on Build GRCh38
rs927708794 Chr.17: 76153481-76153481 on Build GRCh38
rs996056622 Chr.17: 76153662-76153662 on Build GRCh38
rs188095055 Chr.17: 76153703-76153703 on Build GRCh38
rs891175513 Chr.17: 76153440-76153440 on Build GRCh38
rs183858537 Chr.17: 76153531-76153531 on Build GRCh38
rs1868822 Chr.17: 76153443-76153443 on Build GRCh38
rs79666138 Chr.17: 76153866-76153866 on Build GRCh38
rs539970395 Chr.17: 76153541-76153541 on Build GRCh38
rs754708 Chr.17: 76153888-76153888 on Build GRCh38
rs898677088 Chr.17: 76153657-76153657 on Build GRCh38
rs772826312 Chr.17: 76153799-76153799 on Build GRCh38
rs556785851 Chr.17: 76153704-76153704 on Build GRCh38
rs371227634 Chr.17: 76153478-76153478 on Build GRCh38
rs775758458 Chr.17: 76153889-76153889 on Build GRCh38
rs879942425 Chr.17: 76153818-76153818 on Build GRCh38
rs968746081 Chr.17: 76153888-76153889 on Build GRCh38
rs1051068529 Chr.17: 76153883-76153883 on Build GRCh38
rs376723711 Chr.17: 76153453-76153453 on Build GRCh38
rs970264854 Chr.17: 76153701-76153701 on Build GRCh38
rs898534668 Chr.17: 76153891-76153891 on Build GRCh38
rs539546906 Chr.17: 76153492-76153492 on Build GRCh38
rs1036288785 Chr.17: 76153838-76153838 on Build GRCh38
rs866946514 Chr.17: 76153643-76153643 on Build GRCh38
rs560080091 Chr.17: 76153562-76153568 on Build GRCh38
rs1023211597 Chr.17: 76153800-76153800 on Build GRCh38
rs746814585 Chr.17: 76153822-76153822 on Build GRCh38
rs1051038053 Chr.17: 76153602-76153602 on Build GRCh38
rs748233528 Chr.17: 76153804-76153804 on Build GRCh38
rs932660297 Chr.17: 76153590-76153590 on Build GRCh38
rs551523646 Chr.17: 76153873-76153873 on Build GRCh38
rs921844712 Chr.17: 76153650-76153650 on Build GRCh38
rs370610143 Chr.17: 76153892-76153892 on Build GRCh38
rs553484224 Chr.17: 76153782-76153782 on Build GRCh38
rs1037424600 Chr.17: 76153785-76153786 on Build GRCh38
rs866457122 Chr.17: 76153479-76153479 on Build GRCh38
rs149306290 Chr.17: 76153811-76153811 on Build GRCh38
rs947337113 Chr.17: 76153625-76153625 on Build GRCh38

View Full Product Details