Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 6730539-6731043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00487
Entrez Gene ID
400941
Gene Name
long intergenic non-protein coding RNA 487
Gene Aliases
-
UniGene
Hs.446195
Gene Chromosome Location
Chr.2: 6729168-6770311 on Build GRCh38
Transcripts
NR_038369.1

SNP Detail

SNP ID Chromosome Location
rs990036520 Chr.2: 6730568-6730568 on Build GRCh38
rs919804613 Chr.2: 6730777-6730777 on Build GRCh38
rs146632002 Chr.2: 6730593-6730593 on Build GRCh38
rs75638545 Chr.2: 6730951-6730951 on Build GRCh38
rs115584841 Chr.2: 6730867-6730867 on Build GRCh38
rs932632365 Chr.2: 6730656-6730656 on Build GRCh38
rs956466462 Chr.2: 6731000-6731000 on Build GRCh38
rs918439606 Chr.2: 6731006-6731006 on Build GRCh38
rs752952175 Chr.2: 6730605-6730605 on Build GRCh38
rs1043329249 Chr.2: 6730932-6730932 on Build GRCh38
rs79485832 Chr.2: 6730700-6730700 on Build GRCh38
rs1021084996 Chr.2: 6730889-6730889 on Build GRCh38
rs1045652276 Chr.2: 6730702-6730702 on Build GRCh38
rs563844037 Chr.2: 6730573-6730573 on Build GRCh38
rs1037272303 Chr.2: 6730667-6730667 on Build GRCh38
rs972810008 Chr.2: 6731001-6731001 on Build GRCh38
rs75428033 Chr.2: 6730699-6730699 on Build GRCh38
rs552050096 Chr.2: 6730899-6730899 on Build GRCh38
rs992251708 Chr.2: 6730713-6730713 on Build GRCh38
rs917571561 Chr.2: 6730748-6730748 on Build GRCh38
rs114904090 Chr.2: 6730659-6730659 on Build GRCh38
rs77807024 Chr.2: 6730697-6730697 on Build GRCh38
rs963111656 Chr.2: 6730998-6730998 on Build GRCh38
rs976936703 Chr.2: 6730604-6730604 on Build GRCh38
rs549425245 Chr.2: 6730752-6730752 on Build GRCh38
rs1021201127 Chr.2: 6730588-6730588 on Build GRCh38
rs1054233117 Chr.2: 6730820-6730820 on Build GRCh38
rs997544574 Chr.2: 6730913-6730913 on Build GRCh38
rs764250374 Chr.2: 6730770-6730770 on Build GRCh38
rs969745672 Chr.2: 6730761-6730761 on Build GRCh38
rs766663496 Chr.2: 6730900-6730900 on Build GRCh38
rs78298137 Chr.2: 6730896-6730896 on Build GRCh38
rs1025101816 Chr.2: 6731007-6731007 on Build GRCh38
rs16865565 Chr.2: 6730567-6730567 on Build GRCh38
rs980711807 Chr.2: 6730774-6730774 on Build GRCh38
rs910009875 Chr.2: 6730972-6730972 on Build GRCh38
rs904739283 Chr.2: 6730941-6730941 on Build GRCh38
rs141059729 Chr.2: 6730595-6730595 on Build GRCh38
rs941229023 Chr.2: 6730666-6730666 on Build GRCh38
rs909944832 Chr.2: 6730662-6730662 on Build GRCh38
rs751392431 Chr.2: 6730870-6730870 on Build GRCh38
rs1001592338 Chr.2: 6730773-6730773 on Build GRCh38
rs949958147 Chr.2: 6730688-6730688 on Build GRCh38
rs949834921 Chr.2: 6731019-6731019 on Build GRCh38
rs528850746 Chr.2: 6730679-6730679 on Build GRCh38
rs985849416 Chr.2: 6730955-6730955 on Build GRCh38
rs376157641 Chr.2: 6730886-6730886 on Build GRCh38

View Full Product Details