Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 4731436-4731949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRNT
Entrez Gene ID
149830
Gene Name
prion protein (testis specific)
Gene Aliases
M8
UniGene
Hs.126516
Gene Chromosome Location
Chr.20: 4731282-4740668 on Build GRCh38
Transcripts
NR_024267.1 NR_024268.1 NR_024269.1

SNP Detail

SNP ID Chromosome Location
rs955439493 Chr.20: 4731831-4731831 on Build GRCh38
rs918221999 Chr.20: 4731675-4731675 on Build GRCh38
rs35939434 Chr.20: 4731895-4731896 on Build GRCh38
rs561872318 Chr.20: 4731857-4731857 on Build GRCh38
rs1031045128 Chr.20: 4731618-4731618 on Build GRCh38
rs73896144 Chr.20: 4731493-4731493 on Build GRCh38
rs1044936933 Chr.20: 4731760-4731760 on Build GRCh38
rs780555679 Chr.20: 4731886-4731886 on Build GRCh38
rs964133630 Chr.20: 4731658-4731658 on Build GRCh38
rs34617801 Chr.20: 4731824-4731825 on Build GRCh38
rs984669647 Chr.20: 4731688-4731688 on Build GRCh38
rs531950814 Chr.20: 4731478-4731478 on Build GRCh38
rs918280640 Chr.20: 4731900-4731900 on Build GRCh38
rs188966659 Chr.20: 4731856-4731856 on Build GRCh38
rs758144292 Chr.20: 4731741-4731741 on Build GRCh38
rs988683195 Chr.20: 4731591-4731591 on Build GRCh38
rs112639485 Chr.20: 4731747-4731747 on Build GRCh38
rs181956711 Chr.20: 4731592-4731592 on Build GRCh38
rs904017291 Chr.20: 4731852-4731852 on Build GRCh38
rs891793692 Chr.20: 4731813-4731813 on Build GRCh38
rs947789514 Chr.20: 4731822-4731822 on Build GRCh38
rs113632499 Chr.20: 4731728-4731728 on Build GRCh38
rs1010129981 Chr.20: 4731820-4731820 on Build GRCh38
rs1000205227 Chr.20: 4731777-4731777 on Build GRCh38
rs973735095 Chr.20: 4731667-4731667 on Build GRCh38
rs548314922 Chr.20: 4731650-4731650 on Build GRCh38
rs766561398 Chr.20: 4731636-4731636 on Build GRCh38
rs756860993 Chr.20: 4731788-4731788 on Build GRCh38
rs974999748 Chr.20: 4731662-4731662 on Build GRCh38
rs1001086015 Chr.20: 4731616-4731616 on Build GRCh38
rs181787986 Chr.20: 4731926-4731926 on Build GRCh38
rs184518707 Chr.20: 4731830-4731830 on Build GRCh38
rs1033010119 Chr.20: 4731805-4731805 on Build GRCh38
rs962460623 Chr.20: 4731890-4731890 on Build GRCh38
rs1020105289 Chr.20: 4731654-4731654 on Build GRCh38
rs750721504 Chr.20: 4731457-4731457 on Build GRCh38
rs552465789 Chr.20: 4731737-4731737 on Build GRCh38
rs79912297 Chr.20: 4731539-4731539 on Build GRCh38
rs905951154 Chr.20: 4731769-4731769 on Build GRCh38
rs538988555 Chr.20: 4731803-4731803 on Build GRCh38
rs541563119 Chr.20: 4731683-4731683 on Build GRCh38
rs929584314 Chr.20: 4731682-4731682 on Build GRCh38
rs955966156 Chr.20: 4731578-4731578 on Build GRCh38
rs569731863 Chr.20: 4731664-4731664 on Build GRCh38
rs1001158131 Chr.20: 4731885-4731885 on Build GRCh38
rs917675197 Chr.20: 4731776-4731776 on Build GRCh38
rs1000173804 Chr.20: 4731535-4731535 on Build GRCh38
rs941064190 Chr.20: 4731655-4731655 on Build GRCh38
rs150186066 Chr.20: 4731891-4731891 on Build GRCh38
rs951996776 Chr.20: 4731671-4731671 on Build GRCh38
rs1045649617 Chr.20: 4731833-4731833 on Build GRCh38
rs955214128 Chr.20: 4731613-4731613 on Build GRCh38
rs985641385 Chr.20: 4731627-4731627 on Build GRCh38
rs376110282 Chr.20: 4731828-4731828 on Build GRCh38
rs558981085 Chr.20: 4731573-4731573 on Build GRCh38

View Full Product Details