Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38425278-38425782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNHG17
Entrez Gene ID
388796
Gene Name
small nucleolar RNA host gene 17
Gene Aliases
-
UniGene
Hs.400876
Gene Chromosome Location
Chr.20: 38420592-38435375 on Build GRCh38
Transcripts
NR_015366.4 NR_027241.2 NR_002910.2

Gene Symbol
SNORA71B
Entrez Gene ID
26776
Gene Name
small nucleolar RNA, H/ACA box 71B
Gene Aliases
RNU71B, U71b
UniGene
Hs.400876
Gene Chromosome Location
Chr.20: 38425195-38425330 on Build GRCh38
Transcripts
NR_015366.4 NR_027241.2 NR_002910.2

SNP Detail

SNP ID Chromosome Location
rs761110859 Chr.20: 38425407-38425407 on Build GRCh38
rs925776455 Chr.20: 38425338-38425338 on Build GRCh38
rs41310014 Chr.20: 38425619-38425619 on Build GRCh38
rs750934738 Chr.20: 38425311-38425311 on Build GRCh38
rs75984216 Chr.20: 38425459-38425459 on Build GRCh38
rs567880230 Chr.20: 38425536-38425536 on Build GRCh38
rs1045662087 Chr.20: 38425614-38425614 on Build GRCh38
rs994407023 Chr.20: 38425569-38425569 on Build GRCh38
rs986339557 Chr.20: 38425698-38425698 on Build GRCh38
rs1050185134 Chr.20: 38425463-38425463 on Build GRCh38
rs879122068 Chr.20: 38425441-38425441 on Build GRCh38
rs939110012 Chr.20: 38425621-38425622 on Build GRCh38
rs1040419411 Chr.20: 38425544-38425544 on Build GRCh38
rs892134330 Chr.20: 38425649-38425650 on Build GRCh38
rs182029357 Chr.20: 38425457-38425457 on Build GRCh38
rs910317955 Chr.20: 38425702-38425702 on Build GRCh38
rs992861575 Chr.20: 38425630-38425630 on Build GRCh38
rs550307203 Chr.20: 38425542-38425542 on Build GRCh38
rs748833357 Chr.20: 38425331-38425331 on Build GRCh38
rs1034449044 Chr.20: 38425584-38425584 on Build GRCh38
rs111597668 Chr.20: 38425527-38425527 on Build GRCh38
rs375885451 Chr.20: 38425322-38425322 on Build GRCh38
rs371518605 Chr.20: 38425317-38425317 on Build GRCh38
rs1025849531 Chr.20: 38425570-38425570 on Build GRCh38
rs779774886 Chr.20: 38425344-38425347 on Build GRCh38
rs903808855 Chr.20: 38425736-38425736 on Build GRCh38
rs925832233 Chr.20: 38425649-38425649 on Build GRCh38
rs546793684 Chr.20: 38425533-38425533 on Build GRCh38
rs910167885 Chr.20: 38425581-38425581 on Build GRCh38
rs888615687 Chr.20: 38425571-38425571 on Build GRCh38
rs778842414 Chr.20: 38425360-38425360 on Build GRCh38
rs764371022 Chr.20: 38425461-38425461 on Build GRCh38
rs892103032 Chr.20: 38425445-38425445 on Build GRCh38
rs557912196 Chr.20: 38425398-38425398 on Build GRCh38
rs759701383 Chr.20: 38425742-38425742 on Build GRCh38
rs767876786 Chr.20: 38425362-38425362 on Build GRCh38
rs185826677 Chr.20: 38425546-38425546 on Build GRCh38
rs535250749 Chr.20: 38425541-38425541 on Build GRCh38
rs977301868 Chr.20: 38425645-38425645 on Build GRCh38
rs370308973 Chr.20: 38425720-38425720 on Build GRCh38
rs967916109 Chr.20: 38425512-38425512 on Build GRCh38
rs1024709785 Chr.20: 38425643-38425643 on Build GRCh38
rs774244582 Chr.20: 38425314-38425314 on Build GRCh38
rs1003276139 Chr.20: 38425354-38425354 on Build GRCh38
rs190628220 Chr.20: 38425665-38425665 on Build GRCh38
rs770761270 Chr.20: 38425353-38425353 on Build GRCh38
rs755537467 Chr.20: 38425321-38425321 on Build GRCh38
rs368535357 Chr.20: 38425312-38425312 on Build GRCh38
rs75295914 Chr.20: 38425644-38425644 on Build GRCh38
rs41314902 Chr.20: 38425573-38425573 on Build GRCh38
rs747624722 Chr.20: 38425320-38425320 on Build GRCh38
rs750313451 Chr.20: 38425522-38425522 on Build GRCh38
rs75727390 Chr.20: 38425562-38425562 on Build GRCh38
rs1022129465 Chr.20: 38425480-38425480 on Build GRCh38
rs754263050 Chr.20: 38425557-38425557 on Build GRCh38
rs935844341 Chr.20: 38425693-38425693 on Build GRCh38

View Full Product Details