Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26555145-26555666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR548J
Entrez Gene ID
100313914
Gene Name
microRNA 548j
Gene Aliases
MIRN548J, hsa-mir-548j
UniGene
Gene Chromosome Location
Chr.22: 26555212-26555323 on Build GRCh38
Transcripts
NR_031615.1 NM_001008566.1 NM_003595.3 XM_006724338.3

Gene Symbol
TPST2
Entrez Gene ID
8459
Gene Name
tyrosylprotein sulfotransferase 2
Gene Aliases
TANGO13B
UniGene
Hs.632768
Gene Chromosome Location
Chr.22: 26524082-26590125 on Build GRCh38
Transcripts
NR_031615.1 NM_001008566.1 NM_003595.3 XM_006724338.3

SNP Detail

SNP ID Chromosome Location
rs758519723 Chr.22: 26555509-26555509 on Build GRCh38
rs974829347 Chr.22: 26555476-26555476 on Build GRCh38
rs770280725 Chr.22: 26555230-26555230 on Build GRCh38
rs763737409 Chr.22: 26555187-26555187 on Build GRCh38
rs185307887 Chr.22: 26555404-26555404 on Build GRCh38
rs4820685 Chr.22: 26555623-26555623 on Build GRCh38
rs916272082 Chr.22: 26555559-26555559 on Build GRCh38
rs976434292 Chr.22: 26555292-26555292 on Build GRCh38
rs559170967 Chr.22: 26555354-26555354 on Build GRCh38
rs767118859 Chr.22: 26555256-26555256 on Build GRCh38
rs769059822 Chr.22: 26555321-26555321 on Build GRCh38
rs73880125 Chr.22: 26555480-26555480 on Build GRCh38
rs760105319 Chr.22: 26555273-26555273 on Build GRCh38
rs1046180267 Chr.22: 26555621-26555621 on Build GRCh38
rs750378876 Chr.22: 26555201-26555201 on Build GRCh38
rs909753931 Chr.22: 26555353-26555353 on Build GRCh38
rs538834475 Chr.22: 26555620-26555623 on Build GRCh38
rs751780445 Chr.22: 26555284-26555284 on Build GRCh38
rs771781711 Chr.22: 26555326-26555326 on Build GRCh38
rs886136783 Chr.22: 26555175-26555175 on Build GRCh38
rs780586827 Chr.22: 26555302-26555302 on Build GRCh38
rs1035624702 Chr.22: 26555231-26555231 on Build GRCh38
rs1005885258 Chr.22: 26555183-26555183 on Build GRCh38
rs1031819873 Chr.22: 26555599-26555599 on Build GRCh38
rs756431448 Chr.22: 26555223-26555236 on Build GRCh38
rs765859130 Chr.22: 26555348-26555348 on Build GRCh38
rs908126660 Chr.22: 26555515-26555515 on Build GRCh38
rs545900525 Chr.22: 26555280-26555280 on Build GRCh38
rs569240631 Chr.22: 26555639-26555639 on Build GRCh38
rs773027463 Chr.22: 26555330-26555330 on Build GRCh38
rs188578117 Chr.22: 26555507-26555507 on Build GRCh38
rs1037390970 Chr.22: 26555557-26555557 on Build GRCh38
rs751278896 Chr.22: 26555191-26555191 on Build GRCh38
rs548302511 Chr.22: 26555406-26555406 on Build GRCh38
rs149611690 Chr.22: 26555420-26555420 on Build GRCh38
rs1021550978 Chr.22: 26555453-26555453 on Build GRCh38
rs997899646 Chr.22: 26555617-26555617 on Build GRCh38
rs546943051 Chr.22: 26555331-26555331 on Build GRCh38
rs12161068 Chr.22: 26555249-26555249 on Build GRCh38
rs371252751 Chr.22: 26555223-26555223 on Build GRCh38
rs779864803 Chr.22: 26555202-26555202 on Build GRCh38
rs749890635 Chr.22: 26555271-26555271 on Build GRCh38
rs767174170 Chr.22: 26555196-26555196 on Build GRCh38
rs758195449 Chr.22: 26555369-26555369 on Build GRCh38
rs757304959 Chr.22: 26555641-26555641 on Build GRCh38
rs919358317 Chr.22: 26555399-26555399 on Build GRCh38
rs28402932 Chr.22: 26555351-26555351 on Build GRCh38
rs964665741 Chr.22: 26555471-26555471 on Build GRCh38
rs138333951 Chr.22: 26555413-26555413 on Build GRCh38
rs752431410 Chr.22: 26555359-26555359 on Build GRCh38
rs1037782151 Chr.22: 26555393-26555393 on Build GRCh38
rs985416656 Chr.22: 26555630-26555630 on Build GRCh38
rs193172216 Chr.22: 26555171-26555171 on Build GRCh38
rs942162812 Chr.22: 26555180-26555180 on Build GRCh38
rs144361364 Chr.22: 26555629-26555629 on Build GRCh38
rs530474414 Chr.22: 26555436-26555436 on Build GRCh38
rs1030955297 Chr.22: 26555428-26555428 on Build GRCh38
rs769796998 Chr.22: 26555299-26555299 on Build GRCh38
rs4822739 Chr.22: 26555219-26555219 on Build GRCh38
rs760186245 Chr.22: 26555335-26555335 on Build GRCh38
rs746874366 Chr.22: 26555328-26555328 on Build GRCh38
rs757533254 Chr.22: 26555285-26555285 on Build GRCh38
rs368755594 Chr.22: 26555222-26555222 on Build GRCh38
rs942231128 Chr.22: 26555516-26555516 on Build GRCh38
rs894910223 Chr.22: 26555372-26555372 on Build GRCh38
rs367750295 Chr.22: 26555290-26555290 on Build GRCh38
rs895788232 Chr.22: 26555426-26555426 on Build GRCh38
rs763326764 Chr.22: 26555239-26555239 on Build GRCh38
rs1048012083 Chr.22: 26555336-26555336 on Build GRCh38
rs1023140850 Chr.22: 26555265-26555265 on Build GRCh38
rs906394881 Chr.22: 26555411-26555412 on Build GRCh38
rs371948789 Chr.22: 26555303-26555303 on Build GRCh38
rs565141718 Chr.22: 26555283-26555283 on Build GRCh38
rs753744163 Chr.22: 26555282-26555282 on Build GRCh38
rs775867210 Chr.22: 26555237-26555237 on Build GRCh38
rs551130138 Chr.22: 26555584-26555584 on Build GRCh38
rs8141834 Chr.22: 26555642-26555642 on Build GRCh38
rs375330417 Chr.22: 26555325-26555325 on Build GRCh38
rs753598986 Chr.22: 26555210-26555210 on Build GRCh38
rs539695691 Chr.22: 26555600-26555600 on Build GRCh38
rs576736626 Chr.22: 26555313-26555313 on Build GRCh38
rs969282802 Chr.22: 26555272-26555272 on Build GRCh38
rs955775888 Chr.22: 26555613-26555613 on Build GRCh38
rs768949575 Chr.22: 26555244-26555244 on Build GRCh38
rs370861124 Chr.22: 26555214-26555214 on Build GRCh38
rs774912752 Chr.22: 26555323-26555323 on Build GRCh38
rs957907567 Chr.22: 26555435-26555435 on Build GRCh38
rs754330349 Chr.22: 26555349-26555349 on Build GRCh38
rs745895224 Chr.22: 26555298-26555298 on Build GRCh38
rs192691159 Chr.22: 26555519-26555519 on Build GRCh38

View Full Product Details