Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52268166-52268666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIRLET7G
Entrez Gene ID
406890
Gene Name
microRNA let-7g
Gene Aliases
LET7G, MIRNLET7G, hsa-let-7g, let-7g
UniGene
Gene Chromosome Location
Chr.3: 52268278-52268361 on Build GRCh38
Transcripts
NR_029660.1 NM_025222.3 XM_011534136.2

Gene Symbol
WDR82
Entrez Gene ID
80335
Gene Name
WD repeat domain 82
Gene Aliases
MST107, MSTP107, PRO2730, PRO34047, SWD2, TMEM113, WDR82A
UniGene
Hs.194110
Gene Chromosome Location
Chr.3: 52254422-52278643 on Build GRCh38
Transcripts
NR_029660.1 NM_025222.3 XM_011534136.2

SNP Detail

SNP ID Chromosome Location
rs757576721 Chr.3: 52268267-52268267 on Build GRCh38
rs528135469 Chr.3: 52268382-52268382 on Build GRCh38
rs374668887 Chr.3: 52268384-52268384 on Build GRCh38
rs1022640944 Chr.3: 52268290-52268290 on Build GRCh38
rs1022588710 Chr.3: 52268531-52268531 on Build GRCh38
rs528905858 Chr.3: 52268403-52268403 on Build GRCh38
rs772443356 Chr.3: 52268554-52268554 on Build GRCh38
rs897930276 Chr.3: 52268512-52268512 on Build GRCh38
rs12171285 Chr.3: 52268245-52268245 on Build GRCh38
rs1008105194 Chr.3: 52268314-52268314 on Build GRCh38
rs1016446622 Chr.3: 52268450-52268450 on Build GRCh38
rs941680150 Chr.3: 52268438-52268438 on Build GRCh38
rs542794187 Chr.3: 52268315-52268315 on Build GRCh38
rs531474068 Chr.3: 52268597-52268597 on Build GRCh38
rs145909524 Chr.3: 52268270-52268270 on Build GRCh38
rs886600249 Chr.3: 52268443-52268443 on Build GRCh38
rs764998060 Chr.3: 52268318-52268318 on Build GRCh38
rs1012502332 Chr.3: 52268506-52268506 on Build GRCh38
rs371455918 Chr.3: 52268503-52268503 on Build GRCh38
rs79178707 Chr.3: 52268508-52268508 on Build GRCh38
rs542312900 Chr.3: 52268447-52268447 on Build GRCh38
rs956046214 Chr.3: 52268619-52268619 on Build GRCh38
rs1030515521 Chr.3: 52268203-52268203 on Build GRCh38
rs778561804 Chr.3: 52268269-52268269 on Build GRCh38
rs1017773990 Chr.3: 52268325-52268326 on Build GRCh38
rs773092062 Chr.3: 52268316-52268318 on Build GRCh38
rs1006418856 Chr.3: 52268446-52268446 on Build GRCh38
rs564699743 Chr.3: 52268472-52268472 on Build GRCh38
rs776344208 Chr.3: 52268259-52268259 on Build GRCh38
rs370150007 Chr.3: 52268386-52268386 on Build GRCh38
rs962340675 Chr.3: 52268236-52268236 on Build GRCh38
rs1016550148 Chr.3: 52268201-52268201 on Build GRCh38
rs934544154 Chr.3: 52268381-52268381 on Build GRCh38
rs1006881785 Chr.3: 52268192-52268192 on Build GRCh38
rs746565785 Chr.3: 52268465-52268465 on Build GRCh38
rs971128611 Chr.3: 52268313-52268313 on Build GRCh38
rs933405066 Chr.3: 52268426-52268426 on Build GRCh38
rs9631505 Chr.3: 52268263-52268263 on Build GRCh38
rs542149216 Chr.3: 52268566-52268566 on Build GRCh38
rs776605542 Chr.3: 52268561-52268561 on Build GRCh38
rs971075350 Chr.3: 52268540-52268540 on Build GRCh38
rs941221860 Chr.3: 52268474-52268474 on Build GRCh38
rs532857455 Chr.3: 52268636-52268636 on Build GRCh38
rs371266565 Chr.3: 52268326-52268326 on Build GRCh38
rs1047848617 Chr.3: 52268432-52268432 on Build GRCh38
rs1037733381 Chr.3: 52268490-52268492 on Build GRCh38
rs1007806713 Chr.3: 52268618-52268618 on Build GRCh38
rs901896159 Chr.3: 52268416-52268416 on Build GRCh38
rs1031447218 Chr.3: 52268578-52268578 on Build GRCh38
rs186206259 Chr.3: 52268387-52268387 on Build GRCh38

View Full Product Details