Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 109184962-109185497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00488
Entrez Gene ID
677779
Gene Name
long intergenic non-protein coding RNA 488
Gene Aliases
C3orf66
UniGene
Hs.618669
Gene Chromosome Location
Chr.3: 109178165-109185261 on Build GRCh38
Transcripts
NR_026767.1

SNP Detail

SNP ID Chromosome Location
rs183610733 Chr.3: 109185023-109185023 on Build GRCh38
rs941140990 Chr.3: 109185340-109185340 on Build GRCh38
rs960146014 Chr.3: 109185046-109185046 on Build GRCh38
rs772855813 Chr.3: 109185007-109185007 on Build GRCh38
rs911417075 Chr.3: 109185159-109185159 on Build GRCh38
rs970249487 Chr.3: 109185025-109185025 on Build GRCh38
rs980240654 Chr.3: 109185034-109185034 on Build GRCh38
rs972827853 Chr.3: 109185077-109185077 on Build GRCh38
rs577758804 Chr.3: 109185391-109185391 on Build GRCh38
rs577852045 Chr.3: 109185032-109185032 on Build GRCh38
rs909633648 Chr.3: 109185328-109185328 on Build GRCh38
rs578069230 Chr.3: 109185016-109185016 on Build GRCh38
rs952874004 Chr.3: 109185416-109185416 on Build GRCh38
rs141746612 Chr.3: 109185021-109185021 on Build GRCh38
rs550935647 Chr.3: 109185294-109185294 on Build GRCh38
rs977036558 Chr.3: 109185215-109185215 on Build GRCh38
rs1035817055 Chr.3: 109185035-109185035 on Build GRCh38
rs1055532618 Chr.3: 109185392-109185392 on Build GRCh38
rs866340366 Chr.3: 109185029-109185029 on Build GRCh38
rs984501563 Chr.3: 109185170-109185170 on Build GRCh38
rs1014117085 Chr.3: 109185441-109185441 on Build GRCh38
rs923093298 Chr.3: 109185275-109185275 on Build GRCh38
rs891516548 Chr.3: 109185301-109185301 on Build GRCh38
rs891447424 Chr.3: 109185021-109185021 on Build GRCh38
rs1031788612 Chr.3: 109185003-109185003 on Build GRCh38
rs960176775 Chr.3: 109185384-109185384 on Build GRCh38
rs931346644 Chr.3: 109185130-109185130 on Build GRCh38
rs541632902 Chr.3: 109185065-109185065 on Build GRCh38
rs922261051 Chr.3: 109185288-109185288 on Build GRCh38
rs906163289 Chr.3: 109185367-109185367 on Build GRCh38
rs909139273 Chr.3: 109185199-109185199 on Build GRCh38
rs7639593 Chr.3: 109185454-109185454 on Build GRCh38
rs958164000 Chr.3: 109185116-109185116 on Build GRCh38
rs753259210 Chr.3: 109185177-109185177 on Build GRCh38
rs1046785505 Chr.3: 109185317-109185317 on Build GRCh38
rs528306738 Chr.3: 109185039-109185039 on Build GRCh38
rs949817 Chr.3: 109185411-109185411 on Build GRCh38
rs1035846712 Chr.3: 109185378-109185378 on Build GRCh38
rs1043077078 Chr.3: 109185332-109185332 on Build GRCh38
rs1011248893 Chr.3: 109185311-109185311 on Build GRCh38
rs999582770 Chr.3: 109185084-109185084 on Build GRCh38
rs1001732779 Chr.3: 109185371-109185371 on Build GRCh38
rs570155598 Chr.3: 109185432-109185432 on Build GRCh38
rs943229611 Chr.3: 109185204-109185204 on Build GRCh38
rs1038927108 Chr.3: 109185274-109185274 on Build GRCh38
rs989635612 Chr.3: 109185150-109185150 on Build GRCh38
rs561826632 Chr.3: 109185092-109185092 on Build GRCh38

View Full Product Details