Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122889374-122889903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100129550
Entrez Gene ID
100129550
Gene Name
uncharacterized LOC100129550
Gene Aliases
-
UniGene
Hs.61558
Gene Chromosome Location
Chr.3: 122886513-122892416 on Build GRCh38
Transcripts
NR_024618.1

SNP Detail

SNP ID Chromosome Location
rs1049100929 Chr.3: 122889443-122889443 on Build GRCh38
rs532474612 Chr.3: 122889452-122889452 on Build GRCh38
rs922788795 Chr.3: 122889576-122889576 on Build GRCh38
rs929297022 Chr.3: 122889442-122889442 on Build GRCh38
rs538445095 Chr.3: 122889629-122889629 on Build GRCh38
rs949989958 Chr.3: 122889713-122889713 on Build GRCh38
rs200100971 Chr.3: 122889619-122889619 on Build GRCh38
rs565623331 Chr.3: 122889747-122889747 on Build GRCh38
rs888464576 Chr.3: 122889812-122889812 on Build GRCh38
rs758513325 Chr.3: 122889605-122889606 on Build GRCh38
rs945828717 Chr.3: 122889600-122889600 on Build GRCh38
rs977289080 Chr.3: 122889558-122889558 on Build GRCh38
rs745941605 Chr.3: 122889536-122889536 on Build GRCh38
rs147893504 Chr.3: 122889400-122889400 on Build GRCh38
rs894357605 Chr.3: 122889664-122889664 on Build GRCh38
rs994260550 Chr.3: 122889408-122889408 on Build GRCh38
rs1002489282 Chr.3: 122889466-122889466 on Build GRCh38
rs1040488120 Chr.3: 122889410-122889410 on Build GRCh38
rs749269608 Chr.3: 122889407-122889407 on Build GRCh38
rs1055284627 Chr.3: 122889480-122889480 on Build GRCh38
rs925639849 Chr.3: 122889631-122889631 on Build GRCh38
rs779151283 Chr.3: 122889466-122889466 on Build GRCh38
rs985373327 Chr.3: 122889574-122889574 on Build GRCh38
rs1025623110 Chr.3: 122889416-122889416 on Build GRCh38
rs964244235 Chr.3: 122889539-122889539 on Build GRCh38
rs111673104 Chr.3: 122889837-122889837 on Build GRCh38
rs1020673684 Chr.3: 122889498-122889498 on Build GRCh38
rs28397570 Chr.3: 122889497-122889497 on Build GRCh38
rs888025376 Chr.3: 122889446-122889446 on Build GRCh38
rs935625560 Chr.3: 122889654-122889654 on Build GRCh38
rs919355736 Chr.3: 122889435-122889435 on Build GRCh38
rs769258999 Chr.3: 122889867-122889867 on Build GRCh38
rs558358973 Chr.3: 122889657-122889657 on Build GRCh38
rs953957296 Chr.3: 122889548-122889548 on Build GRCh38
rs964681525 Chr.3: 122889868-122889868 on Build GRCh38
rs139985329 Chr.3: 122889770-122889770 on Build GRCh38
rs1046015170 Chr.3: 122889743-122889743 on Build GRCh38
rs896673545 Chr.3: 122889491-122889491 on Build GRCh38
rs370959365 Chr.3: 122889527-122889527 on Build GRCh38
rs185028695 Chr.3: 122889810-122889810 on Build GRCh38
rs977031183 Chr.3: 122889872-122889872 on Build GRCh38
rs747570595 Chr.3: 122889799-122889799 on Build GRCh38
rs949545355 Chr.3: 122889866-122889866 on Build GRCh38
rs984348870 Chr.3: 122889531-122889531 on Build GRCh38
rs1055358348 Chr.3: 122889808-122889808 on Build GRCh38
rs938223413 Chr.3: 122889786-122889786 on Build GRCh38
rs1030269264 Chr.3: 122889877-122889877 on Build GRCh38

View Full Product Details