Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99133451-99133916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH4
Entrez Gene ID
127
Gene Name
alcohol dehydrogenase 4 (class II), pi polypeptide
Gene Aliases
ADH-2, HEL-S-4
UniGene
Hs.1219
Gene Chromosome Location
Chr.4: 99123657-99144330 on Build GRCh38
Transcripts
NM_000670.4 NM_001306171.1 NM_001306172.1 XM_017007713.1 XM_017007714.1 XM_017007715.1 NR_037884.1

Gene Symbol
LOC100507053
Entrez Gene ID
100507053
Gene Name
uncharacterized LOC100507053
Gene Aliases
-
UniGene
Hs.635084
Gene Chromosome Location
Chr.4: 99088857-99301356 on Build GRCh38
Transcripts
NM_000670.4 NM_001306171.1 NM_001306172.1 XM_017007713.1 XM_017007714.1 XM_017007715.1 NR_037884.1

SNP Detail

SNP ID Chromosome Location
rs796100782 Chr.4: 99133640-99133640 on Build GRCh38
rs951341126 Chr.4: 99133794-99133794 on Build GRCh38
rs912395528 Chr.4: 99133508-99133508 on Build GRCh38
rs17218003 Chr.4: 99133572-99133572 on Build GRCh38
rs117189719 Chr.4: 99133571-99133571 on Build GRCh38
rs1054055033 Chr.4: 99133579-99133579 on Build GRCh38
rs29001193 Chr.4: 99133757-99133757 on Build GRCh38
rs975543577 Chr.4: 99133873-99133873 on Build GRCh38
rs955975965 Chr.4: 99133822-99133822 on Build GRCh38
rs1011161570 Chr.4: 99133837-99133837 on Build GRCh38
rs1019413927 Chr.4: 99133828-99133828 on Build GRCh38
rs1018176719 Chr.4: 99133882-99133882 on Build GRCh38
rs29001194 Chr.4: 99133693-99133693 on Build GRCh38
rs556755117 Chr.4: 99133646-99133646 on Build GRCh38
rs894158578 Chr.4: 99133750-99133750 on Build GRCh38
rs13143133 Chr.4: 99133819-99133819 on Build GRCh38
rs29001191 Chr.4: 99133875-99133875 on Build GRCh38
rs929267899 Chr.4: 99133673-99133673 on Build GRCh38
rs29001192 Chr.4: 99133860-99133860 on Build GRCh38
rs965468982 Chr.4: 99133511-99133511 on Build GRCh38
rs985259643 Chr.4: 99133878-99133878 on Build GRCh38
rs995610798 Chr.4: 99133759-99133759 on Build GRCh38
rs6532798 Chr.4: 99133676-99133676 on Build GRCh38
rs975869917 Chr.4: 99133529-99133529 on Build GRCh38
rs906689956 Chr.4: 99133689-99133689 on Build GRCh38
rs1055911119 Chr.4: 99133722-99133722 on Build GRCh38
rs542958248 Chr.4: 99133645-99133645 on Build GRCh38
rs1003192407 Chr.4: 99133802-99133802 on Build GRCh38
rs566586632 Chr.4: 99133484-99133484 on Build GRCh38
rs4148890 Chr.4: 99133638-99133638 on Build GRCh38
rs921279316 Chr.4: 99133535-99133535 on Build GRCh38
rs982605393 Chr.4: 99133487-99133487 on Build GRCh38
rs759745002 Chr.4: 99133769-99133769 on Build GRCh38
rs989841171 Chr.4: 99133498-99133498 on Build GRCh38
rs1044618758 Chr.4: 99133758-99133758 on Build GRCh38
rs907071210 Chr.4: 99133771-99133771 on Build GRCh38
rs533541664 Chr.4: 99133559-99133559 on Build GRCh38
rs749325095 Chr.4: 99133638-99133638 on Build GRCh38

View Full Product Details