Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149425443-149425988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARMN
Entrez Gene ID
728264
Gene Name
cardiac mesoderm enhancer-associated non-coding RNA
Gene Aliases
CARMEN, MIR143HG
UniGene
Hs.519666
Gene Chromosome Location
Chr.5: 149406845-149432836 on Build GRCh38
Transcripts
NR_105059.1 NR_105060.1

SNP Detail

SNP ID Chromosome Location
rs773215237 Chr.5: 149425750-149425750 on Build GRCh38
rs948546923 Chr.5: 149425732-149425732 on Build GRCh38
rs544995730 Chr.5: 149425866-149425866 on Build GRCh38
rs1040583545 Chr.5: 149425734-149425734 on Build GRCh38
rs553277845 Chr.5: 149425793-149425793 on Build GRCh38
rs534594044 Chr.5: 149425762-149425762 on Build GRCh38
rs979781062 Chr.5: 149425598-149425598 on Build GRCh38
rs1047423799 Chr.5: 149425791-149425791 on Build GRCh38
rs900233809 Chr.5: 149425807-149425807 on Build GRCh38
rs138365224 Chr.5: 149425862-149425862 on Build GRCh38
rs563201430 Chr.5: 149425877-149425877 on Build GRCh38
rs1028588943 Chr.5: 149425513-149425513 on Build GRCh38
rs758669208 Chr.5: 149425554-149425554 on Build GRCh38
rs984538057 Chr.5: 149425679-149425679 on Build GRCh38
rs542775747 Chr.5: 149425953-149425953 on Build GRCh38
rs183017248 Chr.5: 149425942-149425942 on Build GRCh38
rs187544379 Chr.5: 149425561-149425561 on Build GRCh38
rs990233614 Chr.5: 149425691-149425691 on Build GRCh38
rs142840162 Chr.5: 149425946-149425946 on Build GRCh38
rs938584336 Chr.5: 149425755-149425755 on Build GRCh38
rs748432101 Chr.5: 149425543-149425543 on Build GRCh38
rs910633623 Chr.5: 149425703-149425703 on Build GRCh38
rs1034056858 Chr.5: 149425923-149425923 on Build GRCh38
rs956031746 Chr.5: 149425917-149425917 on Build GRCh38
rs1000275419 Chr.5: 149425800-149425800 on Build GRCh38
rs760778372 Chr.5: 149425794-149425794 on Build GRCh38
rs374428176 Chr.5: 149425676-149425676 on Build GRCh38
rs924595788 Chr.5: 149425665-149425665 on Build GRCh38
rs889775908 Chr.5: 149425797-149425797 on Build GRCh38
rs190535039 Chr.5: 149425895-149425895 on Build GRCh38
rs1028256469 Chr.5: 149425486-149425486 on Build GRCh38
rs878008 Chr.5: 149425488-149425488 on Build GRCh38
rs1033173426 Chr.5: 149425511-149425511 on Build GRCh38
rs74518737 Chr.5: 149425469-149425469 on Build GRCh38
rs75777785 Chr.5: 149425475-149425475 on Build GRCh38
rs955791717 Chr.5: 149425539-149425539 on Build GRCh38
rs955680221 Chr.5: 149425855-149425855 on Build GRCh38
rs997044480 Chr.5: 149425820-149425820 on Build GRCh38
rs891838044 Chr.5: 149425812-149425812 on Build GRCh38
rs914624560 Chr.5: 149425705-149425705 on Build GRCh38
rs956229666 Chr.5: 149425527-149425527 on Build GRCh38
rs972025747 Chr.5: 149425563-149425563 on Build GRCh38
rs940544594 Chr.5: 149425724-149425724 on Build GRCh38
rs772283228 Chr.5: 149425741-149425741 on Build GRCh38
rs1033098940 Chr.5: 149425806-149425806 on Build GRCh38
rs1028893791 Chr.5: 149425836-149425836 on Build GRCh38
rs971704567 Chr.5: 149425867-149425867 on Build GRCh38
rs1000223068 Chr.5: 149425491-149425491 on Build GRCh38
rs907578972 Chr.5: 149425704-149425704 on Build GRCh38
rs926935571 Chr.5: 149425619-149425619 on Build GRCh38
rs917513586 Chr.5: 149425558-149425558 on Build GRCh38
rs1010414969 Chr.5: 149425821-149425821 on Build GRCh38

View Full Product Details