Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149429189-149429735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARMN
Entrez Gene ID
728264
Gene Name
cardiac mesoderm enhancer-associated non-coding RNA
Gene Aliases
CARMEN, MIR143HG
UniGene
Hs.519666
Gene Chromosome Location
Chr.5: 149406845-149432836 on Build GRCh38
Transcripts
NR_105059.1 NR_105060.1

SNP Detail

SNP ID Chromosome Location
rs895229002 Chr.5: 149429650-149429650 on Build GRCh38
rs397817694 Chr.5: 149429322-149429323 on Build GRCh38
rs1007923471 Chr.5: 149429477-149429477 on Build GRCh38
rs992248926 Chr.5: 149429582-149429582 on Build GRCh38
rs963846195 Chr.5: 149429495-149429495 on Build GRCh38
rs745461431 Chr.5: 149429657-149429657 on Build GRCh38
rs962695582 Chr.5: 149429228-149429228 on Build GRCh38
rs558474362 Chr.5: 149429308-149429308 on Build GRCh38
rs772549058 Chr.5: 149429601-149429601 on Build GRCh38
rs951111725 Chr.5: 149429245-149429245 on Build GRCh38
rs948163450 Chr.5: 149429419-149429419 on Build GRCh38
rs1025412052 Chr.5: 149429549-149429549 on Build GRCh38
rs997100423 Chr.5: 149429700-149429700 on Build GRCh38
rs756258352 Chr.5: 149429415-149429415 on Build GRCh38
rs547505654 Chr.5: 149429689-149429689 on Build GRCh38
rs1045733672 Chr.5: 149429431-149429431 on Build GRCh38
rs397811827 Chr.5: 149429215-149429216 on Build GRCh38
rs902707702 Chr.5: 149429467-149429467 on Build GRCh38
rs1025829057 Chr.5: 149429244-149429244 on Build GRCh38
rs34290100 Chr.5: 149429312-149429313 on Build GRCh38
rs914453999 Chr.5: 149429294-149429294 on Build GRCh38
rs36093629 Chr.5: 149429322-149429322 on Build GRCh38
rs541539542 Chr.5: 149429512-149429512 on Build GRCh38
rs561418751 Chr.5: 149429652-149429652 on Build GRCh38
rs140713581 Chr.5: 149429492-149429492 on Build GRCh38
rs764042524 Chr.5: 149429278-149429278 on Build GRCh38
rs77228332 Chr.5: 149429265-149429265 on Build GRCh38
rs991115852 Chr.5: 149429236-149429236 on Build GRCh38
rs376057792 Chr.5: 149429653-149429653 on Build GRCh38
rs547695078 Chr.5: 149429263-149429263 on Build GRCh38
rs543032245 Chr.5: 149429644-149429644 on Build GRCh38
rs780707022 Chr.5: 149429613-149429613 on Build GRCh38
rs528934216 Chr.5: 149429682-149429682 on Build GRCh38
rs553668742 Chr.5: 149429527-149429527 on Build GRCh38
rs753976434 Chr.5: 149429470-149429470 on Build GRCh38
rs1025290039 Chr.5: 149429454-149429454 on Build GRCh38
rs935391626 Chr.5: 149429387-149429387 on Build GRCh38
rs1007810229 Chr.5: 149429654-149429654 on Build GRCh38
rs910853329 Chr.5: 149429310-149429310 on Build GRCh38
rs946713857 Chr.5: 149429295-149429295 on Build GRCh38
rs896655405 Chr.5: 149429414-149429414 on Build GRCh38
rs910978626 Chr.5: 149429592-149429592 on Build GRCh38
rs970609483 Chr.5: 149429506-149429506 on Build GRCh38
rs531170348 Chr.5: 149429602-149429602 on Build GRCh38
rs574201343 Chr.5: 149429496-149429496 on Build GRCh38
rs1032441568 Chr.5: 149429486-149429486 on Build GRCh38
rs762819853 Chr.5: 149429269-149429269 on Build GRCh38
rs763038908 Chr.5: 149429646-149429646 on Build GRCh38
rs975298285 Chr.5: 149429288-149429288 on Build GRCh38
rs755490529 Chr.5: 149429585-149429585 on Build GRCh38
rs572242941 Chr.5: 149429551-149429551 on Build GRCh38
rs970495005 Chr.5: 149429262-149429262 on Build GRCh38
rs1019327636 Chr.5: 149429655-149429655 on Build GRCh38
rs886772459 Chr.5: 149429648-149429648 on Build GRCh38
rs770320628 Chr.5: 149429594-149429594 on Build GRCh38
rs968639340 Chr.5: 149429498-149429498 on Build GRCh38
rs533535993 Chr.5: 149429383-149429383 on Build GRCh38
rs570017044 Chr.5: 149429356-149429356 on Build GRCh38
rs113013728 Chr.5: 149429614-149429614 on Build GRCh38
rs566291664 Chr.5: 149429264-149429264 on Build GRCh38
rs35551363 Chr.5: 149429411-149429412 on Build GRCh38
rs187827091 Chr.5: 149429401-149429401 on Build GRCh38
rs55945735 Chr.5: 149429595-149429595 on Build GRCh38
rs376971648 Chr.5: 149429230-149429230 on Build GRCh38
rs984246166 Chr.5: 149429565-149429565 on Build GRCh38
rs1021972225 Chr.5: 149429497-149429497 on Build GRCh38
rs899561567 Chr.5: 149429661-149429661 on Build GRCh38
rs936863471 Chr.5: 149429640-149429640 on Build GRCh38
rs983909494 Chr.5: 149429306-149429306 on Build GRCh38
rs564412086 Chr.5: 149429596-149429596 on Build GRCh38
rs751274194 Chr.5: 149429329-149429329 on Build GRCh38
rs767610412 Chr.5: 149429359-149429359 on Build GRCh38
rs922292776 Chr.5: 149429597-149429597 on Build GRCh38
rs59810862 Chr.5: 149429214-149429215 on Build GRCh38
rs935444010 Chr.5: 149429557-149429557 on Build GRCh38
rs898719507 Chr.5: 149429447-149429447 on Build GRCh38
rs369052151 Chr.5: 149429453-149429453 on Build GRCh38

View Full Product Details