Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159452474-159452982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC285627
Entrez Gene ID
285627
Gene Name
uncharacterized LOC285627
Gene Aliases
-
UniGene
Hs.552766
Gene Chromosome Location
Chr.5: 159448556-159466276 on Build GRCh38
Transcripts
NR_027110.1

SNP Detail

SNP ID Chromosome Location
rs537197172 Chr.5: 159452697-159452697 on Build GRCh38
rs924232316 Chr.5: 159452838-159452838 on Build GRCh38
rs535738216 Chr.5: 159452730-159452730 on Build GRCh38
rs925946678 Chr.5: 159452755-159452755 on Build GRCh38
rs111849868 Chr.5: 159452525-159452525 on Build GRCh38
rs867709245 Chr.5: 159452845-159452845 on Build GRCh38
rs549704458 Chr.5: 159452532-159452532 on Build GRCh38
rs1005204152 Chr.5: 159452660-159452660 on Build GRCh38
rs766525008 Chr.5: 159452945-159452945 on Build GRCh38
rs529130917 Chr.5: 159452548-159452548 on Build GRCh38
rs576587345 Chr.5: 159452723-159452723 on Build GRCh38
rs370523872 Chr.5: 159452943-159452943 on Build GRCh38
rs944753425 Chr.5: 159452530-159452530 on Build GRCh38
rs555574343 Chr.5: 159452871-159452871 on Build GRCh38
rs147138709 Chr.5: 159452814-159452814 on Build GRCh38
rs909019095 Chr.5: 159452938-159452938 on Build GRCh38
rs114452418 Chr.5: 159452960-159452960 on Build GRCh38
rs769769396 Chr.5: 159452703-159452703 on Build GRCh38
rs957566732 Chr.5: 159452768-159452768 on Build GRCh38
rs192964523 Chr.5: 159452545-159452545 on Build GRCh38
rs375950343 Chr.5: 159452939-159452939 on Build GRCh38
rs1049172149 Chr.5: 159452908-159452908 on Build GRCh38
rs915457231 Chr.5: 159452515-159452515 on Build GRCh38
rs978737812 Chr.5: 159452829-159452829 on Build GRCh38
rs1024122134 Chr.5: 159452685-159452685 on Build GRCh38
rs7702827 Chr.5: 159452861-159452861 on Build GRCh38
rs989933121 Chr.5: 159452769-159452769 on Build GRCh38
rs188609495 Chr.5: 159452623-159452623 on Build GRCh38
rs900615146 Chr.5: 159452572-159452572 on Build GRCh38
rs946950964 Chr.5: 159452516-159452516 on Build GRCh38
rs934102278 Chr.5: 159452840-159452840 on Build GRCh38
rs185095987 Chr.5: 159452777-159452777 on Build GRCh38
rs958446498 Chr.5: 159452767-159452767 on Build GRCh38
rs149467426 Chr.5: 159452698-159452698 on Build GRCh38
rs986950734 Chr.5: 159452841-159452841 on Build GRCh38
rs1014865484 Chr.5: 159452692-159452692 on Build GRCh38
rs377537189 Chr.5: 159452717-159452717 on Build GRCh38
rs906849787 Chr.5: 159452700-159452700 on Build GRCh38
rs142847278 Chr.5: 159452961-159452961 on Build GRCh38
rs994522118 Chr.5: 159452550-159452550 on Build GRCh38
rs1014475864 Chr.5: 159452683-159452683 on Build GRCh38
rs150316601 Chr.5: 159452596-159452596 on Build GRCh38
rs565649090 Chr.5: 159452611-159452611 on Build GRCh38
rs534576908 Chr.5: 159452612-159452612 on Build GRCh38
rs533284741 Chr.5: 159452524-159452524 on Build GRCh38
rs958501981 Chr.5: 159452644-159452644 on Build GRCh38
rs181058534 Chr.5: 159452749-159452749 on Build GRCh38
rs1004493855 Chr.5: 159452521-159452521 on Build GRCh38
rs921323357 Chr.5: 159452782-159452782 on Build GRCh38
rs140648435 Chr.5: 159452959-159452959 on Build GRCh38
rs1036659909 Chr.5: 159452944-159452944 on Build GRCh38
rs370010501 Chr.5: 159452848-159452848 on Build GRCh38
rs1048887741 Chr.5: 159452591-159452591 on Build GRCh38
rs564180373 Chr.5: 159452834-159452834 on Build GRCh38
rs137924668 Chr.5: 159452624-159452624 on Build GRCh38
rs931779326 Chr.5: 159452587-159452587 on Build GRCh38
rs763023548 Chr.5: 159452903-159452903 on Build GRCh38
rs184240143 Chr.5: 159452571-159452571 on Build GRCh38

View Full Product Details