Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31173586-31174092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSORS1C3
Entrez Gene ID
100130889
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
Gene Aliases
NCRNA00196
UniGene
Hs.670091
Gene Chromosome Location
Chr.6: 31173735-31177899 on Build GRCh38
Transcripts
NR_026816.1

SNP Detail

SNP ID Chromosome Location
rs950118788 Chr.6: 31173704-31173704 on Build GRCh38
rs926578164 Chr.6: 31173778-31173778 on Build GRCh38
rs72856757 Chr.6: 31173894-31173894 on Build GRCh38
rs887468 Chr.6: 31173746-31173746 on Build GRCh38
rs959409238 Chr.6: 31173800-31173800 on Build GRCh38
rs531278882 Chr.6: 31174028-31174028 on Build GRCh38
rs530188510 Chr.6: 31173897-31173897 on Build GRCh38
rs372829560 Chr.6: 31173901-31173901 on Build GRCh38
rs915345089 Chr.6: 31173824-31173824 on Build GRCh38
rs991450876 Chr.6: 31173972-31173972 on Build GRCh38
rs961928800 Chr.6: 31173904-31173904 on Build GRCh38
rs565710835 Chr.6: 31173649-31173649 on Build GRCh38
rs893452813 Chr.6: 31173787-31173787 on Build GRCh38
rs958956301 Chr.6: 31173969-31173969 on Build GRCh38
rs564882791 Chr.6: 31173700-31173700 on Build GRCh38
rs1004670539 Chr.6: 31173987-31173987 on Build GRCh38
rs971259815 Chr.6: 31173992-31173992 on Build GRCh38
rs989496443 Chr.6: 31173807-31173807 on Build GRCh38
rs115568381 Chr.6: 31173612-31173612 on Build GRCh38
rs138004452 Chr.6: 31173789-31173789 on Build GRCh38
rs11507949 Chr.6: 31173899-31173899 on Build GRCh38
rs945492220 Chr.6: 31173826-31173826 on Build GRCh38
rs576903104 Chr.6: 31173861-31173861 on Build GRCh38
rs766271573 Chr.6: 31173741-31173741 on Build GRCh38
rs887467 Chr.6: 31173887-31173887 on Build GRCh38
rs890169737 Chr.6: 31173871-31173871 on Build GRCh38
rs886178082 Chr.6: 31173986-31173986 on Build GRCh38
rs2269711 Chr.6: 31173835-31173835 on Build GRCh38
rs749569311 Chr.6: 31173993-31173993 on Build GRCh38
rs3132517 Chr.6: 31174059-31174059 on Build GRCh38
rs1061503 Chr.6: 31173765-31173765 on Build GRCh38
rs995149772 Chr.6: 31173949-31173949 on Build GRCh38
rs73728280 Chr.6: 31174050-31174050 on Build GRCh38
rs532344910 Chr.6: 31173703-31173703 on Build GRCh38
rs867625605 Chr.6: 31173641-31173641 on Build GRCh38
rs754336091 Chr.6: 31174021-31174021 on Build GRCh38
rs563818545 Chr.6: 31173991-31173991 on Build GRCh38
rs11507948 Chr.6: 31173902-31173902 on Build GRCh38
rs773046219 Chr.6: 31173692-31173692 on Build GRCh38
rs553989344 Chr.6: 31173647-31173647 on Build GRCh38
rs1048782212 Chr.6: 31173944-31173944 on Build GRCh38
rs994530116 Chr.6: 31173911-31173911 on Build GRCh38
rs777292407 Chr.6: 31173966-31173966 on Build GRCh38
rs980084291 Chr.6: 31173758-31173758 on Build GRCh38
rs751401965 Chr.6: 31173744-31173744 on Build GRCh38
rs912101156 Chr.6: 31173859-31173859 on Build GRCh38
rs116763857 Chr.6: 31173705-31173705 on Build GRCh38
rs536465428 Chr.6: 31173668-31173668 on Build GRCh38

View Full Product Details