Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35735303-35735805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC12
Entrez Gene ID
221481
Gene Name
armadillo repeat containing 12
Gene Aliases
C6orf81
UniGene
Hs.533066
Gene Chromosome Location
Chr.6: 35733726-35748913 on Build GRCh38
Transcripts
XM_011514381.2 XM_017010435.1 NR_027117.2

Gene Symbol
LOC285847
Entrez Gene ID
285847
Gene Name
uncharacterized LOC285847
Gene Aliases
-
UniGene
Hs.407190
Gene Chromosome Location
Chr.6: 35726762-35736947 on Build GRCh38
Transcripts
XM_011514381.2 XM_017010435.1 NR_027117.2

SNP Detail

SNP ID Chromosome Location
rs922328875 Chr.6: 35735395-35735395 on Build GRCh38
rs929746727 Chr.6: 35735418-35735418 on Build GRCh38
rs540942280 Chr.6: 35735505-35735505 on Build GRCh38
rs561695563 Chr.6: 35735471-35735471 on Build GRCh38
rs1010323128 Chr.6: 35735751-35735751 on Build GRCh38
rs77821345 Chr.6: 35735538-35735538 on Build GRCh38
rs558498027 Chr.6: 35735326-35735326 on Build GRCh38
rs548121144 Chr.6: 35735686-35735686 on Build GRCh38
rs576600555 Chr.6: 35735428-35735428 on Build GRCh38
rs922364729 Chr.6: 35735495-35735495 on Build GRCh38
rs993256355 Chr.6: 35735426-35735426 on Build GRCh38
rs917336973 Chr.6: 35735635-35735635 on Build GRCh38
rs141784256 Chr.6: 35735721-35735721 on Build GRCh38
rs139559685 Chr.6: 35735746-35735746 on Build GRCh38
rs976451589 Chr.6: 35735378-35735378 on Build GRCh38
rs897676536 Chr.6: 35735419-35735419 on Build GRCh38
rs751775215 Chr.6: 35735642-35735642 on Build GRCh38
rs565503787 Chr.6: 35735606-35735606 on Build GRCh38
rs558590649 Chr.6: 35735781-35735781 on Build GRCh38
rs942156295 Chr.6: 35735712-35735712 on Build GRCh38
rs982530081 Chr.6: 35735657-35735657 on Build GRCh38
rs1047868170 Chr.6: 35735465-35735465 on Build GRCh38
rs549557811 Chr.6: 35735616-35735617 on Build GRCh38
rs370783966 Chr.6: 35735718-35735718 on Build GRCh38
rs904226827 Chr.6: 35735776-35735778 on Build GRCh38
rs1051471473 Chr.6: 35735755-35735755 on Build GRCh38
rs546437122 Chr.6: 35735778-35735778 on Build GRCh38
rs1020118983 Chr.6: 35735546-35735546 on Build GRCh38
rs185003931 Chr.6: 35735506-35735506 on Build GRCh38
rs150572115 Chr.6: 35735722-35735722 on Build GRCh38
rs116307714 Chr.6: 35735605-35735605 on Build GRCh38
rs968995857 Chr.6: 35735553-35735553 on Build GRCh38
rs761986582 Chr.6: 35735726-35735726 on Build GRCh38
rs532963444 Chr.6: 35735773-35735773 on Build GRCh38
rs1037526830 Chr.6: 35735409-35735409 on Build GRCh38
rs1010352572 Chr.6: 35735539-35735539 on Build GRCh38
rs747831078 Chr.6: 35735749-35735749 on Build GRCh38
rs1017263877 Chr.6: 35735775-35735775 on Build GRCh38
rs58429232 Chr.6: 35735583-35735583 on Build GRCh38
rs529642663 Chr.6: 35735534-35735534 on Build GRCh38
rs886638339 Chr.6: 35735470-35735470 on Build GRCh38
rs532720817 Chr.6: 35735658-35735658 on Build GRCh38
rs1001908287 Chr.6: 35735525-35735525 on Build GRCh38
rs1018574383 Chr.6: 35735574-35735574 on Build GRCh38
rs541505802 Chr.6: 35735536-35735536 on Build GRCh38
rs570017562 Chr.6: 35735731-35735731 on Build GRCh38
rs890226269 Chr.6: 35735763-35735763 on Build GRCh38
rs758539555 Chr.6: 35735558-35735558 on Build GRCh38

View Full Product Details