Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160005213-160005713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIRN
Entrez Gene ID
100271873
Gene Name
antisense of IGF2R non-protein coding RNA
Gene Aliases
AIR, IGF2R-AS, IGF2R-AS1, IGF2RAS, NCRNA00088
UniGene
Hs.742166
Gene Chromosome Location
Chr.6: 160003291-160007664 on Build GRCh38
Transcripts
NR_047511.1 NR_047514.1 NM_000876.3

Gene Symbol
IGF2R
Entrez Gene ID
3482
Gene Name
insulin like growth factor 2 receptor
Gene Aliases
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
UniGene
Hs.487062
Gene Chromosome Location
Chr.6: 159969099-160111504 on Build GRCh38
Transcripts
NR_047511.1 NR_047514.1 NM_000876.3

SNP Detail

SNP ID Chromosome Location
rs974338038 Chr.6: 160005478-160005478 on Build GRCh38
rs558673496 Chr.6: 160005634-160005634 on Build GRCh38
rs1046299629 Chr.6: 160005348-160005348 on Build GRCh38
rs902480918 Chr.6: 160005264-160005264 on Build GRCh38
rs890016094 Chr.6: 160005451-160005451 on Build GRCh38
rs890917102 Chr.6: 160005566-160005566 on Build GRCh38
rs937852516 Chr.6: 160005417-160005417 on Build GRCh38
rs1042383676 Chr.6: 160005263-160005263 on Build GRCh38
rs962435038 Chr.6: 160005610-160005610 on Build GRCh38
rs182471731 Chr.6: 160005551-160005551 on Build GRCh38
rs8191720 Chr.6: 160005401-160005401 on Build GRCh38
rs552904848 Chr.6: 160005531-160005531 on Build GRCh38
rs942962372 Chr.6: 160005658-160005658 on Build GRCh38
rs908203575 Chr.6: 160005272-160005272 on Build GRCh38
rs677882 Chr.6: 160005424-160005424 on Build GRCh38
rs1038999721 Chr.6: 160005548-160005548 on Build GRCh38
rs934770332 Chr.6: 160005521-160005521 on Build GRCh38
rs979234794 Chr.6: 160005405-160005427 on Build GRCh38
rs1051378530 Chr.6: 160005450-160005450 on Build GRCh38
rs994714172 Chr.6: 160005685-160005685 on Build GRCh38
rs1038549425 Chr.6: 160005659-160005659 on Build GRCh38
rs561285112 Chr.6: 160005269-160005269 on Build GRCh38
rs962341432 Chr.6: 160005454-160005454 on Build GRCh38
rs978215610 Chr.6: 160005572-160005572 on Build GRCh38
rs950394179 Chr.6: 160005384-160005384 on Build GRCh38
rs34753168 Chr.6: 160005617-160005617 on Build GRCh38
rs542362286 Chr.6: 160005689-160005689 on Build GRCh38
rs865778910 Chr.6: 160005410-160005410 on Build GRCh38
rs1010739569 Chr.6: 160005244-160005244 on Build GRCh38
rs994598896 Chr.6: 160005486-160005486 on Build GRCh38
rs567460327 Chr.6: 160005459-160005459 on Build GRCh38
rs1025987244 Chr.6: 160005379-160005379 on Build GRCh38
rs1029498072 Chr.6: 160005560-160005560 on Build GRCh38
rs959268347 Chr.6: 160005527-160005527 on Build GRCh38
rs990650700 Chr.6: 160005418-160005418 on Build GRCh38
rs933957729 Chr.6: 160005442-160005442 on Build GRCh38
rs1038491657 Chr.6: 160005482-160005482 on Build GRCh38
rs546882038 Chr.6: 160005354-160005354 on Build GRCh38
rs8191721 Chr.6: 160005404-160005404 on Build GRCh38
rs1030305413 Chr.6: 160005303-160005303 on Build GRCh38
rs934029247 Chr.6: 160005593-160005593 on Build GRCh38
rs376596080 Chr.6: 160005547-160005547 on Build GRCh38
rs868838230 Chr.6: 160005324-160005324 on Build GRCh38
rs1022145704 Chr.6: 160005411-160005411 on Build GRCh38
rs916093703 Chr.6: 160005301-160005301 on Build GRCh38
rs8191722 Chr.6: 160005427-160005427 on Build GRCh38
rs189755865 Chr.6: 160005244-160005244 on Build GRCh38
rs967802369 Chr.6: 160005280-160005294 on Build GRCh38
rs893157867 Chr.6: 160005545-160005545 on Build GRCh38
rs915082791 Chr.6: 160005550-160005550 on Build GRCh38
rs1026458756 Chr.6: 160005495-160005495 on Build GRCh38
rs982505211 Chr.6: 160005266-160005266 on Build GRCh38
rs1017329461 Chr.6: 160005353-160005353 on Build GRCh38
rs889964287 Chr.6: 160005314-160005314 on Build GRCh38
rs772647876 Chr.6: 160005490-160005490 on Build GRCh38
rs947416579 Chr.6: 160005548-160005548 on Build GRCh38
rs999337144 Chr.6: 160005385-160005385 on Build GRCh38
rs1051529807 Chr.6: 160005600-160005600 on Build GRCh38
rs973762644 Chr.6: 160005620-160005620 on Build GRCh38
rs575324554 Chr.6: 160005663-160005663 on Build GRCh38
rs941594041 Chr.6: 160005278-160005278 on Build GRCh38
rs530377993 Chr.6: 160005287-160005287 on Build GRCh38
rs73021677 Chr.6: 160005590-160005590 on Build GRCh38
rs973512420 Chr.6: 160005365-160005365 on Build GRCh38
rs975588522 Chr.6: 160005259-160005259 on Build GRCh38
rs551536491 Chr.6: 160005421-160005421 on Build GRCh38
rs536516445 Chr.6: 160005496-160005496 on Build GRCh38
rs768892523 Chr.6: 160005234-160005234 on Build GRCh38
rs373592732 Chr.6: 160005530-160005530 on Build GRCh38
rs571345941 Chr.6: 160005422-160005422 on Build GRCh38
rs1015327626 Chr.6: 160005594-160005594 on Build GRCh38
rs898685519 Chr.6: 160005667-160005667 on Build GRCh38
rs1034974981 Chr.6: 160005515-160005515 on Build GRCh38
rs922401755 Chr.6: 160005260-160005260 on Build GRCh38
rs1023045621 Chr.6: 160005621-160005621 on Build GRCh38
rs9347379 Chr.6: 160005252-160005252 on Build GRCh38
rs898634703 Chr.6: 160005483-160005483 on Build GRCh38
rs1003603772 Chr.6: 160005514-160005514 on Build GRCh38

View Full Product Details