Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27151451-27151987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA-AS3
Entrez Gene ID
100133311
Gene Name
HOXA cluster antisense RNA 3
Gene Aliases
HOXA6as
UniGene
Hs.610216
Gene Chromosome Location
Chr.7: 27140364-27155928 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 XM_005249730.2 XM_006715715.2 XM_011515343.2

Gene Symbol
HOXA3
Entrez Gene ID
3200
Gene Name
homeobox A3
Gene Aliases
HOX1, HOX1E
UniGene
Hs.659337
Gene Chromosome Location
Chr.7: 27106190-27152581 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 XM_005249730.2 XM_006715715.2 XM_011515343.2

SNP Detail

SNP ID Chromosome Location
rs954963222 Chr.7: 27151719-27151719 on Build GRCh38
rs547411960 Chr.7: 27151726-27151726 on Build GRCh38
rs1043198281 Chr.7: 27151480-27151480 on Build GRCh38
rs1037382354 Chr.7: 27151967-27151967 on Build GRCh38
rs146655311 Chr.7: 27151825-27151827 on Build GRCh38
rs534923435 Chr.7: 27151514-27151514 on Build GRCh38
rs750094141 Chr.7: 27151522-27151522 on Build GRCh38
rs934682157 Chr.7: 27151547-27151547 on Build GRCh38
rs1006370483 Chr.7: 27151683-27151683 on Build GRCh38
rs949293403 Chr.7: 27151922-27151922 on Build GRCh38
rs1013814250 Chr.7: 27151596-27151596 on Build GRCh38
rs892397861 Chr.7: 27151574-27151574 on Build GRCh38
rs149404500 Chr.7: 27151701-27151701 on Build GRCh38
rs943984177 Chr.7: 27151576-27151576 on Build GRCh38
rs1046198724 Chr.7: 27151608-27151608 on Build GRCh38
rs777536914 Chr.7: 27151700-27151700 on Build GRCh38
rs905065404 Chr.7: 27151616-27151616 on Build GRCh38
rs958053844 Chr.7: 27151855-27151855 on Build GRCh38
rs1001904471 Chr.7: 27151625-27151625 on Build GRCh38
rs760434986 Chr.7: 27151571-27151571 on Build GRCh38
rs766264828 Chr.7: 27151607-27151607 on Build GRCh38
rs753705294 Chr.7: 27151638-27151638 on Build GRCh38
rs936476859 Chr.7: 27151515-27151515 on Build GRCh38
rs184790425 Chr.7: 27151835-27151835 on Build GRCh38
rs925940854 Chr.7: 27151516-27151516 on Build GRCh38
rs192017387 Chr.7: 27151832-27151832 on Build GRCh38
rs540558312 Chr.7: 27151682-27151682 on Build GRCh38
rs578057995 Chr.7: 27151654-27151654 on Build GRCh38
rs923307586 Chr.7: 27151619-27151620 on Build GRCh38
rs74747623 Chr.7: 27151948-27151948 on Build GRCh38
rs753016156 Chr.7: 27151628-27151628 on Build GRCh38
rs909283053 Chr.7: 27151953-27151953 on Build GRCh38
rs1017777956 Chr.7: 27151740-27151740 on Build GRCh38
rs80171741 Chr.7: 27151957-27151957 on Build GRCh38
rs926067363 Chr.7: 27151842-27151842 on Build GRCh38
rs752263842 Chr.7: 27151682-27151683 on Build GRCh38
rs369041286 Chr.7: 27151844-27151844 on Build GRCh38
rs934628178 Chr.7: 27151866-27151866 on Build GRCh38
rs917262710 Chr.7: 27151889-27151889 on Build GRCh38
rs746760026 Chr.7: 27151871-27151871 on Build GRCh38
rs564523936 Chr.7: 27151629-27151629 on Build GRCh38
rs35373589 Chr.7: 27151921-27151922 on Build GRCh38
rs551155215 Chr.7: 27151929-27151929 on Build GRCh38
rs577854126 Chr.7: 27151658-27151658 on Build GRCh38
rs146378191 Chr.7: 27151807-27151807 on Build GRCh38
rs1036966772 Chr.7: 27151698-27151698 on Build GRCh38
rs1040368874 Chr.7: 27151591-27151591 on Build GRCh38
rs138874296 Chr.7: 27151552-27151552 on Build GRCh38
rs768631776 Chr.7: 27151928-27151928 on Build GRCh38
rs780962204 Chr.7: 27151944-27151944 on Build GRCh38
rs887874891 Chr.7: 27151659-27151659 on Build GRCh38
rs1026958076 Chr.7: 27151703-27151703 on Build GRCh38
rs765093010 Chr.7: 27151646-27151646 on Build GRCh38
rs759462829 Chr.7: 27151644-27151644 on Build GRCh38
rs531457252 Chr.7: 27151921-27151921 on Build GRCh38
rs530714073 Chr.7: 27151828-27151828 on Build GRCh38
rs765156333 Chr.7: 27151656-27151657 on Build GRCh38
rs931941745 Chr.7: 27151946-27151946 on Build GRCh38
rs386711528 Chr.7: 27151825-27151828 on Build GRCh38
rs371969993 Chr.7: 27151963-27151963 on Build GRCh38

View Full Product Details