Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 93362422-93362931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00535
Entrez Gene ID
642924
Gene Name
long intergenic non-protein coding RNA 535
Gene Aliases
-
UniGene
Hs.434882
Gene Chromosome Location
Chr.8: 93346467-93700433 on Build GRCh38
Transcripts
NR_033858.1

SNP Detail

SNP ID Chromosome Location
rs925419924 Chr.8: 93362657-93362657 on Build GRCh38
rs10087128 Chr.8: 93362815-93362815 on Build GRCh38
rs999643208 Chr.8: 93362898-93362899 on Build GRCh38
rs1010462426 Chr.8: 93362899-93362899 on Build GRCh38
rs984384751 Chr.8: 93362717-93362717 on Build GRCh38
rs367742494 Chr.8: 93362655-93362655 on Build GRCh38
rs1004034743 Chr.8: 93362472-93362472 on Build GRCh38
rs907948967 Chr.8: 93362722-93362722 on Build GRCh38
rs573229742 Chr.8: 93362822-93362822 on Build GRCh38
rs1028693216 Chr.8: 93362682-93362682 on Build GRCh38
rs922214188 Chr.8: 93362675-93362675 on Build GRCh38
rs1051842756 Chr.8: 93362470-93362470 on Build GRCh38
rs571596456 Chr.8: 93362711-93362711 on Build GRCh38
rs927358193 Chr.8: 93362821-93362821 on Build GRCh38
rs576592997 Chr.8: 93362828-93362828 on Build GRCh38
rs966886118 Chr.8: 93362619-93362619 on Build GRCh38
rs371361895 Chr.8: 93362490-93362490 on Build GRCh38
rs976129520 Chr.8: 93362559-93362559 on Build GRCh38
rs532815732 Chr.8: 93362453-93362453 on Build GRCh38
rs891802403 Chr.8: 93362889-93362889 on Build GRCh38
rs966627136 Chr.8: 93362658-93362658 on Build GRCh38
rs748248738 Chr.8: 93362874-93362876 on Build GRCh38
rs892013398 Chr.8: 93362577-93362577 on Build GRCh38
rs905974408 Chr.8: 93362564-93362564 on Build GRCh38
rs1024625919 Chr.8: 93362860-93362860 on Build GRCh38
rs1037303335 Chr.8: 93362513-93362513 on Build GRCh38
rs144020718 Chr.8: 93362576-93362576 on Build GRCh38
rs529010845 Chr.8: 93362549-93362549 on Build GRCh38
rs1010910310 Chr.8: 93362621-93362621 on Build GRCh38
rs754829175 Chr.8: 93362670-93362670 on Build GRCh38
rs372997810 Chr.8: 93362534-93362534 on Build GRCh38
rs1000188382 Chr.8: 93362573-93362573 on Build GRCh38
rs114479997 Chr.8: 93362560-93362560 on Build GRCh38
rs187408659 Chr.8: 93362545-93362545 on Build GRCh38
rs75019067 Chr.8: 93362546-93362546 on Build GRCh38
rs898281051 Chr.8: 93362786-93362786 on Build GRCh38
rs192237976 Chr.8: 93362816-93362816 on Build GRCh38
rs907555136 Chr.8: 93362752-93362752 on Build GRCh38
rs548766502 Chr.8: 93362558-93362558 on Build GRCh38
rs2060091 Chr.8: 93362812-93362812 on Build GRCh38
rs902189379 Chr.8: 93362887-93362887 on Build GRCh38
rs917865895 Chr.8: 93362813-93362813 on Build GRCh38
rs533025914 Chr.8: 93362772-93362772 on Build GRCh38
rs958995808 Chr.8: 93362562-93362562 on Build GRCh38
rs1054421893 Chr.8: 93362877-93362877 on Build GRCh38
rs534207573 Chr.8: 93362756-93362756 on Build GRCh38
rs951840230 Chr.8: 93362697-93362697 on Build GRCh38
rs144060562 Chr.8: 93362899-93362899 on Build GRCh38

View Full Product Details