Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127448294-127448789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL12
Entrez Gene ID
6136
Gene Name
ribosomal protein L12
Gene Aliases
L12
UniGene
Hs.408054
Gene Chromosome Location
Chr.9: 127447674-127451432 on Build GRCh38
Transcripts
NM_000976.3 NR_002449.2

Gene Symbol
SNORA65
Entrez Gene ID
26783
Gene Name
small nucleolar RNA, H/ACA box 65
Gene Aliases
RNU65, U65
UniGene
Hs.408054
Gene Chromosome Location
Chr.9: 127448502-127448637 on Build GRCh38
Transcripts
NM_000976.3 NR_002449.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3664109 c.355C>G p.R119G Chr.9: 127448361-127448361 on Build GRCh38
6451853 c.349C>T p.R117W Chr.9: 127448367-127448367 on Build GRCh38
3325499 c.350G>A p.R117Q Chr.9: 127448366-127448366 on Build GRCh38
6336929 c.327T>C p.I109I Chr.9: 127448389-127448389 on Build GRCh38
1314467 c.375C>G p.L125L Chr.9: 127448341-127448341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554191130 Chr.9: 127448643-127448647 on Build GRCh38
rs941818816 Chr.9: 127448646-127448646 on Build GRCh38
rs776907957 Chr.9: 127448484-127448484 on Build GRCh38
rs377381430 Chr.9: 127448398-127448398 on Build GRCh38
rs756551267 Chr.9: 127448524-127448525 on Build GRCh38
rs747031609 Chr.9: 127448479-127448479 on Build GRCh38
rs1001119093 Chr.9: 127448517-127448517 on Build GRCh38
rs759877673 Chr.9: 127448434-127448434 on Build GRCh38
rs777619535 Chr.9: 127448592-127448592 on Build GRCh38
rs953993778 Chr.9: 127448524-127448524 on Build GRCh38
rs754019463 Chr.9: 127448432-127448432 on Build GRCh38
rs779702831 Chr.9: 127448452-127448452 on Build GRCh38
rs879958777 Chr.9: 127448378-127448378 on Build GRCh38
rs1000811215 Chr.9: 127448746-127448746 on Build GRCh38
rs763117672 Chr.9: 127448338-127448339 on Build GRCh38
rs760774103 Chr.9: 127448419-127448419 on Build GRCh38
rs780904603 Chr.9: 127448361-127448361 on Build GRCh38
rs772127641 Chr.9: 127448399-127448399 on Build GRCh38
rs777602924 Chr.9: 127448670-127448670 on Build GRCh38
rs775754923 Chr.9: 127448495-127448495 on Build GRCh38
rs773536765 Chr.9: 127448408-127448408 on Build GRCh38
rs201208415 Chr.9: 127448375-127448375 on Build GRCh38
rs902173975 Chr.9: 127448712-127448712 on Build GRCh38
rs763444755 Chr.9: 127448503-127448503 on Build GRCh38
rs559068212 Chr.9: 127448636-127448636 on Build GRCh38
rs2247573 Chr.9: 127448667-127448667 on Build GRCh38
rs759019940 Chr.9: 127448443-127448443 on Build GRCh38
rs11542425 Chr.9: 127448352-127448352 on Build GRCh38
rs762468316 Chr.9: 127448629-127448629 on Build GRCh38
rs774278549 Chr.9: 127448390-127448390 on Build GRCh38
rs1049202695 Chr.9: 127448402-127448402 on Build GRCh38
rs370429010 Chr.9: 127448614-127448614 on Build GRCh38
rs202110248 Chr.9: 127448631-127448631 on Build GRCh38
rs748902530 Chr.9: 127448455-127448455 on Build GRCh38
rs768099107 Chr.9: 127448453-127448456 on Build GRCh38
rs752129262 Chr.9: 127448512-127448512 on Build GRCh38
rs184123213 Chr.9: 127448367-127448367 on Build GRCh38
rs545496612 Chr.9: 127448627-127448627 on Build GRCh38
rs79093095 Chr.9: 127448617-127448617 on Build GRCh38
rs61734642 Chr.9: 127448358-127448358 on Build GRCh38
rs749065420 Chr.9: 127448376-127448376 on Build GRCh38
rs1042006103 Chr.9: 127448697-127448697 on Build GRCh38
rs753149477 Chr.9: 127448334-127448334 on Build GRCh38
rs11542422 Chr.9: 127448418-127448418 on Build GRCh38
rs753230100 Chr.9: 127448439-127448439 on Build GRCh38
rs1049760989 Chr.9: 127448658-127448658 on Build GRCh38
rs141909964 Chr.9: 127448366-127448366 on Build GRCh38
rs771001689 Chr.9: 127448480-127448480 on Build GRCh38
rs764200850 Chr.9: 127448398-127448399 on Build GRCh38
rs771025129 Chr.9: 127448603-127448603 on Build GRCh38
rs143393244 Chr.9: 127448478-127448478 on Build GRCh38
rs1055282142 Chr.9: 127448444-127448444 on Build GRCh38
rs763354581 Chr.9: 127448626-127448626 on Build GRCh38
rs138383878 Chr.9: 127448414-127448414 on Build GRCh38
rs751700511 Chr.9: 127448436-127448436 on Build GRCh38
rs555470116 Chr.9: 127448339-127448339 on Build GRCh38
rs766537993 Chr.9: 127448429-127448429 on Build GRCh38
rs1008321691 Chr.9: 127448320-127448320 on Build GRCh38
rs954691308 Chr.9: 127448319-127448319 on Build GRCh38
rs749915174 Chr.9: 127448450-127448450 on Build GRCh38
rs543292920 Chr.9: 127448447-127448447 on Build GRCh38
rs772350512 Chr.9: 127448389-127448389 on Build GRCh38
rs375784646 Chr.9: 127448605-127448605 on Build GRCh38
rs759730819 Chr.9: 127448492-127448492 on Build GRCh38
rs747015233 Chr.9: 127448595-127448595 on Build GRCh38
rs781218704 Chr.9: 127448681-127448681 on Build GRCh38
rs896190570 Chr.9: 127448466-127448466 on Build GRCh38
rs764737056 Chr.9: 127448642-127448642 on Build GRCh38
rs773331924 Chr.9: 127448415-127448415 on Build GRCh38
rs774998519 Chr.9: 127448628-127448628 on Build GRCh38
rs779559857 Chr.9: 127448538-127448540 on Build GRCh38
rs561502305 Chr.9: 127448676-127448676 on Build GRCh38
rs958246068 Chr.9: 127448664-127448664 on Build GRCh38
rs762086285 Chr.9: 127448381-127448381 on Build GRCh38
rs372553174 Chr.9: 127448518-127448518 on Build GRCh38
rs986269868 Chr.9: 127448584-127448584 on Build GRCh38
rs944684155 Chr.9: 127448363-127448363 on Build GRCh38
rs747887823 Chr.9: 127448573-127448573 on Build GRCh38
rs551259016 Chr.9: 127448761-127448761 on Build GRCh38
rs75030264 Chr.9: 127448683-127448683 on Build GRCh38
rs750721979 Chr.9: 127448521-127448522 on Build GRCh38
rs755320138 Chr.9: 127448329-127448329 on Build GRCh38
rs758201398 Chr.9: 127448582-127448582 on Build GRCh38
rs778739615 Chr.9: 127448565-127448565 on Build GRCh38
rs754549858 Chr.9: 127448641-127448641 on Build GRCh38
rs753556208 Chr.9: 127448549-127448550 on Build GRCh38
rs753516153 Chr.9: 127448537-127448537 on Build GRCh38
rs751473374 Chr.9: 127448673-127448673 on Build GRCh38
rs1040680896 Chr.9: 127448371-127448371 on Build GRCh38
rs765871710 Chr.9: 127448519-127448519 on Build GRCh38
rs898822162 Chr.9: 127448566-127448566 on Build GRCh38
rs778632621 Chr.9: 127448460-127448460 on Build GRCh38
rs754636743 Chr.9: 127448548-127448548 on Build GRCh38
rs563854425 Chr.9: 127448749-127448751 on Build GRCh38
rs112031868 Chr.9: 127448655-127448656 on Build GRCh38
rs748097311 Chr.9: 127448464-127448464 on Build GRCh38
rs770378789 Chr.9: 127448687-127448687 on Build GRCh38
rs779417892 Chr.9: 127448332-127448332 on Build GRCh38
rs528164261 Chr.9: 127448666-127448666 on Build GRCh38
rs764650592 Chr.9: 127448505-127448505 on Build GRCh38
rs757788903 Chr.9: 127448451-127448452 on Build GRCh38
rs376996721 Chr.9: 127448477-127448477 on Build GRCh38
rs765582967 Chr.9: 127448435-127448435 on Build GRCh38
rs754188666 Chr.9: 127448328-127448328 on Build GRCh38
rs755711277 Chr.9: 127448451-127448451 on Build GRCh38
rs769865461 Chr.9: 127448616-127448616 on Build GRCh38
rs1032669489 Chr.9: 127448757-127448757 on Build GRCh38
rs370343082 Chr.9: 127448736-127448736 on Build GRCh38
rs765609450 Chr.9: 127448493-127448493 on Build GRCh38

View Full Product Details