Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73811721-73812254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSIX
Entrez Gene ID
9383
Gene Name
TSIX transcript, XIST antisense RNA
Gene Aliases
LINC00013, NCRNA00013, XIST-AS, XIST-AS1, XISTAS
UniGene
Hs.529901
Gene Chromosome Location
Chr.X: 73792205-73829231 on Build GRCh38
Transcripts
NR_003255.2

SNP Detail

SNP ID Chromosome Location
rs188913381 Chr.X: 73812108-73812108 on Build GRCh38
rs979130254 Chr.X: 73812047-73812047 on Build GRCh38
rs746345765 Chr.X: 73811859-73811859 on Build GRCh38
rs1011672250 Chr.X: 73812058-73812058 on Build GRCh38
rs998644047 Chr.X: 73812149-73812149 on Build GRCh38
rs930102207 Chr.X: 73811919-73811919 on Build GRCh38
rs756411190 Chr.X: 73812059-73812059 on Build GRCh38
rs1022092665 Chr.X: 73812074-73812074 on Build GRCh38
rs757670819 Chr.X: 73811972-73811972 on Build GRCh38
rs1037834489 Chr.X: 73811884-73811884 on Build GRCh38
rs751126475 Chr.X: 73811865-73811865 on Build GRCh38
rs775685427 Chr.X: 73812035-73812035 on Build GRCh38
rs1025696871 Chr.X: 73811771-73811771 on Build GRCh38
rs760987579 Chr.X: 73812036-73812036 on Build GRCh38
rs920121012 Chr.X: 73811892-73811892 on Build GRCh38
rs779327701 Chr.X: 73811765-73811765 on Build GRCh38
rs987860507 Chr.X: 73812008-73812008 on Build GRCh38
rs182035963 Chr.X: 73812171-73812171 on Build GRCh38
rs781693408 Chr.X: 73812006-73812006 on Build GRCh38
rs967585960 Chr.X: 73811808-73811808 on Build GRCh38
rs959937007 Chr.X: 73812177-73812177 on Build GRCh38
rs915693315 Chr.X: 73811879-73811879 on Build GRCh38
rs772534959 Chr.X: 73811862-73811862 on Build GRCh38
rs915598778 Chr.X: 73812200-73812200 on Build GRCh38
rs959740438 Chr.X: 73811833-73811833 on Build GRCh38
rs977314630 Chr.X: 73811818-73811818 on Build GRCh38
rs975742211 Chr.X: 73811938-73811938 on Build GRCh38
rs895956355 Chr.X: 73812201-73812201 on Build GRCh38
rs771442731 Chr.X: 73811763-73811764 on Build GRCh38
rs991140986 Chr.X: 73812197-73812197 on Build GRCh38
rs58123571 Chr.X: 73811763-73811763 on Build GRCh38
rs891374821 Chr.X: 73811973-73811973 on Build GRCh38
rs928516925 Chr.X: 73811829-73811829 on Build GRCh38
rs1021475373 Chr.X: 73811797-73811797 on Build GRCh38
rs1017263233 Chr.X: 73811842-73811842 on Build GRCh38
rs184729529 Chr.X: 73812219-73812219 on Build GRCh38
rs941771846 Chr.X: 73811880-73811880 on Build GRCh38
rs951373527 Chr.X: 73811805-73811805 on Build GRCh38
rs750913942 Chr.X: 73812016-73812016 on Build GRCh38
rs201649500 Chr.X: 73811759-73811760 on Build GRCh38
rs1047383569 Chr.X: 73811947-73811947 on Build GRCh38
rs73624268 Chr.X: 73811763-73811763 on Build GRCh38
rs1056469470 Chr.X: 73812163-73812164 on Build GRCh38

View Full Product Details