Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144974673-144975223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF252P
Entrez Gene ID
286101
Gene Name
zinc finger protein 252, pseudogene
Gene Aliases
ZNF252
UniGene
Hs.367856
Gene Chromosome Location
Chr.8: 144973589-145002899 on Build GRCh38
Transcripts
NR_023392.1

SNP Detail

SNP ID Chromosome Location
rs753005642 Chr.8: 144975017-144975017 on Build GRCh38
rs574168066 Chr.8: 144975124-144975124 on Build GRCh38
rs1044035991 Chr.8: 144974837-144974837 on Build GRCh38
rs756266808 Chr.8: 144975141-144975142 on Build GRCh38
rs568532132 Chr.8: 144974785-144974785 on Build GRCh38
rs1001585215 Chr.8: 144974813-144974813 on Build GRCh38
rs915456679 Chr.8: 144975079-144975079 on Build GRCh38
rs530294584 Chr.8: 144975108-144975108 on Build GRCh38
rs142161269 Chr.8: 144975018-144975018 on Build GRCh38
rs547315348 Chr.8: 144974902-144974902 on Build GRCh38
rs1054456732 Chr.8: 144975078-144975078 on Build GRCh38
rs769272720 Chr.8: 144974739-144974739 on Build GRCh38
rs753991016 Chr.8: 144975105-144975105 on Build GRCh38
rs548717014 Chr.8: 144975113-144975113 on Build GRCh38
rs987139055 Chr.8: 144974710-144974710 on Build GRCh38
rs955649667 Chr.8: 144974701-144974701 on Build GRCh38
rs972904495 Chr.8: 144974780-144974780 on Build GRCh38
rs151184555 Chr.8: 144975050-144975050 on Build GRCh38
rs1045701173 Chr.8: 144974805-144974805 on Build GRCh38
rs925750655 Chr.8: 144974758-144974758 on Build GRCh38
rs953934252 Chr.8: 144974723-144974723 on Build GRCh38
rs911579158 Chr.8: 144974712-144974716 on Build GRCh38
rs2979290 Chr.8: 144975106-144975106 on Build GRCh38
rs1042554520 Chr.8: 144975104-144975104 on Build GRCh38
rs370757743 Chr.8: 144975161-144975161 on Build GRCh38
rs781288993 Chr.8: 144974964-144974965 on Build GRCh38
rs926982800 Chr.8: 144975058-144975058 on Build GRCh38
rs957088825 Chr.8: 144975132-144975132 on Build GRCh38
rs909687813 Chr.8: 144974747-144974747 on Build GRCh38
rs10104365 Chr.8: 144975052-144975052 on Build GRCh38
rs983983179 Chr.8: 144974733-144974733 on Build GRCh38
rs939846153 Chr.8: 144974771-144974771 on Build GRCh38
rs962823258 Chr.8: 144974831-144974831 on Build GRCh38
rs552336707 Chr.8: 144975184-144975184 on Build GRCh38
rs994439443 Chr.8: 144975168-144975168 on Build GRCh38
rs536783026 Chr.8: 144974757-144974757 on Build GRCh38
rs183171470 Chr.8: 144974962-144974962 on Build GRCh38
rs74628153 Chr.8: 144974841-144974841 on Build GRCh38
rs186053469 Chr.8: 144974903-144974903 on Build GRCh38
rs761531006 Chr.8: 144974698-144974698 on Build GRCh38
rs905875398 Chr.8: 144974811-144974811 on Build GRCh38
rs187495030 Chr.8: 144975098-144975098 on Build GRCh38
rs905482873 Chr.8: 144974857-144974857 on Build GRCh38
rs112938039 Chr.8: 144974963-144974963 on Build GRCh38
rs961508329 Chr.8: 144975046-144975046 on Build GRCh38
rs763843098 Chr.8: 144974704-144974705 on Build GRCh38
rs935828010 Chr.8: 144974766-144974766 on Build GRCh38
rs543141809 Chr.8: 144975153-144975153 on Build GRCh38
rs935642689 Chr.8: 144975059-144975059 on Build GRCh38
rs201189993 Chr.8: 144974749-144974750 on Build GRCh38
rs913025912 Chr.8: 144975157-144975157 on Build GRCh38
rs1017264770 Chr.8: 144975148-144975148 on Build GRCh38
rs62531555 Chr.8: 144974961-144974961 on Build GRCh38
rs182388376 Chr.8: 144974835-144974835 on Build GRCh38
rs935775288 Chr.8: 144974927-144974927 on Build GRCh38
rs1049816521 Chr.8: 144975111-144975111 on Build GRCh38
rs773905832 Chr.8: 144974802-144974802 on Build GRCh38
rs565610593 Chr.8: 144974889-144974899 on Build GRCh38
rs1018589559 Chr.8: 144974889-144974889 on Build GRCh38
rs146160211 Chr.8: 144974904-144974904 on Build GRCh38
rs570818311 Chr.8: 144975193-144975193 on Build GRCh38
rs202224661 Chr.8: 144974823-144974824 on Build GRCh38
rs62531554 Chr.8: 144974958-144974958 on Build GRCh38
rs955597885 Chr.8: 144974879-144974879 on Build GRCh38
rs916969835 Chr.8: 144974794-144974794 on Build GRCh38
rs544152369 Chr.8: 144975103-144975103 on Build GRCh38
rs61391943 Chr.8: 144974880-144974880 on Build GRCh38
rs112846659 Chr.8: 144974951-144974951 on Build GRCh38
rs972551403 Chr.8: 144975049-144975049 on Build GRCh38
rs953966430 Chr.8: 144974973-144974973 on Build GRCh38
rs758168234 Chr.8: 144974794-144974795 on Build GRCh38
rs191043836 Chr.8: 144974829-144974829 on Build GRCh38
rs979762922 Chr.8: 144975055-144975055 on Build GRCh38
rs1039213740 Chr.8: 144974750-144974750 on Build GRCh38
rs1028283880 Chr.8: 144974699-144974699 on Build GRCh38
rs62531553 Chr.8: 144974709-144974709 on Build GRCh38
rs1016823422 Chr.8: 144975043-144975043 on Build GRCh38

View Full Product Details