Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16625746-16626306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROCCP2
Entrez Gene ID
84809
Gene Name
ciliary rootlet coiled-coil, rootletin pseudogene 2
Gene Aliases
CROCCL1
UniGene
Hs.631865
Gene Chromosome Location
Chr.1: 16618256-16630906 on Build GRCh38
Transcripts
NR_026752.1

SNP Detail

SNP ID Chromosome Location
rs2005302 Chr.1: 16626127-16626127 on Build GRCh38
rs540370028 Chr.1: 16626261-16626261 on Build GRCh38
rs576242178 Chr.1: 16626169-16626169 on Build GRCh38
rs11799496 Chr.1: 16626082-16626082 on Build GRCh38
rs57507062 Chr.1: 16625876-16625876 on Build GRCh38
rs200246015 Chr.1: 16625837-16625837 on Build GRCh38
rs560066938 Chr.1: 16625859-16625859 on Build GRCh38
rs879111010 Chr.1: 16626007-16626007 on Build GRCh38
rs533234587 Chr.1: 16625802-16625802 on Build GRCh38
rs1762944 Chr.1: 16626208-16626208 on Build GRCh38
rs371001725 Chr.1: 16625769-16625769 on Build GRCh38
rs1765551 Chr.1: 16626015-16626015 on Build GRCh38
rs571343173 Chr.1: 16626064-16626064 on Build GRCh38
rs553813767 Chr.1: 16626090-16626090 on Build GRCh38
rs373883693 Chr.1: 16625963-16625963 on Build GRCh38
rs1765550 Chr.1: 16626269-16626269 on Build GRCh38
rs777992248 Chr.1: 16625938-16625938 on Build GRCh38
rs578050635 Chr.1: 16626230-16626230 on Build GRCh38
rs744025 Chr.1: 16625887-16625887 on Build GRCh38
rs775382695 Chr.1: 16625877-16625877 on Build GRCh38
rs569427934 Chr.1: 16625975-16625975 on Build GRCh38
rs71259460 Chr.1: 16626068-16626068 on Build GRCh38
rs545203254 Chr.1: 16626184-16626184 on Build GRCh38
rs2002232 Chr.1: 16626155-16626155 on Build GRCh38
rs1765552 Chr.1: 16625772-16625772 on Build GRCh38
rs564902172 Chr.1: 16626224-16626224 on Build GRCh38
rs868254396 Chr.1: 16625961-16625961 on Build GRCh38
rs1056509340 Chr.1: 16626281-16626281 on Build GRCh38
rs995697280 Chr.1: 16625854-16625854 on Build GRCh38
rs915311 Chr.1: 16625986-16625986 on Build GRCh38
rs573717771 Chr.1: 16626128-16626128 on Build GRCh38
rs56766248 Chr.1: 16625856-16625856 on Build GRCh38
rs182896779 Chr.1: 16626161-16626161 on Build GRCh38
rs9658928 Chr.1: 16625882-16625882 on Build GRCh38
rs551652004 Chr.1: 16626003-16626003 on Build GRCh38
rs868751546 Chr.1: 16625858-16625858 on Build GRCh38

View Full Product Details