Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32239330-32239885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR9LP
Entrez Gene ID
339483
Gene Name
myotubularin related protein 9-like, pseudogene
Gene Aliases
MTMR9L
UniGene
Hs.471067
Gene Chromosome Location
Chr.1: 32231660-32241710 on Build GRCh38
Transcripts
NR_026850.1

SNP Detail

SNP ID Chromosome Location
rs7544661 Chr.1: 32239514-32239514 on Build GRCh38
rs1044572706 Chr.1: 32239611-32239612 on Build GRCh38
rs979684506 Chr.1: 32239765-32239765 on Build GRCh38
rs922039402 Chr.1: 32239489-32239489 on Build GRCh38
rs992370098 Chr.1: 32239531-32239531 on Build GRCh38
rs1003871859 Chr.1: 32239719-32239719 on Build GRCh38
rs956936074 Chr.1: 32239832-32239832 on Build GRCh38
rs1013060317 Chr.1: 32239740-32239740 on Build GRCh38
rs959842506 Chr.1: 32239731-32239731 on Build GRCh38
rs995811405 Chr.1: 32239383-32239383 on Build GRCh38
rs879864555 Chr.1: 32239563-32239563 on Build GRCh38
rs925283010 Chr.1: 32239587-32239587 on Build GRCh38
rs905600517 Chr.1: 32239659-32239659 on Build GRCh38
rs925566245 Chr.1: 32239831-32239831 on Build GRCh38
rs561829299 Chr.1: 32239862-32239862 on Build GRCh38
rs1023821366 Chr.1: 32239746-32239746 on Build GRCh38
rs1033710257 Chr.1: 32239363-32239363 on Build GRCh38
rs912225918 Chr.1: 32239693-32239693 on Build GRCh38
rs886775751 Chr.1: 32239714-32239714 on Build GRCh38
rs892067944 Chr.1: 32239722-32239722 on Build GRCh38
rs7521501 Chr.1: 32239592-32239592 on Build GRCh38
rs528587075 Chr.1: 32239418-32239418 on Build GRCh38
rs530842920 Chr.1: 32239840-32239840 on Build GRCh38
rs1035362213 Chr.1: 32239451-32239451 on Build GRCh38
rs751591129 Chr.1: 32239560-32239560 on Build GRCh38
rs781471392 Chr.1: 32239672-32239672 on Build GRCh38
rs35607417 Chr.1: 32239857-32239858 on Build GRCh38
rs34549705 Chr.1: 32239394-32239394 on Build GRCh38
rs1035307151 Chr.1: 32239725-32239725 on Build GRCh38
rs765632825 Chr.1: 32239386-32239386 on Build GRCh38
rs969927221 Chr.1: 32239750-32239750 on Build GRCh38
rs975195036 Chr.1: 32239434-32239434 on Build GRCh38
rs909819000 Chr.1: 32239534-32239534 on Build GRCh38
rs184780117 Chr.1: 32239428-32239428 on Build GRCh38
rs780571018 Chr.1: 32239529-32239529 on Build GRCh38
rs899775867 Chr.1: 32239352-32239352 on Build GRCh38
rs1010461062 Chr.1: 32239752-32239752 on Build GRCh38
rs139252487 Chr.1: 32239613-32239613 on Build GRCh38
rs944106822 Chr.1: 32239637-32239637 on Build GRCh38
rs916718405 Chr.1: 32239539-32239539 on Build GRCh38
rs1010428370 Chr.1: 32239416-32239416 on Build GRCh38
rs151256776 Chr.1: 32239565-32239565 on Build GRCh38
rs995396099 Chr.1: 32239669-32239669 on Build GRCh38
rs952168988 Chr.1: 32239503-32239503 on Build GRCh38
rs866344404 Chr.1: 32239769-32239769 on Build GRCh38
rs917039177 Chr.1: 32239591-32239591 on Build GRCh38
rs748357144 Chr.1: 32239800-32239800 on Build GRCh38
rs556602774 Chr.1: 32239476-32239476 on Build GRCh38
rs899639815 Chr.1: 32239651-32239651 on Build GRCh38
rs1052434319 Chr.1: 32239600-32239600 on Build GRCh38
rs952138502 Chr.1: 32239856-32239856 on Build GRCh38
rs879724332 Chr.1: 32239615-32239615 on Build GRCh38
rs1019127192 Chr.1: 32239785-32239785 on Build GRCh38
rs966242068 Chr.1: 32239788-32239788 on Build GRCh38
rs951020680 Chr.1: 32239396-32239396 on Build GRCh38
rs1036833669 Chr.1: 32239588-32239588 on Build GRCh38
rs1039561945 Chr.1: 32239638-32239638 on Build GRCh38
rs979653710 Chr.1: 32239585-32239585 on Build GRCh38
rs375597260 Chr.1: 32239801-32239802 on Build GRCh38

View Full Product Details