Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178007221-178007784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC730102
Entrez Gene ID
730102
Gene Name
quinone oxidoreductase-like protein 2 pseudogene
Gene Aliases
-
UniGene
Hs.733192
Gene Chromosome Location
Chr.1: 178006136-178038007 on Build GRCh38
Transcripts
NR_037167.1

SNP Detail

SNP ID Chromosome Location
rs957519062 Chr.1: 178007642-178007642 on Build GRCh38
rs11588190 Chr.1: 178007305-178007305 on Build GRCh38
rs560093768 Chr.1: 178007283-178007288 on Build GRCh38
rs976517525 Chr.1: 178007499-178007499 on Build GRCh38
rs374774071 Chr.1: 178007612-178007612 on Build GRCh38
rs770467392 Chr.1: 178007743-178007743 on Build GRCh38
rs183224674 Chr.1: 178007251-178007251 on Build GRCh38
rs529141049 Chr.1: 178007434-178007434 on Build GRCh38
rs994366385 Chr.1: 178007359-178007359 on Build GRCh38
rs564400614 Chr.1: 178007265-178007265 on Build GRCh38
rs1018864623 Chr.1: 178007465-178007465 on Build GRCh38
rs925931600 Chr.1: 178007608-178007608 on Build GRCh38
rs193143028 Chr.1: 178007604-178007604 on Build GRCh38
rs187934326 Chr.1: 178007537-178007537 on Build GRCh38
rs139398902 Chr.1: 178007506-178007506 on Build GRCh38
rs577962988 Chr.1: 178007671-178007671 on Build GRCh38
rs534075850 Chr.1: 178007714-178007714 on Build GRCh38
rs41267166 Chr.1: 178007362-178007362 on Build GRCh38
rs150913893 Chr.1: 178007487-178007487 on Build GRCh38
rs548874053 Chr.1: 178007486-178007486 on Build GRCh38
rs1054278055 Chr.1: 178007295-178007295 on Build GRCh38
rs967066962 Chr.1: 178007597-178007597 on Build GRCh38
rs77296534 Chr.1: 178007741-178007741 on Build GRCh38
rs948144573 Chr.1: 178007328-178007328 on Build GRCh38
rs149678379 Chr.1: 178007602-178007602 on Build GRCh38
rs777550783 Chr.1: 178007366-178007366 on Build GRCh38
rs75692447 Chr.1: 178007592-178007592 on Build GRCh38
rs41267168 Chr.1: 178007399-178007399 on Build GRCh38
rs756841243 Chr.1: 178007484-178007484 on Build GRCh38
rs550717992 Chr.1: 178007257-178007257 on Build GRCh38
rs1046493988 Chr.1: 178007757-178007757 on Build GRCh38
rs746411787 Chr.1: 178007662-178007662 on Build GRCh38
rs566714880 Chr.1: 178007394-178007394 on Build GRCh38
rs1046357585 Chr.1: 178007330-178007330 on Build GRCh38
rs935884364 Chr.1: 178007271-178007271 on Build GRCh38
rs752575677 Chr.1: 178007342-178007342 on Build GRCh38
rs1029316048 Chr.1: 178007583-178007583 on Build GRCh38

View Full Product Details