Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6720361-6720912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GVINP1
Entrez Gene ID
387751
Gene Name
GTPase, very large interferon inducible pseudogene 1
Gene Aliases
GVIN1, GVIN1P, VLIG-1, VLIG1
UniGene
Hs.494757
Gene Chromosome Location
Chr.11: 6713139-6721879 on Build GRCh38
Transcripts
NR_003945.1

SNP Detail

SNP ID Chromosome Location
rs749764936 Chr.11: 6720696-6720696 on Build GRCh38
rs750195142 Chr.11: 6720765-6720765 on Build GRCh38
rs541953484 Chr.11: 6720826-6720826 on Build GRCh38
rs1037318810 Chr.11: 6720566-6720566 on Build GRCh38
rs1020576887 Chr.11: 6720644-6720644 on Build GRCh38
rs1057283440 Chr.11: 6720490-6720490 on Build GRCh38
rs930123916 Chr.11: 6720551-6720551 on Build GRCh38
rs1023074230 Chr.11: 6720547-6720547 on Build GRCh38
rs765704324 Chr.11: 6720625-6720625 on Build GRCh38
rs537266965 Chr.11: 6720410-6720410 on Build GRCh38
rs757456818 Chr.11: 6720431-6720431 on Build GRCh38
rs896066360 Chr.11: 6720491-6720491 on Build GRCh38
rs934918778 Chr.11: 6720486-6720486 on Build GRCh38
rs976756071 Chr.11: 6720774-6720774 on Build GRCh38
rs745948972 Chr.11: 6720569-6720569 on Build GRCh38
rs768945809 Chr.11: 6720699-6720699 on Build GRCh38
rs941547787 Chr.11: 6720887-6720887 on Build GRCh38
rs553136199 Chr.11: 6720593-6720593 on Build GRCh38
rs760462658 Chr.11: 6720876-6720876 on Build GRCh38
rs943885913 Chr.11: 6720387-6720387 on Build GRCh38
rs367916775 Chr.11: 6720457-6720457 on Build GRCh38
rs1041294365 Chr.11: 6720506-6720506 on Build GRCh38
rs778155690 Chr.11: 6720763-6720763 on Build GRCh38
rs577070321 Chr.11: 6720418-6720418 on Build GRCh38
rs901422743 Chr.11: 6720546-6720546 on Build GRCh38
rs769851774 Chr.11: 6720574-6720574 on Build GRCh38
rs867411593 Chr.11: 6720493-6720493 on Build GRCh38
rs965270604 Chr.11: 6720797-6720797 on Build GRCh38
rs752255215 Chr.11: 6720679-6720679 on Build GRCh38
rs1005952352 Chr.11: 6720564-6720564 on Build GRCh38
rs781312445 Chr.11: 6720492-6720492 on Build GRCh38
rs188464350 Chr.11: 6720889-6720889 on Build GRCh38
rs1010064008 Chr.11: 6720636-6720636 on Build GRCh38
rs987460240 Chr.11: 6720705-6720705 on Build GRCh38
rs930258939 Chr.11: 6720812-6720812 on Build GRCh38
rs573275139 Chr.11: 6720721-6720721 on Build GRCh38
rs922870334 Chr.11: 6720791-6720791 on Build GRCh38
rs1035574867 Chr.11: 6720612-6720612 on Build GRCh38
rs375532094 Chr.11: 6720739-6720739 on Build GRCh38
rs546130518 Chr.11: 6720516-6720516 on Build GRCh38
rs1019582490 Chr.11: 6720776-6720776 on Build GRCh38
rs902826519 Chr.11: 6720458-6720458 on Build GRCh38
rs561924174 Chr.11: 6720854-6720854 on Build GRCh38
rs568179300 Chr.11: 6720384-6720384 on Build GRCh38
rs774837243 Chr.11: 6720764-6720764 on Build GRCh38
rs1047709764 Chr.11: 6720558-6720558 on Build GRCh38
rs150366871 Chr.11: 6720413-6720413 on Build GRCh38
rs886185532 Chr.11: 6720560-6720560 on Build GRCh38
rs968818701 Chr.11: 6720675-6720675 on Build GRCh38

View Full Product Details