Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 23827302-23827885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPSAP58
Entrez Gene ID
388524
Gene Name
ribosomal protein SA pseudogene 58
Gene Aliases
RPSA_30_1642
UniGene
Hs.560655
Gene Chromosome Location
Chr.19: 23763014-23828135 on Build GRCh38
Transcripts
NR_003662.3

SNP Detail

SNP ID Chromosome Location
rs907220726 Chr.19: 23827732-23827732 on Build GRCh38
rs747956426 Chr.19: 23827553-23827554 on Build GRCh38
rs200043817 Chr.19: 23827527-23827527 on Build GRCh38
rs775469510 Chr.19: 23827482-23827482 on Build GRCh38
rs551019266 Chr.19: 23827672-23827672 on Build GRCh38
rs756905866 Chr.19: 23827543-23827543 on Build GRCh38
rs552928473 Chr.19: 23827447-23827447 on Build GRCh38
rs561364007 Chr.19: 23827589-23827589 on Build GRCh38
rs943613113 Chr.19: 23827398-23827398 on Build GRCh38
rs772320811 Chr.19: 23827616-23827618 on Build GRCh38
rs200798327 Chr.19: 23827387-23827387 on Build GRCh38
rs9797535 Chr.19: 23827510-23827510 on Build GRCh38
rs745696386 Chr.19: 23827475-23827475 on Build GRCh38
rs773319019 Chr.19: 23827683-23827685 on Build GRCh38
rs776785904 Chr.19: 23827598-23827598 on Build GRCh38
rs577244205 Chr.19: 23827463-23827463 on Build GRCh38
rs200743636 Chr.19: 23827519-23827519 on Build GRCh38
rs765643553 Chr.19: 23827618-23827618 on Build GRCh38
rs761935300 Chr.19: 23827356-23827356 on Build GRCh38
rs976321737 Chr.19: 23827800-23827800 on Build GRCh38
rs749174878 Chr.19: 23827561-23827561 on Build GRCh38
rs749979025 Chr.19: 23827744-23827744 on Build GRCh38
rs546322631 Chr.19: 23827825-23827825 on Build GRCh38
rs749120075 Chr.19: 23827656-23827656 on Build GRCh38
rs150262364 Chr.19: 23827366-23827366 on Build GRCh38
rs770697721 Chr.19: 23827339-23827339 on Build GRCh38
rs201346716 Chr.19: 23827565-23827565 on Build GRCh38
rs778644570 Chr.19: 23827518-23827518 on Build GRCh38
rs530726617 Chr.19: 23827715-23827715 on Build GRCh38
rs9797600 Chr.19: 23827736-23827736 on Build GRCh38
rs1042542504 Chr.19: 23827625-23827625 on Build GRCh38
rs756780509 Chr.19: 23827644-23827644 on Build GRCh38
rs770175253 Chr.19: 23827703-23827703 on Build GRCh38
rs755357217 Chr.19: 23827526-23827526 on Build GRCh38
rs769403802 Chr.19: 23827551-23827551 on Build GRCh38
rs751065074 Chr.19: 23827631-23827631 on Build GRCh38
rs201963256 Chr.19: 23827595-23827595 on Build GRCh38
rs9797605 Chr.19: 23827570-23827570 on Build GRCh38
rs745451204 Chr.19: 23827647-23827647 on Build GRCh38
rs534690539 Chr.19: 23827799-23827799 on Build GRCh38
rs760718579 Chr.19: 23827591-23827591 on Build GRCh38
rs952188843 Chr.19: 23827738-23827738 on Build GRCh38
rs563530333 Chr.19: 23827700-23827700 on Build GRCh38
rs200425946 Chr.19: 23827718-23827718 on Build GRCh38
rs754836246 Chr.19: 23827790-23827790 on Build GRCh38
rs903700741 Chr.19: 23827486-23827486 on Build GRCh38
rs760542340 Chr.19: 23827337-23827337 on Build GRCh38
rs780415340 Chr.19: 23827384-23827384 on Build GRCh38
rs565264699 Chr.19: 23827646-23827646 on Build GRCh38
rs771828936 Chr.19: 23827332-23827332 on Build GRCh38
rs936398735 Chr.19: 23827640-23827640 on Build GRCh38
rs867391250 Chr.19: 23827365-23827365 on Build GRCh38
rs762391345 Chr.19: 23827742-23827742 on Build GRCh38
rs769755380 Chr.19: 23827481-23827481 on Build GRCh38
rs765120047 Chr.19: 23827353-23827353 on Build GRCh38
rs9797606 Chr.19: 23827599-23827599 on Build GRCh38
rs777431188 Chr.19: 23827456-23827456 on Build GRCh38
rs575554058 Chr.19: 23827539-23827539 on Build GRCh38
rs746382473 Chr.19: 23827336-23827338 on Build GRCh38
rs767527128 Chr.19: 23827502-23827502 on Build GRCh38
rs923494545 Chr.19: 23827807-23827807 on Build GRCh38
rs775277450 Chr.19: 23827553-23827553 on Build GRCh38
rs758936403 Chr.19: 23827624-23827624 on Build GRCh38
rs1046307658 Chr.19: 23827714-23827714 on Build GRCh38
rs751941656 Chr.19: 23827741-23827741 on Build GRCh38
rs202081352 Chr.19: 23827465-23827465 on Build GRCh38
rs767717464 Chr.19: 23827358-23827358 on Build GRCh38
rs373029044 Chr.19: 23827422-23827422 on Build GRCh38
rs776307175 Chr.19: 23827342-23827342 on Build GRCh38
rs779340214 Chr.19: 23827418-23827418 on Build GRCh38
rs780763933 Chr.19: 23827550-23827550 on Build GRCh38
rs770121800 Chr.19: 23827844-23827844 on Build GRCh38
rs199953756 Chr.19: 23827474-23827474 on Build GRCh38
rs540400554 Chr.19: 23827620-23827620 on Build GRCh38
rs763463086 Chr.19: 23827730-23827730 on Build GRCh38
rs779728419 Chr.19: 23827773-23827773 on Build GRCh38
rs754210826 Chr.19: 23827522-23827522 on Build GRCh38
rs753619590 Chr.19: 23827779-23827779 on Build GRCh38
rs555278603 Chr.19: 23827348-23827348 on Build GRCh38
rs770850630 Chr.19: 23827693-23827693 on Build GRCh38
rs532517427 Chr.19: 23827670-23827670 on Build GRCh38
rs751613228 Chr.19: 23827354-23827354 on Build GRCh38
rs1013516243 Chr.19: 23827710-23827710 on Build GRCh38
rs567392952 Chr.19: 23827797-23827797 on Build GRCh38
rs930789501 Chr.19: 23827363-23827363 on Build GRCh38
rs776786774 Chr.19: 23827699-23827699 on Build GRCh38
rs952325977 Chr.19: 23827573-23827573 on Build GRCh38
rs771775563 Chr.19: 23827451-23827451 on Build GRCh38
rs571023267 Chr.19: 23827847-23827847 on Build GRCh38
rs753188173 Chr.19: 23827528-23827528 on Build GRCh38
rs755482636 Chr.19: 23827393-23827393 on Build GRCh38
rs964972425 Chr.19: 23827602-23827602 on Build GRCh38
rs773058113 Chr.19: 23827333-23827333 on Build GRCh38
rs9797512 Chr.19: 23827466-23827466 on Build GRCh38
rs768556523 Chr.19: 23827661-23827661 on Build GRCh38
rs772073440 Chr.19: 23827801-23827801 on Build GRCh38
rs766888745 Chr.19: 23827743-23827743 on Build GRCh38
rs13343419 Chr.19: 23827520-23827520 on Build GRCh38
rs773301065 Chr.19: 23827518-23827518 on Build GRCh38
rs746002880 Chr.19: 23827829-23827829 on Build GRCh38
rs868764803 Chr.19: 23827785-23827785 on Build GRCh38
rs1026374703 Chr.19: 23827566-23827566 on Build GRCh38
rs11546165 Chr.19: 23827759-23827759 on Build GRCh38
rs747702872 Chr.19: 23827329-23827329 on Build GRCh38
rs201171553 Chr.19: 23827863-23827863 on Build GRCh38
rs758991399 Chr.19: 23827531-23827531 on Build GRCh38
rs764740632 Chr.19: 23827627-23827627 on Build GRCh38
rs747109603 Chr.19: 23827681-23827681 on Build GRCh38
rs549149533 Chr.19: 23827733-23827733 on Build GRCh38
rs756341866 Chr.19: 23827371-23827371 on Build GRCh38

View Full Product Details