Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200745798-200746387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOX3P-AOX2P
Entrez Gene ID
107161151
Gene Name
AOX3P-AOX2P readthrough
Gene Aliases
AOX3/AOX3L1
UniGene
Hs.148269
Gene Chromosome Location
Chr.2: 200695723-200794915 on Build GRCh38
Transcripts
NR_135011.1 NR_135012.1

SNP Detail

SNP ID Chromosome Location
rs11886328 Chr.2: 200746072-200746072 on Build GRCh38
rs2348121 Chr.2: 200745861-200745861 on Build GRCh38
rs11897081 Chr.2: 200746000-200746000 on Build GRCh38
rs766321918 Chr.2: 200745992-200745992 on Build GRCh38
rs149416454 Chr.2: 200745923-200745923 on Build GRCh38
rs570968975 Chr.2: 200746331-200746331 on Build GRCh38
rs1021556663 Chr.2: 200746168-200746168 on Build GRCh38
rs373374517 Chr.2: 200746304-200746304 on Build GRCh38
rs933654428 Chr.2: 200746070-200746070 on Build GRCh38
rs904851466 Chr.2: 200746097-200746097 on Build GRCh38
rs566277973 Chr.2: 200746107-200746107 on Build GRCh38
rs139642255 Chr.2: 200746013-200746013 on Build GRCh38
rs1001908579 Chr.2: 200746098-200746098 on Build GRCh38
rs146066719 Chr.2: 200746105-200746105 on Build GRCh38
rs1017848041 Chr.2: 200746305-200746305 on Build GRCh38
rs892582005 Chr.2: 200746254-200746254 on Build GRCh38
rs527811518 Chr.2: 200746257-200746257 on Build GRCh38
rs1035099285 Chr.2: 200745996-200745996 on Build GRCh38
rs187813399 Chr.2: 200746276-200746276 on Build GRCh38
rs140006983 Chr.2: 200746181-200746181 on Build GRCh38
rs1003547793 Chr.2: 200745969-200745969 on Build GRCh38
rs531841343 Chr.2: 200746364-200746364 on Build GRCh38
rs948603586 Chr.2: 200746141-200746141 on Build GRCh38
rs979431545 Chr.2: 200745893-200745893 on Build GRCh38
rs1044325274 Chr.2: 200746154-200746154 on Build GRCh38
rs988294522 Chr.2: 200746014-200746014 on Build GRCh38
rs529767912 Chr.2: 200745932-200745932 on Build GRCh38
rs527718664 Chr.2: 200746225-200746225 on Build GRCh38
rs986489539 Chr.2: 200746360-200746360 on Build GRCh38
rs775523335 Chr.2: 200746303-200746303 on Build GRCh38
rs1038291928 Chr.2: 200745818-200745818 on Build GRCh38
rs960862821 Chr.2: 200745824-200745824 on Build GRCh38
rs1022535077 Chr.2: 200746037-200746037 on Build GRCh38
rs1012848894 Chr.2: 200745864-200745864 on Build GRCh38
rs1024032316 Chr.2: 200745912-200745912 on Build GRCh38
rs982613226 Chr.2: 200746358-200746358 on Build GRCh38
rs774143756 Chr.2: 200746040-200746040 on Build GRCh38
rs933789197 Chr.2: 200746008-200746008 on Build GRCh38
rs1056453509 Chr.2: 200746218-200746218 on Build GRCh38
rs552377889 Chr.2: 200746320-200746320 on Build GRCh38
rs539929896 Chr.2: 200745888-200745888 on Build GRCh38
rs1028609926 Chr.2: 200746350-200746350 on Build GRCh38
rs1001513683 Chr.2: 200746226-200746226 on Build GRCh38
rs142700671 Chr.2: 200746332-200746332 on Build GRCh38
rs1009707255 Chr.2: 200746147-200746147 on Build GRCh38
rs13025464 Chr.2: 200745922-200745922 on Build GRCh38
rs976132094 Chr.2: 200745985-200745985 on Build GRCh38
rs941294061 Chr.2: 200746061-200746061 on Build GRCh38
rs910533302 Chr.2: 200745918-200745918 on Build GRCh38
rs939822541 Chr.2: 200746100-200746100 on Build GRCh38
rs968716842 Chr.2: 200745880-200745880 on Build GRCh38
rs754914208 Chr.2: 200745895-200745895 on Build GRCh38
rs908428111 Chr.2: 200746054-200746054 on Build GRCh38
rs950876874 Chr.2: 200746355-200746355 on Build GRCh38
rs964682130 Chr.2: 200746312-200746312 on Build GRCh38
rs988718159 Chr.2: 200745837-200745837 on Build GRCh38
rs905182275 Chr.2: 200746160-200746160 on Build GRCh38
rs923324501 Chr.2: 200746351-200746351 on Build GRCh38
rs896241866 Chr.2: 200746121-200746121 on Build GRCh38

View Full Product Details