Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 21476228-21476770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PI4KAP2
Entrez Gene ID
375133
Gene Name
phosphatidylinositol 4-kinase alpha pseudogene 2
Gene Aliases
-
UniGene
Hs.448225
Gene Chromosome Location
Chr.22: 21472998-21517491 on Build GRCh38
Transcripts
NR_003700.1

SNP Detail

SNP ID Chromosome Location
rs1031881418 Chr.22: 21476724-21476724 on Build GRCh38
rs1012387759 Chr.22: 21476649-21476649 on Build GRCh38
rs778714307 Chr.22: 21476660-21476660 on Build GRCh38
rs1047664555 Chr.22: 21476575-21476575 on Build GRCh38
rs956252957 Chr.22: 21476728-21476728 on Build GRCh38
rs943414119 Chr.22: 21476534-21476534 on Build GRCh38
rs371473302 Chr.22: 21476279-21476279 on Build GRCh38
rs930569151 Chr.22: 21476572-21476572 on Build GRCh38
rs540589775 Chr.22: 21476464-21476464 on Build GRCh38
rs987440798 Chr.22: 21476745-21476745 on Build GRCh38
rs770515347 Chr.22: 21476301-21476301 on Build GRCh38
rs1035066356 Chr.22: 21476614-21476614 on Build GRCh38
rs575039737 Chr.22: 21476445-21476445 on Build GRCh38
rs459015 Chr.22: 21476393-21476393 on Build GRCh38
rs544768272 Chr.22: 21476434-21476434 on Build GRCh38
rs560081941 Chr.22: 21476566-21476566 on Build GRCh38
rs575904922 Chr.22: 21476375-21476375 on Build GRCh38
rs1003206147 Chr.22: 21476603-21476603 on Build GRCh38
rs979018343 Chr.22: 21476672-21476672 on Build GRCh38
rs911983145 Chr.22: 21476510-21476510 on Build GRCh38
rs466486 Chr.22: 21476251-21476251 on Build GRCh38
rs764698117 Chr.22: 21476604-21476604 on Build GRCh38
rs968845531 Chr.22: 21476663-21476663 on Build GRCh38
rs1038878707 Chr.22: 21476560-21476560 on Build GRCh38
rs886138445 Chr.22: 21476590-21476590 on Build GRCh38
rs555208189 Chr.22: 21476438-21476438 on Build GRCh38
rs1023158947 Chr.22: 21476658-21476658 on Build GRCh38
rs898969449 Chr.22: 21476564-21476565 on Build GRCh38

View Full Product Details