Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28093905-28094485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF165
Entrez Gene ID
7718
Gene Name
zinc finger protein 165
Gene Aliases
CT53, LD65, ZSCAN7
UniGene
Hs.55481
Gene Chromosome Location
Chr.6: 28080704-28104244 on Build GRCh38
Transcripts
XM_017011262.1 NR_024063.2

Gene Symbol
ZSCAN12P1
Entrez Gene ID
221584
Gene Name
zinc finger and SCAN domain containing 12 pseudogene 1
Gene Aliases
ZNF187p1, ZNF305P1, ZNF96L1, ZNF96P1, ZSCAN12L1, dJ313I6.7
UniGene
Hs.634723
Gene Chromosome Location
Chr.6: 28090807-28095715 on Build GRCh38
Transcripts
XM_017011262.1 NR_024063.2

SNP Detail

SNP ID Chromosome Location
rs1014711138 Chr.6: 28094018-28094018 on Build GRCh38
rs1018664933 Chr.6: 28094213-28094213 on Build GRCh38
rs774992934 Chr.6: 28094123-28094123 on Build GRCh38
rs962105880 Chr.6: 28094219-28094219 on Build GRCh38
rs949898710 Chr.6: 28094289-28094289 on Build GRCh38
rs776450489 Chr.6: 28094255-28094255 on Build GRCh38
rs942586655 Chr.6: 28094381-28094381 on Build GRCh38
rs893358661 Chr.6: 28094077-28094077 on Build GRCh38
rs149077044 Chr.6: 28094438-28094438 on Build GRCh38
rs561352678 Chr.6: 28094287-28094287 on Build GRCh38
rs203893 Chr.6: 28094288-28094288 on Build GRCh38
rs530292136 Chr.6: 28094049-28094049 on Build GRCh38
rs1014331003 Chr.6: 28094010-28094010 on Build GRCh38
rs991429384 Chr.6: 28094283-28094283 on Build GRCh38
rs893148591 Chr.6: 28094444-28094444 on Build GRCh38
rs1054447757 Chr.6: 28094440-28094440 on Build GRCh38
rs552152513 Chr.6: 28094229-28094229 on Build GRCh38
rs568864051 Chr.6: 28094243-28094243 on Build GRCh38
rs9368551 Chr.6: 28094014-28094014 on Build GRCh38
rs3823180 Chr.6: 28093966-28093966 on Build GRCh38
rs894342908 Chr.6: 28094000-28094000 on Build GRCh38
rs574582167 Chr.6: 28094355-28094355 on Build GRCh38
rs971735643 Chr.6: 28094239-28094239 on Build GRCh38
rs771502060 Chr.6: 28094090-28094090 on Build GRCh38
rs9393889 Chr.6: 28094169-28094169 on Build GRCh38
rs1055609619 Chr.6: 28093982-28093982 on Build GRCh38
rs768505691 Chr.6: 28094203-28094203 on Build GRCh38
rs186533933 Chr.6: 28094098-28094098 on Build GRCh38
rs766126621 Chr.6: 28093965-28093965 on Build GRCh38
rs146488843 Chr.6: 28094369-28094369 on Build GRCh38
rs987171122 Chr.6: 28094185-28094185 on Build GRCh38
rs574680398 Chr.6: 28094033-28094033 on Build GRCh38
rs957553427 Chr.6: 28094230-28094230 on Build GRCh38
rs554630059 Chr.6: 28094344-28094344 on Build GRCh38
rs561368345 Chr.6: 28094046-28094046 on Build GRCh38
rs903243729 Chr.6: 28094328-28094328 on Build GRCh38
rs547010707 Chr.6: 28094074-28094074 on Build GRCh38

View Full Product Details