Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160094919-160095464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2R
Entrez Gene ID
3482
Gene Name
insulin like growth factor 2 receptor
Gene Aliases
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
UniGene
Hs.487062
Gene Chromosome Location
Chr.6: 159969099-160111504 on Build GRCh38
Transcripts
NM_000876.3 NR_003288.2

Gene Symbol
LOC729603
Entrez Gene ID
729603
Gene Name
calcineurin like EF-hand protein 1 pseudogene
Gene Aliases
-
UniGene
Hs.674810
Gene Chromosome Location
Chr.6: 160093082-160096212 on Build GRCh38
Transcripts
NM_000876.3 NR_003288.2

SNP Detail

SNP ID Chromosome Location
rs1051246065 Chr.6: 160095078-160095078 on Build GRCh38
rs542320690 Chr.6: 160095092-160095092 on Build GRCh38
rs562367519 Chr.6: 160095044-160095044 on Build GRCh38
rs367842529 Chr.6: 160095259-160095259 on Build GRCh38
rs553329865 Chr.6: 160095320-160095320 on Build GRCh38
rs988756699 Chr.6: 160095186-160095186 on Build GRCh38
rs1002967830 Chr.6: 160095164-160095164 on Build GRCh38
rs146639193 Chr.6: 160095371-160095371 on Build GRCh38
rs1010229150 Chr.6: 160095433-160095433 on Build GRCh38
rs921048505 Chr.6: 160095062-160095062 on Build GRCh38
rs556001804 Chr.6: 160095358-160095358 on Build GRCh38
rs946578268 Chr.6: 160094993-160094993 on Build GRCh38
rs545623042 Chr.6: 160094971-160094971 on Build GRCh38
rs906045714 Chr.6: 160095344-160095344 on Build GRCh38
rs568054655 Chr.6: 160095284-160095284 on Build GRCh38
rs533578118 Chr.6: 160095285-160095285 on Build GRCh38
rs561995918 Chr.6: 160095103-160095103 on Build GRCh38
rs191704446 Chr.6: 160095212-160095212 on Build GRCh38
rs538031842 Chr.6: 160095306-160095306 on Build GRCh38
rs571187546 Chr.6: 160095138-160095138 on Build GRCh38
rs986577734 Chr.6: 160095264-160095264 on Build GRCh38
rs932415156 Chr.6: 160095070-160095070 on Build GRCh38
rs531373258 Chr.6: 160095084-160095084 on Build GRCh38
rs567368125 Chr.6: 160095079-160095079 on Build GRCh38
rs557553403 Chr.6: 160094997-160094997 on Build GRCh38
rs577696255 Chr.6: 160095091-160095091 on Build GRCh38
rs80010378 Chr.6: 160095416-160095416 on Build GRCh38
rs74485061 Chr.6: 160095111-160095111 on Build GRCh38
rs186903186 Chr.6: 160095119-160095119 on Build GRCh38
rs996106417 Chr.6: 160095139-160095139 on Build GRCh38
rs905988181 Chr.6: 160095160-160095160 on Build GRCh38
rs1023124576 Chr.6: 160095141-160095141 on Build GRCh38
rs866735650 Chr.6: 160095121-160095121 on Build GRCh38
rs139725654 Chr.6: 160095242-160095242 on Build GRCh38
rs753878384 Chr.6: 160095408-160095408 on Build GRCh38
rs146359149 Chr.6: 160095211-160095211 on Build GRCh38
rs144501044 Chr.6: 160095340-160095340 on Build GRCh38
rs115628416 Chr.6: 160095176-160095176 on Build GRCh38
rs576402356 Chr.6: 160095327-160095327 on Build GRCh38
rs560056682 Chr.6: 160094968-160094968 on Build GRCh38
rs541659197 Chr.6: 160095390-160095390 on Build GRCh38
rs550462289 Chr.6: 160095195-160095195 on Build GRCh38
rs183094588 Chr.6: 160094970-160094970 on Build GRCh38
rs956006522 Chr.6: 160094961-160094961 on Build GRCh38
rs1020859080 Chr.6: 160095188-160095189 on Build GRCh38
rs931587048 Chr.6: 160095332-160095332 on Build GRCh38
rs1036155058 Chr.6: 160095174-160095174 on Build GRCh38
rs754867859 Chr.6: 160095298-160095298 on Build GRCh38
rs1035698132 Chr.6: 160094960-160094960 on Build GRCh38
rs1021580414 Chr.6: 160095436-160095436 on Build GRCh38

View Full Product Details