Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160466415-160466959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPAL2
Entrez Gene ID
80350
Gene Name
lipoprotein(a) like 2, pseudogene
Gene Aliases
APOA2, APOAL, APOARGC, apo(a)rg-C
UniGene
Hs.654503
Gene Chromosome Location
Chr.6: 160466555-160511124 on Build GRCh38
Transcripts
NR_028093.1

SNP Detail

SNP ID Chromosome Location
rs530861775 Chr.6: 160466583-160466583 on Build GRCh38
rs188805861 Chr.6: 160466937-160466937 on Build GRCh38
rs548927074 Chr.6: 160466795-160466795 on Build GRCh38
rs569348427 Chr.6: 160466565-160466565 on Build GRCh38
rs958854790 Chr.6: 160466758-160466758 on Build GRCh38
rs993957750 Chr.6: 160466647-160466647 on Build GRCh38
rs972812933 Chr.6: 160466710-160466710 on Build GRCh38
rs920115210 Chr.6: 160466710-160466710 on Build GRCh38
rs904327460 Chr.6: 160466562-160466563 on Build GRCh38
rs541143853 Chr.6: 160466461-160466461 on Build GRCh38
rs906606688 Chr.6: 160466923-160466923 on Build GRCh38
rs981331919 Chr.6: 160466436-160466436 on Build GRCh38
rs954061157 Chr.6: 160466686-160466686 on Build GRCh38
rs952890952 Chr.6: 160466715-160466715 on Build GRCh38
rs371863455 Chr.6: 160466713-160466713 on Build GRCh38
rs910240202 Chr.6: 160466824-160466824 on Build GRCh38
rs980798807 Chr.6: 160466716-160466716 on Build GRCh38
rs184078619 Chr.6: 160466798-160466798 on Build GRCh38
rs73592970 Chr.6: 160466889-160466889 on Build GRCh38
rs889359190 Chr.6: 160466571-160466571 on Build GRCh38
rs932992122 Chr.6: 160466505-160466505 on Build GRCh38
rs750688468 Chr.6: 160466885-160466885 on Build GRCh38
rs771217329 Chr.6: 160466498-160466498 on Build GRCh38
rs764648288 Chr.6: 160466578-160466579 on Build GRCh38
rs369273832 Chr.6: 160466561-160466561 on Build GRCh38
rs985759381 Chr.6: 160466822-160466822 on Build GRCh38
rs112821455 Chr.6: 160466799-160466799 on Build GRCh38
rs1056845886 Chr.6: 160466553-160466553 on Build GRCh38
rs550916734 Chr.6: 160466814-160466814 on Build GRCh38
rs1019638255 Chr.6: 160466870-160466870 on Build GRCh38
rs749262514 Chr.6: 160466495-160466495 on Build GRCh38
rs967256382 Chr.6: 160466775-160466775 on Build GRCh38
rs539957470 Chr.6: 160466641-160466641 on Build GRCh38
rs1002314136 Chr.6: 160466935-160466935 on Build GRCh38
rs529120582 Chr.6: 160466681-160466681 on Build GRCh38
rs906493698 Chr.6: 160466704-160466704 on Build GRCh38
rs570558777 Chr.6: 160466843-160466843 on Build GRCh38
rs953029018 Chr.6: 160466903-160466903 on Build GRCh38
rs923034857 Chr.6: 160466813-160466813 on Build GRCh38
rs942260844 Chr.6: 160466831-160466831 on Build GRCh38
rs879566979 Chr.6: 160466773-160466773 on Build GRCh38
rs912877326 Chr.6: 160466745-160466745 on Build GRCh38
rs1033712888 Chr.6: 160466754-160466754 on Build GRCh38
rs977212499 Chr.6: 160466469-160466469 on Build GRCh38
rs1027065708 Chr.6: 160466896-160466896 on Build GRCh38
rs371422127 Chr.6: 160466458-160466458 on Build GRCh38
rs1034269507 Chr.6: 160466454-160466454 on Build GRCh38
rs938904810 Chr.6: 160466725-160466725 on Build GRCh38
rs560113947 Chr.6: 160466490-160466490 on Build GRCh38
rs377096051 Chr.6: 160466892-160466892 on Build GRCh38
rs1008794470 Chr.6: 160466863-160466863 on Build GRCh38
rs551099741 Chr.6: 160466595-160466595 on Build GRCh38
rs1038255525 Chr.6: 160466592-160466592 on Build GRCh38
rs898019846 Chr.6: 160466593-160466593 on Build GRCh38
rs1044637169 Chr.6: 160466481-160466481 on Build GRCh38
rs920081852 Chr.6: 160466445-160466445 on Build GRCh38
rs776043854 Chr.6: 160466888-160466888 on Build GRCh38
rs574219253 Chr.6: 160466460-160466460 on Build GRCh38
rs1019586089 Chr.6: 160466703-160466703 on Build GRCh38
rs890380001 Chr.6: 160466844-160466844 on Build GRCh38
rs542938897 Chr.6: 160466484-160466484 on Build GRCh38
rs980248843 Chr.6: 160466924-160466924 on Build GRCh38
rs773358151 Chr.6: 160466466-160466466 on Build GRCh38
rs769825723 Chr.6: 160466462-160466462 on Build GRCh38
rs938349895 Chr.6: 160466510-160466510 on Build GRCh38
rs531750142 Chr.6: 160466714-160466715 on Build GRCh38
rs565287471 Chr.6: 160466562-160466562 on Build GRCh38
rs780185208 Chr.6: 160466459-160466459 on Build GRCh38
rs572114425 Chr.6: 160466455-160466455 on Build GRCh38
rs377545028 Chr.6: 160466448-160466449 on Build GRCh38
rs927544535 Chr.6: 160466720-160466720 on Build GRCh38
rs1008239740 Chr.6: 160466701-160466701 on Build GRCh38
rs990113666 Chr.6: 160466769-160466769 on Build GRCh38
rs891556683 Chr.6: 160466554-160466554 on Build GRCh38
rs563328443 Chr.6: 160466666-160466666 on Build GRCh38
rs1002212742 Chr.6: 160466707-160466707 on Build GRCh38
rs1056729609 Chr.6: 160466737-160466737 on Build GRCh38
rs573598374 Chr.6: 160466503-160466503 on Build GRCh38
rs767836464 Chr.6: 160466868-160466868 on Build GRCh38
rs560775987 Chr.6: 160466557-160466557 on Build GRCh38

View Full Product Details