Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 38685827-38686376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM183BP
Entrez Gene ID
340286
Gene Name
acyloxyacyl hydrolase (neutrophil)
Gene Aliases
FAM183B, THEG6
UniGene
Hs.144075
Gene Chromosome Location
Chr.7: 38685346-38687089 on Build GRCh38
Transcripts
NR_028347.1

SNP Detail

SNP ID Chromosome Location
rs368040568 Chr.7: 38685991-38685991 on Build GRCh38
rs772601721 Chr.7: 38686043-38686043 on Build GRCh38
rs780598503 Chr.7: 38686034-38686034 on Build GRCh38
rs774859366 Chr.7: 38685973-38685973 on Build GRCh38
rs775500401 Chr.7: 38686165-38686165 on Build GRCh38
rs748711096 Chr.7: 38686100-38686100 on Build GRCh38
rs764414668 Chr.7: 38685885-38685885 on Build GRCh38
rs1020679828 Chr.7: 38686175-38686175 on Build GRCh38
rs780173876 Chr.7: 38685919-38685919 on Build GRCh38
rs922409414 Chr.7: 38686021-38686021 on Build GRCh38
rs766561213 Chr.7: 38686059-38686059 on Build GRCh38
rs779730651 Chr.7: 38686239-38686239 on Build GRCh38
rs772717151 Chr.7: 38685855-38685855 on Build GRCh38
rs200110756 Chr.7: 38685978-38685978 on Build GRCh38
rs11980087 Chr.7: 38686204-38686204 on Build GRCh38
rs770567254 Chr.7: 38686033-38686033 on Build GRCh38
rs949717497 Chr.7: 38686344-38686344 on Build GRCh38
rs141721757 Chr.7: 38686193-38686193 on Build GRCh38
rs368311158 Chr.7: 38686054-38686054 on Build GRCh38
rs748743822 Chr.7: 38686250-38686250 on Build GRCh38
rs776182585 Chr.7: 38685958-38685958 on Build GRCh38
rs752619200 Chr.7: 38686014-38686014 on Build GRCh38
rs780838346 Chr.7: 38686089-38686089 on Build GRCh38
rs111578732 Chr.7: 38686124-38686124 on Build GRCh38
rs1001655141 Chr.7: 38686118-38686118 on Build GRCh38
rs769409104 Chr.7: 38686094-38686094 on Build GRCh38
rs747467802 Chr.7: 38686292-38686292 on Build GRCh38
rs914887133 Chr.7: 38686022-38686022 on Build GRCh38
rs745308299 Chr.7: 38686035-38686035 on Build GRCh38
rs368968799 Chr.7: 38685915-38685915 on Build GRCh38
rs752674242 Chr.7: 38686071-38686071 on Build GRCh38
rs765352218 Chr.7: 38686150-38686150 on Build GRCh38
rs757582617 Chr.7: 38685891-38685891 on Build GRCh38
rs976629921 Chr.7: 38686009-38686009 on Build GRCh38
rs906019973 Chr.7: 38686108-38686108 on Build GRCh38
rs532827309 Chr.7: 38686030-38686030 on Build GRCh38
rs777566159 Chr.7: 38686075-38686075 on Build GRCh38
rs758258582 Chr.7: 38686023-38686023 on Build GRCh38
rs1029713785 Chr.7: 38686255-38686255 on Build GRCh38
rs772669575 Chr.7: 38686103-38686103 on Build GRCh38
rs753787240 Chr.7: 38686266-38686266 on Build GRCh38
rs746660101 Chr.7: 38685949-38685949 on Build GRCh38
rs56870388 Chr.7: 38686063-38686063 on Build GRCh38
rs994154306 Chr.7: 38686145-38686145 on Build GRCh38
rs370143311 Chr.7: 38685964-38685964 on Build GRCh38
rs539305097 Chr.7: 38686228-38686228 on Build GRCh38
rs776979598 Chr.7: 38686129-38686129 on Build GRCh38
rs370929680 Chr.7: 38686037-38686037 on Build GRCh38
rs765298994 Chr.7: 38686070-38686070 on Build GRCh38
rs778606254 Chr.7: 38685937-38685937 on Build GRCh38
rs771727053 Chr.7: 38685944-38685944 on Build GRCh38
rs759792874 Chr.7: 38685928-38685928 on Build GRCh38
rs772928965 Chr.7: 38686001-38686001 on Build GRCh38
rs375635929 Chr.7: 38685918-38685918 on Build GRCh38
rs751325256 Chr.7: 38686201-38686201 on Build GRCh38
rs745489994 Chr.7: 38686093-38686093 on Build GRCh38
rs759720696 Chr.7: 38686069-38686069 on Build GRCh38
rs552819797 Chr.7: 38686031-38686031 on Build GRCh38
rs575827341 Chr.7: 38686314-38686314 on Build GRCh38
rs754899319 Chr.7: 38685925-38685925 on Build GRCh38
rs760842236 Chr.7: 38686055-38686055 on Build GRCh38
rs747824586 Chr.7: 38685851-38685851 on Build GRCh38
rs374472663 Chr.7: 38685988-38685988 on Build GRCh38
rs1001602790 Chr.7: 38685866-38685866 on Build GRCh38
rs547012619 Chr.7: 38685878-38685878 on Build GRCh38
rs373489323 Chr.7: 38685852-38685852 on Build GRCh38
rs111509905 Chr.7: 38686099-38686099 on Build GRCh38
rs767207904 Chr.7: 38686209-38686209 on Build GRCh38
rs751314033 Chr.7: 38686081-38686081 on Build GRCh38
rs927669574 Chr.7: 38686305-38686305 on Build GRCh38
rs548267023 Chr.7: 38686123-38686123 on Build GRCh38
rs771368165 Chr.7: 38686304-38686304 on Build GRCh38
rs766394920 Chr.7: 38685992-38685992 on Build GRCh38
rs775928436 Chr.7: 38685879-38685879 on Build GRCh38
rs758311679 Chr.7: 38686073-38686073 on Build GRCh38
rs60277655 Chr.7: 38686082-38686082 on Build GRCh38
rs962878241 Chr.7: 38686285-38686285 on Build GRCh38
rs767780877 Chr.7: 38685912-38685912 on Build GRCh38
rs369965222 Chr.7: 38686173-38686174 on Build GRCh38
rs749219684 Chr.7: 38685921-38685921 on Build GRCh38
rs747873801 Chr.7: 38685940-38685940 on Build GRCh38
rs760122112 Chr.7: 38685873-38685873 on Build GRCh38
rs1029681882 Chr.7: 38686151-38686151 on Build GRCh38
rs750126180 Chr.7: 38686214-38686214 on Build GRCh38
rs772909023 Chr.7: 38685945-38685945 on Build GRCh38
rs778855936 Chr.7: 38686005-38686005 on Build GRCh38
rs568006595 Chr.7: 38686127-38686127 on Build GRCh38
rs765862801 Chr.7: 38685877-38685877 on Build GRCh38
rs767424478 Chr.7: 38686094-38686095 on Build GRCh38
rs757065959 Chr.7: 38686054-38686055 on Build GRCh38
rs778486450 Chr.7: 38686287-38686287 on Build GRCh38
rs762908164 Chr.7: 38686166-38686166 on Build GRCh38
rs533768792 Chr.7: 38686128-38686128 on Build GRCh38
rs12701651 Chr.7: 38686283-38686283 on Build GRCh38
rs188752575 Chr.7: 38686176-38686176 on Build GRCh38
rs985548628 Chr.7: 38686268-38686268 on Build GRCh38
rs751255802 Chr.7: 38686028-38686029 on Build GRCh38
rs763986447 Chr.7: 38685986-38685989 on Build GRCh38
rs34871648 Chr.7: 38686150-38686151 on Build GRCh38
rs773686260 Chr.7: 38686045-38686045 on Build GRCh38
rs759645693 Chr.7: 38686137-38686137 on Build GRCh38
rs1010252914 Chr.7: 38686138-38686138 on Build GRCh38
rs560816398 Chr.7: 38686000-38686000 on Build GRCh38
rs1054921746 Chr.7: 38686350-38686350 on Build GRCh38
rs375485270 Chr.7: 38686042-38686042 on Build GRCh38
rs763486308 Chr.7: 38685881-38685881 on Build GRCh38
rs569780333 Chr.7: 38686060-38686060 on Build GRCh38
rs767977876 Chr.7: 38685980-38685980 on Build GRCh38
rs945005418 Chr.7: 38686295-38686295 on Build GRCh38
rs770388735 Chr.7: 38685956-38685956 on Build GRCh38

View Full Product Details