Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43941155-43941715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2J4
Entrez Gene ID
84820
Gene Name
RNA polymerase II subunit J4, pseudogene
Gene Aliases
RPB11-phi
UniGene
Hs.657028
Gene Chromosome Location
Chr.7: 43940895-44019194 on Build GRCh38
Transcripts
NR_003655.3 NM_015983.3 XM_006715734.3 XM_011515420.2 XM_011515422.2 XM_011515423.2 XM_017012292.1 XM_017012293.1 XM_017012294.1 XM_017012295.1

Gene Symbol
UBE2D4
Entrez Gene ID
51619
Gene Name
ubiquitin conjugating enzyme E2 D4 (putative)
Gene Aliases
HBUCE1
UniGene
Hs.19196
Gene Chromosome Location
Chr.7: 43926436-43956136 on Build GRCh38
Transcripts
NR_003655.3 NM_015983.3 XM_006715734.3 XM_011515420.2 XM_011515422.2 XM_011515423.2 XM_017012292.1 XM_017012293.1 XM_017012294.1 XM_017012295.1

SNP Detail

SNP ID Chromosome Location
rs913937817 Chr.7: 43941559-43941559 on Build GRCh38
rs931767581 Chr.7: 43941564-43941564 on Build GRCh38
rs890205592 Chr.7: 43941255-43941255 on Build GRCh38
rs973699406 Chr.7: 43941495-43941495 on Build GRCh38
rs145283226 Chr.7: 43941412-43941412 on Build GRCh38
rs564648446 Chr.7: 43941293-43941293 on Build GRCh38
rs755698285 Chr.7: 43941528-43941528 on Build GRCh38
rs780852784 Chr.7: 43941452-43941452 on Build GRCh38
rs569828592 Chr.7: 43941561-43941561 on Build GRCh38
rs879096056 Chr.7: 43941675-43941675 on Build GRCh38
rs375092946 Chr.7: 43941634-43941634 on Build GRCh38
rs961905754 Chr.7: 43941411-43941411 on Build GRCh38
rs926655217 Chr.7: 43941459-43941459 on Build GRCh38
rs190063475 Chr.7: 43941509-43941509 on Build GRCh38
rs551539318 Chr.7: 43941649-43941649 on Build GRCh38
rs545345396 Chr.7: 43941281-43941281 on Build GRCh38
rs1052084985 Chr.7: 43941325-43941325 on Build GRCh38
rs1051915304 Chr.7: 43941248-43941248 on Build GRCh38
rs1001720625 Chr.7: 43941384-43941384 on Build GRCh38
rs574363397 Chr.7: 43941510-43941510 on Build GRCh38
rs370268499 Chr.7: 43941653-43941653 on Build GRCh38
rs143087818 Chr.7: 43941448-43941448 on Build GRCh38
rs776950585 Chr.7: 43941670-43941670 on Build GRCh38
rs999877464 Chr.7: 43941338-43941338 on Build GRCh38
rs528834473 Chr.7: 43941207-43941207 on Build GRCh38
rs1040130054 Chr.7: 43941657-43941657 on Build GRCh38
rs796924651 Chr.7: 43941360-43941360 on Build GRCh38
rs925545070 Chr.7: 43941586-43941586 on Build GRCh38
rs118125137 Chr.7: 43941478-43941478 on Build GRCh38
rs993701207 Chr.7: 43941414-43941414 on Build GRCh38
rs904072368 Chr.7: 43941283-43941283 on Build GRCh38
rs897539132 Chr.7: 43941662-43941662 on Build GRCh38
rs28665738 Chr.7: 43941211-43941211 on Build GRCh38
rs1056938194 Chr.7: 43941206-43941206 on Build GRCh38
rs939474924 Chr.7: 43941188-43941188 on Build GRCh38
rs751369169 Chr.7: 43941347-43941347 on Build GRCh38
rs2527811 Chr.7: 43941435-43941435 on Build GRCh38
rs368146340 Chr.7: 43941677-43941677 on Build GRCh38
rs1049993851 Chr.7: 43941609-43941609 on Build GRCh38
rs1043891137 Chr.7: 43941575-43941575 on Build GRCh38
rs999145979 Chr.7: 43941287-43941287 on Build GRCh38
rs779653480 Chr.7: 43941608-43941608 on Build GRCh38
rs192950507 Chr.7: 43941684-43941684 on Build GRCh38
rs945533985 Chr.7: 43941221-43941221 on Build GRCh38
rs1028642148 Chr.7: 43941348-43941348 on Build GRCh38
rs946418315 Chr.7: 43941682-43941682 on Build GRCh38
rs960143900 Chr.7: 43941422-43941422 on Build GRCh38
rs1027466731 Chr.7: 43941689-43941689 on Build GRCh38
rs528750423 Chr.7: 43941367-43941367 on Build GRCh38
rs1052033412 Chr.7: 43941654-43941654 on Build GRCh38
rs1044005592 Chr.7: 43941253-43941253 on Build GRCh38
rs888763843 Chr.7: 43941352-43941352 on Build GRCh38
rs1019105017 Chr.7: 43941408-43941408 on Build GRCh38

View Full Product Details