Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99289189-99289657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH16
Entrez Gene ID
84176
Gene Name
myosin heavy chain 16 pseudogene
Gene Aliases
MHC20, MYH16P, MYH5
UniGene
Hs.621401
Gene Chromosome Location
Chr.7: 99273301-99297972 on Build GRCh38
Transcripts
NR_002147.2

SNP Detail

SNP ID Chromosome Location
rs557612958 Chr.7: 99289363-99289363 on Build GRCh38
rs541036744 Chr.7: 99289459-99289459 on Build GRCh38
rs191278629 Chr.7: 99289367-99289367 on Build GRCh38
rs779568864 Chr.7: 99289300-99289300 on Build GRCh38
rs901546711 Chr.7: 99289574-99289574 on Build GRCh38
rs772256663 Chr.7: 99289384-99289384 on Build GRCh38
rs538511224 Chr.7: 99289247-99289247 on Build GRCh38
rs561315139 Chr.7: 99289424-99289424 on Build GRCh38
rs953505504 Chr.7: 99289258-99289259 on Build GRCh38
rs994687277 Chr.7: 99289251-99289251 on Build GRCh38
rs879863872 Chr.7: 99289381-99289381 on Build GRCh38
rs747018344 Chr.7: 99289418-99289418 on Build GRCh38
rs1016272789 Chr.7: 99289224-99289224 on Build GRCh38
rs748134996 Chr.7: 99289305-99289305 on Build GRCh38
rs545515767 Chr.7: 99289534-99289534 on Build GRCh38
rs765902931 Chr.7: 99289453-99289453 on Build GRCh38
rs936796218 Chr.7: 99289423-99289423 on Build GRCh38
rs968594266 Chr.7: 99289377-99289377 on Build GRCh38
rs762739854 Chr.7: 99289613-99289613 on Build GRCh38
rs976402479 Chr.7: 99289242-99289242 on Build GRCh38
rs114672351 Chr.7: 99289395-99289395 on Build GRCh38
rs966002168 Chr.7: 99289609-99289609 on Build GRCh38
rs573653136 Chr.7: 99289296-99289296 on Build GRCh38
rs1006017827 Chr.7: 99289222-99289222 on Build GRCh38
rs529175241 Chr.7: 99289444-99289444 on Build GRCh38
rs917938687 Chr.7: 99289295-99289295 on Build GRCh38
rs748184153 Chr.7: 99289502-99289502 on Build GRCh38
rs138207582 Chr.7: 99289553-99289553 on Build GRCh38
rs995053484 Chr.7: 99289509-99289509 on Build GRCh38
rs897753108 Chr.7: 99289467-99289467 on Build GRCh38
rs988354520 Chr.7: 99289347-99289347 on Build GRCh38
rs143692087 Chr.7: 99289607-99289607 on Build GRCh38
rs771077398 Chr.7: 99289606-99289606 on Build GRCh38
rs981467452 Chr.7: 99289373-99289373 on Build GRCh38
rs937120935 Chr.7: 99289419-99289419 on Build GRCh38
rs545571229 Chr.7: 99289301-99289301 on Build GRCh38
rs945808864 Chr.7: 99289524-99289524 on Build GRCh38
rs553373900 Chr.7: 99289470-99289470 on Build GRCh38
rs748673669 Chr.7: 99289340-99289340 on Build GRCh38
rs578031160 Chr.7: 99289522-99289524 on Build GRCh38
rs1007544935 Chr.7: 99289259-99289259 on Build GRCh38
rs768802263 Chr.7: 99289441-99289441 on Build GRCh38
rs747226326 Chr.7: 99289582-99289582 on Build GRCh38
rs556873452 Chr.7: 99289252-99289252 on Build GRCh38

View Full Product Details