Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148086301-148086644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPINK5
Entrez Gene ID
11005
Gene Name
serine peptidase inhibitor, Kazal type 5
Gene Aliases
LEKTI, LETKI, NETS, NS, VAKTI
UniGene
Hs.331555
Gene Chromosome Location
Chr.5: 148063972-148137362 on Build GRCh38
Transcripts
NM_001127698.1 NM_001127699.1 NM_006846.3 XM_011537551.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
351167 c.364G>C p.D122H Chr.5: 148086486-148086486 on Build GRCh38
1435006 c.326T>G p.F109C Chr.5: 148086448-148086448 on Build GRCh38
1063965 c.410C>T p.A137V Chr.5: 148086532-148086532 on Build GRCh38
5805494 c.357T>A p.C119* Chr.5: 148086479-148086479 on Build GRCh38
7303364 c.381C>T p.D127D Chr.5: 148086503-148086503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992538921 Chr.5: 148086335-148086335 on Build GRCh38
rs867936703 Chr.5: 148086490-148086491 on Build GRCh38
rs753523025 Chr.5: 148086426-148086426 on Build GRCh38
rs879132267 Chr.5: 148086611-148086611 on Build GRCh38
rs777435572 Chr.5: 148086412-148086412 on Build GRCh38
rs775311063 Chr.5: 148086548-148086548 on Build GRCh38
rs767428266 Chr.5: 148086542-148086542 on Build GRCh38
rs559487611 Chr.5: 148086340-148086340 on Build GRCh38
rs371334869 Chr.5: 148086435-148086435 on Build GRCh38
rs563549882 Chr.5: 148086376-148086376 on Build GRCh38
rs1024621128 Chr.5: 148086356-148086356 on Build GRCh38
rs17860502 Chr.5: 148086438-148086438 on Build GRCh38
rs542109572 Chr.5: 148086578-148086578 on Build GRCh38
rs746365387 Chr.5: 148086439-148086439 on Build GRCh38
rs757967707 Chr.5: 148086431-148086434 on Build GRCh38
rs866691321 Chr.5: 148086452-148086452 on Build GRCh38
rs185771462 Chr.5: 148086357-148086357 on Build GRCh38
rs753467836 Chr.5: 148086565-148086565 on Build GRCh38
rs140485487 Chr.5: 148086381-148086381 on Build GRCh38
rs371877583 Chr.5: 148086524-148086524 on Build GRCh38
rs587777749 Chr.5: 148086403-148086403 on Build GRCh38
rs756962716 Chr.5: 148086568-148086568 on Build GRCh38
rs201524438 Chr.5: 148086373-148086373 on Build GRCh38
rs371569547 Chr.5: 148086498-148086498 on Build GRCh38
rs1040416742 Chr.5: 148086593-148086593 on Build GRCh38
rs747506940 Chr.5: 148086576-148086576 on Build GRCh38
rs763649250 Chr.5: 148086435-148086436 on Build GRCh38
rs969024148 Chr.5: 148086372-148086372 on Build GRCh38
rs751285979 Chr.5: 148086497-148086498 on Build GRCh38
rs201204643 Chr.5: 148086380-148086380 on Build GRCh38
rs772706833 Chr.5: 148086450-148086450 on Build GRCh38
rs368159711 Chr.5: 148086516-148086516 on Build GRCh38
rs774354593 Chr.5: 148086532-148086532 on Build GRCh38
rs764853169 Chr.5: 148086463-148086463 on Build GRCh38
rs780564429 Chr.5: 148086571-148086571 on Build GRCh38
rs747056451 Chr.5: 148086466-148086466 on Build GRCh38
rs965514325 Chr.5: 148086512-148086512 on Build GRCh38
rs763636029 Chr.5: 148086557-148086557 on Build GRCh38
rs376123101 Chr.5: 148086499-148086499 on Build GRCh38
rs73269187 Chr.5: 148086569-148086569 on Build GRCh38
rs375224787 Chr.5: 148086533-148086533 on Build GRCh38
rs926170879 Chr.5: 148086552-148086552 on Build GRCh38
rs756874525 Chr.5: 148086510-148086511 on Build GRCh38
rs752183279 Chr.5: 148086393-148086393 on Build GRCh38
rs1013741887 Chr.5: 148086341-148086341 on Build GRCh38
rs1053321621 Chr.5: 148086599-148086599 on Build GRCh38
rs768859182 Chr.5: 148086369-148086369 on Build GRCh38
rs766931877 Chr.5: 148086384-148086384 on Build GRCh38
rs770833255 Chr.5: 148086527-148086527 on Build GRCh38
rs895955973 Chr.5: 148086621-148086621 on Build GRCh38
rs755590932 Chr.5: 148086410-148086410 on Build GRCh38
rs546888008 Chr.5: 148086603-148086603 on Build GRCh38
rs760194192 Chr.5: 148086549-148086549 on Build GRCh38
rs781070285 Chr.5: 148086552-148086552 on Build GRCh38

View Full Product Details