Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193138000-193138167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC73
Entrez Gene ID
79577
Gene Name
cell division cycle 73
Gene Aliases
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX
UniGene
Hs.378996
Gene Chromosome Location
Chr.1: 193121958-193254815 on Build GRCh38
Transcripts
NM_024529.4 XM_006711537.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
901381 c.437T>C p.V146A Chr.1: 193138098-193138098 on Build GRCh38
4517923 c.465_466CC>TT p.R156C Chr.1: 193138126-193138127 on Build GRCh38
6917279 c.496C>G p.Q166E Chr.1: 193138157-193138157 on Build GRCh38
94479 c.439C>T p.R147C Chr.1: 193138100-193138100 on Build GRCh38
6949870 c.431delA p.E144fs*58 Chr.1: 193138092-193138092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752785568 Chr.1: 193138135-193138135 on Build GRCh38
rs71639185 Chr.1: 193138110-193138110 on Build GRCh38
rs934976346 Chr.1: 193138093-193138093 on Build GRCh38
rs766638782 Chr.1: 193138075-193138075 on Build GRCh38
rs755468043 Chr.1: 193138099-193138099 on Build GRCh38
rs571687597 Chr.1: 193138048-193138048 on Build GRCh38
rs750008798 Chr.1: 193138068-193138068 on Build GRCh38
rs773796014 Chr.1: 193138046-193138046 on Build GRCh38
rs779144943 Chr.1: 193138111-193138111 on Build GRCh38
rs370666592 Chr.1: 193138078-193138078 on Build GRCh38
rs559275109 Chr.1: 193138126-193138126 on Build GRCh38
rs76717731 Chr.1: 193138062-193138062 on Build GRCh38
rs115964909 Chr.1: 193138057-193138057 on Build GRCh38
rs1052060325 Chr.1: 193138104-193138104 on Build GRCh38
rs201012067 Chr.1: 193138069-193138069 on Build GRCh38

View Full Product Details