Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 148868638-148868485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE4DIP
Entrez Gene ID
9659
Gene Name
phosphodiesterase 4D interacting protein
Gene Aliases
CMYA2, MMGL
UniGene
Hs.584841
Gene Chromosome Location
Chr.1: 148808167-149051277 on Build GRCh38
Transcripts
NM_001198832.2 NM_022359.5 XM_006711646.1 XM_011510172.1 XM_011510173.1 XM_011510175.1 XM_011510176.1 XM_017002878.1 XM_017002880.1 XM_017002887.1 XM_017002888.1 XM_017002889.1 XM_017002891.1 XM_017002892.1 XM_017002893.1 XM_017002894.1 XM_017002898.1 XM_017002899.1 XM_017002902.1 XM_017002903.1

View Full Product Details